Assay Details
| Gene Symbol: BLK | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | BC004473.2 | - | 1 - 2 | 424 | 80 | |
| BC038555.1 | - | 1 - 2 | 424 | 80 | ||
More Information
Set Membership: |
|
MicroArray Probes:
Affymetrix
ABI
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0800006746.hg.1 | ENST00000259089.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006746.hg.1 | ENST00000259089.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006746.hg.1 | ENST00000259089.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006746.hg.1 | ENST00000259089.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006746.hg.1 | ENST00000259089.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006746.hg.1 | ENST00000259089.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006746.hg.1 | ENST00000259089.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006746.hg.1 | ENST00000259089.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006746.hg.1 | ENST00000259089.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006746.hg.1 | ENST00000259089.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006746.hg.1 | ENST00000259089.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006746.hg.1 | ENST00000259089.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006746.hg.1 | ENST00000259089.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006746.hg.1 | ENST00000259089.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006746.hg.1 | ENST00000259089.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006746.hg.1 | ENST00000259089.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006746.hg.1 | ENST00000259089.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006746.hg.1 | ENST00000259089.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006746.hg.1 | ENST00000259089.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006746.hg.1 | ENST00000525389.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006746.hg.1 | ENST00000525389.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006746.hg.1 | ENST00000525389.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006746.hg.1 | ENST00000525389.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006746.hg.1 | ENST00000526097.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006746.hg.1 | ENST00000526097.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006746.hg.1 | ENST00000526097.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006746.hg.1 | ENST00000526097.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006746.hg.1 | ENST00000526097.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006746.hg.1 | ENST00000526097.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006746.hg.1 | ENST00000526778.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006746.hg.1 | ENST00000526778.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006746.hg.1 | ENST00000526778.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006746.hg.1 | ENST00000526778.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006746.hg.1 | ENST00000526778.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006746.hg.1 | ENST00000529894.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006746.hg.1 | ENST00000529894.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006746.hg.1 | ENST00000529894.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006746.hg.1 | ENST00000529894.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006746.hg.1 | ENST00000529894.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006746.hg.1 | ENST00000529894.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006746.hg.1 | ENST00000529894.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006746.hg.1 | ENST00000529894.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006746.hg.1 | ENST00000529894.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006746.hg.1 | ENST00000529894.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006746.hg.1 | ENST00000529894.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006746.hg.1 | ENST00000529894.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006746.hg.1 | ENST00000529894.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006746.hg.1 | ENST00000529894.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006746.hg.1 | ENST00000529894.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006746.hg.1 | ENST00000533828.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006746.hg.1 | ENST00000533828.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006746.hg.1 | ENST00000533828.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006746.hg.1 | ENST00000533828.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006746.hg.1 | NM_001715.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006746.hg.1 | NM_001715.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006746.hg.1 | NM_001715.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006746.hg.1 | NM_001715.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006746.hg.1 | NM_001715.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006746.hg.1 | NM_001715.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006746.hg.1 | NM_001715.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006746.hg.1 | NM_001715.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006746.hg.1 | NM_001715.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006746.hg.1 | NM_001715.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006746.hg.1 | NM_001715.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006746.hg.1 | NM_001715.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006746.hg.1 | NM_001715.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006746.hg.1 | NM_001715.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006746.hg.1 | NM_001715.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006746.hg.1 | NM_001715.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006746.hg.1 | NM_001715.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006746.hg.1 | NM_001715.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006746.hg.1 | NM_001715.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800006746.hg.1 | ENST00000259089.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800006746.hg.1 | ENST00000259089.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800006746.hg.1 | ENST00000259089.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800006746.hg.1 | ENST00000259089.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800006746.hg.1 | ENST00000259089.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800006746.hg.1 | ENST00000259089.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800006746.hg.1 | ENST00000259089.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800006746.hg.1 | ENST00000259089.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800006746.hg.1 | ENST00000259089.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800006746.hg.1 | ENST00000259089.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800006746.hg.1 | ENST00000259089.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800006746.hg.1 | ENST00000259089.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800006746.hg.1 | ENST00000259089.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800006746.hg.1 | ENST00000259089.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800006746.hg.1 | ENST00000259089.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800006746.hg.1 | ENST00000259089.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800006746.hg.1 | ENST00000259089.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800006746.hg.1 | ENST00000259089.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800006746.hg.1 | ENST00000259089.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800006746.hg.1 | ENST00000525389.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800006746.hg.1 | ENST00000525389.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800006746.hg.1 | ENST00000525389.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800006746.hg.1 | ENST00000525389.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800006746.hg.1 | ENST00000526097.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800006746.hg.1 | ENST00000526097.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800006746.hg.1 | ENST00000526097.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800006746.hg.1 | ENST00000526097.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800006746.hg.1 | ENST00000526097.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800006746.hg.1 | ENST00000526097.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800006746.hg.1 | ENST00000526778.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800006746.hg.1 | ENST00000526778.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800006746.hg.1 | ENST00000526778.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800006746.hg.1 | ENST00000526778.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800006746.hg.1 | ENST00000526778.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800006746.hg.1 | ENST00000529894.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800006746.hg.1 | ENST00000529894.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800006746.hg.1 | ENST00000529894.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800006746.hg.1 | ENST00000529894.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800006746.hg.1 | ENST00000529894.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800006746.hg.1 | ENST00000529894.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800006746.hg.1 | ENST00000529894.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800006746.hg.1 | ENST00000529894.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800006746.hg.1 | ENST00000529894.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800006746.hg.1 | ENST00000529894.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800006746.hg.1 | ENST00000529894.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800006746.hg.1 | ENST00000529894.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800006746.hg.1 | ENST00000529894.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800006746.hg.1 | ENST00000529894.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800006746.hg.1 | ENST00000529894.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800006746.hg.1 | ENST00000533828.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800006746.hg.1 | ENST00000533828.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800006746.hg.1 | ENST00000533828.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800006746.hg.1 | ENST00000533828.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800006746.hg.1 | NM_001715.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800006746.hg.1 | NM_001715.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800006746.hg.1 | NM_001715.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800006746.hg.1 | NM_001715.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800006746.hg.1 | NM_001715.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800006746.hg.1 | NM_001715.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800006746.hg.1 | NM_001715.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800006746.hg.1 | NM_001715.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800006746.hg.1 | NM_001715.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800006746.hg.1 | NM_001715.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800006746.hg.1 | NM_001715.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800006746.hg.1 | NM_001715.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800006746.hg.1 | NM_001715.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800006746.hg.1 | NM_001715.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800006746.hg.1 | NM_001715.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800006746.hg.1 | NM_001715.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800006746.hg.1 | NM_001715.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800006746.hg.1 | NM_001715.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800006746.hg.1 | NM_001715.2 | Refseq |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Human Genome Survey Microarray Version 2.0 | 224975 | - | - |

BLK
Genomic Map