Assay Details
Gene Symbol: ANKRD36BP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_026844.1 | 1 | 1199 | 126 | ||
GenBank mRNA | BC007072.1 | - | 1 | 1112 | 126 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC01003486.hg.1
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ArrayStar
ABI
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent SurePrint G3 Human CGH Microarray 4x180K | 127583 | AB023171.2 | genbank_mrna_human |
Agilent SurePrint G3 Human CGH Microarray 4x180K | 127583 | AF086762.1 | genbank_mrna_human |
Agilent SurePrint G3 Human CGH Microarray 4x180K | 127583 | BC004938.1 | genbank_mrna_human |
Agilent SurePrint G3 Human CGH Microarray 4x180K | 127583 | BC020196.1 | genbank_mrna_human |
Agilent SurePrint G3 Human CGH Microarray 4x180K | 127583 | NM_013279.1 | refseq_mrna_human_NM |
Agilent SurePrint G3 Human CGH Microarray 8x60K | 30751 | - | - |
Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M | 342153 | - | - |
Agilent Whole Human Genome Microarray 4x44K v2 | A_23_P97123 | - | - |
Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k | GT_44k_23_P97123 | - | - |
Agilent Whole Human Genome Oligo Microarray 4x44K | 22252 | - | - |
Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray | 22324 | - | - |
Agilent Whole Human Genome Microarray 4x44K v2 | A_23_P97123 | - | - |
SurePrint G3 Human GE Microarray G4851A | A_23_P97123 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20640162 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00580639 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20640183 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20640177 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00580644 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20640184 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00580615 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00580640 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20640172 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00580629 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20640173 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00580622 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00580617 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00580630 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20640168 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20640186 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00580625 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00580626 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00580647 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20640174 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P15141584 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20640187 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00580621 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20640157 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20640166 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P15141586 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P15141587 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00580618 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20640151 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20640179 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00580634 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00580651 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20640173 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20640162 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20640187 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20640177 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00580647 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00580626 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20640186 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00580639 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P15141584 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00580617 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P15141586 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20640151 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P15141587 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20640183 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00580622 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00580621 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00580634 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00580640 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20640168 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20640157 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00580618 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20640172 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20640184 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00580629 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00580630 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20640174 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00580615 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20640166 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20640179 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00580625 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00580644 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00580651 | - | - |
Whole Human Genome Oligo Microarray G4112A | A_23_P97123 | - | - |
Agilent Human Genome CGH Microarray 4x44K | A_14_P111113 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P00580649 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P00580634 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P00580625 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P00580630 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P00580564 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P00580619 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P00580627 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P00580651 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P15141579 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P00580645 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P00580561 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P00580637 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P00580639 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P15141585 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P00580648 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P00580642 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P00580632 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P00580559 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P00580635 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P00580614 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P15141589 | - | - |
Agilent SurePrint G3 Human CGH Microarray 2x400K | A_14_P111113 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Gene 1.0 ST Array | 7922123 | - | - |
Affymetrix Human Gene 1.0 ST Array | 7922122 | - | - |
Affymetrix Human Gene 1.0 ST Array | 7922124 | - | - |
Affymetrix Human Gene 1.1 ST Array | 7922121 | - | - |
Affymetrix Human Transcriptome Array 2.0 | TC0100016328.hg.1 | ENST00000358576.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100016328.hg.1 | ENST00000358576.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100016328.hg.1 | ENST00000358576.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100016328.hg.1 | ENST00000604892.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100016328.hg.1 | NR_026844.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100016328.hg.1 | NR_026844.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100016328.hg.1 | NR_026844.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100016328.hg.1 | NR_026844.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR01054243.hg.1 | TC01003486.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR01054243.hg.1 | ENST00000358576.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR01054243.hg.1 | NR_026844.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR01054244.hg.1 | TC01003486.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR01054244.hg.1 | NR_026844.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100224207.hg.1 | ENST00000358576.4 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100224207.hg.1 | NR_026844.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100224207.hg.1 | ENST00000358576.4 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100224207.hg.1 | NR_026844.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100224207.hg.1 | TC0100016328.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0100224206.hg.1 | ENST00000358576.4 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100224206.hg.1 | ENST00000604892.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100224206.hg.1 | NR_026844.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100224206.hg.1 | ENST00000358576.4 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100224206.hg.1 | ENST00000604892.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100224206.hg.1 | NR_026844.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100224206.hg.1 | TC0100016328.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0100224208.hg.1 | NR_026844.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100224208.hg.1 | NR_026844.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100224208.hg.1 | TC0100016328.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | TC0100016328.hg.1 | ENST00000358576.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100016328.hg.1 | ENST00000358576.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100016328.hg.1 | ENST00000358576.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100016328.hg.1 | ENST00000604892.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100016328.hg.1 | NR_026844.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100016328.hg.1 | NR_026844.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100016328.hg.1 | NR_026844.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100016328.hg.1 | NR_026844.1 | Refseq |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Arraystar Human LncRNA microarray V2.0 | ASHG19A3A054890 | - | - |
Arraystar Human LncRNA microarray V2.0 | ASHG19A3A054890 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Human Genome Survey Microarray Version 2.0 | 103642 | - | - |
