| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100009926.hg.1 |
ENST00000328813.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100009926.hg.1 |
ENST00000624064.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100009926.hg.1 |
ENST00000624064.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100009926.hg.1 |
ENST00000624064.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100009926.hg.1 |
ENST00000624187.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100009926.hg.1 |
NM_033180.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100009926.hg.1 |
NM_033180.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100009926.hg.1 |
NM_033180.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR11016834.hg.1 |
ENST00000328813.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR11016834.hg.1 |
NM_033180.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR11016834.hg.1 |
TC11001335.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR11016835.hg.1 |
ENST00000328813.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR11016835.hg.1 |
NM_033180.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR11016835.hg.1 |
TC11001335.hg.1 |
HTA TC |
| Affymetrix Human Clariom D Assay |
PSR1100174965.hg.1 |
ENST00000624064.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1100174965.hg.1 |
NM_033180.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1100174965.hg.1 |
ENST00000624064.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1100174965.hg.1 |
NM_033180.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1100174965.hg.1 |
TC1100009926.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR1100174964.hg.1 |
ENST00000328813.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1100174964.hg.1 |
ENST00000624064.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1100174964.hg.1 |
ENST00000624187.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1100174964.hg.1 |
NM_033180.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1100174964.hg.1 |
ENST00000328813.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1100174964.hg.1 |
ENST00000624064.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1100174964.hg.1 |
ENST00000624187.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1100174964.hg.1 |
NM_033180.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1100174964.hg.1 |
TC1100009926.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
TC1100009926.hg.1 |
ENST00000328813.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100009926.hg.1 |
ENST00000624064.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100009926.hg.1 |
ENST00000624064.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100009926.hg.1 |
ENST00000624064.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100009926.hg.1 |
ENST00000624187.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100009926.hg.1 |
NM_033180.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1100009926.hg.1 |
NM_033180.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1100009926.hg.1 |
NM_033180.4 |
Refseq |