Assay Details
| Gene Symbol: OPALIN | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_033207.4 | NP_149984.1 | 2 - 3 | 511 | 97 | |
| GenBank mRNA | AF367761.2 | - | 2 - 3 | 451 | 97 | |
| AK289835.1 | - | 2 - 3 | 155 | 97 | ||
| AL834229.1 | - | 2 - 3 | 128 | 97 | ||
| BC033737.1 | - | 2 - 3 | 128 | 5177902 | 97 | |
More Information
Updated Mapping Information
Additional Information:
This assay is in the 5'UTR region of the AF367761.2,AK289835.1,AL834229.1,BC033737.1,NM_033207.4 transcripts.
Set Membership: |
|

OPALIN
Genomic Map