Assay Details
Gene Symbol: SLC8A3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_033262.4 | NP_150287.1 | 3 - 4 | 2649 | 64 | |
GenBank mRNA | AJ304852.1 | - | 2 - 3 | 1957 | 64 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
