Assay Details
Gene Symbol: PRRG1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AF009242.1 | - | 1 - 2 | 34 | 123 |

More Information
Additional Information:
This assay is in the 5'UTR region of the AF009242.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
