Assay Details
| Gene Symbol: SNRPB2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_003092.4 | NP_003083.1 | 1 - 2 | 196 | 82 | |
| GenBank mRNA | AK313004.1 | - | 1 - 2 | 130 | 82 | |
| BC018022.1 | - | 1 - 2 | 148 | 4286926 | 82 | |
| BF243328.1 | - | 175 | 83 | |||
| M15841.1 | - | 1 - 2 | 85 | 82 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC20000104.hg.1
Additional Information:
This assay is in the 5'UTR region of the AK313004.1,BC018022.1,M15841.1,NM_003092.4 transcripts.
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
ABI
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0500010814.hg.1 | ENST00000429057.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR20001396.hg.1 | ENST00000246071.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR20001396.hg.1 | ENST00000377943.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR20001396.hg.1 | NM_003092.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR20001396.hg.1 | NM_198220.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR20001396.hg.1 | TC20000104.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013510.hg.1 | ENST00000407763.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR20001400.hg.1 | ENST00000246071.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR20001400.hg.1 | ENST00000478522.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR20001400.hg.1 | NM_003092.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR20001400.hg.1 | TC20000104.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR20001401.hg.1 | ENST00000246071.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR20001401.hg.1 | ENST00000377943.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR20001401.hg.1 | ENST00000478522.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR20001401.hg.1 | NM_003092.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR20001401.hg.1 | NM_198220.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR20001401.hg.1 | TC20000104.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR20001402.hg.1 | ENST00000246071.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR20001402.hg.1 | ENST00000377943.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR20001402.hg.1 | ENST00000478522.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR20001402.hg.1 | NM_003092.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR20001402.hg.1 | NM_198220.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR20001402.hg.1 | TC20000104.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC2000006781.hg.1 | ENST00000246071.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000006781.hg.1 | ENST00000246071.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000006781.hg.1 | ENST00000246071.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000006781.hg.1 | ENST00000246071.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000006781.hg.1 | ENST00000246071.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000006781.hg.1 | ENST00000246071.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000006781.hg.1 | ENST00000246071.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000006781.hg.1 | ENST00000246071.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000006781.hg.1 | ENST00000246071.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000006781.hg.1 | ENST00000246071.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000006781.hg.1 | ENST00000246071.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000006781.hg.1 | ENST00000246071.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000006781.hg.1 | ENST00000246071.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000006781.hg.1 | ENST00000246071.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000006781.hg.1 | ENST00000377943.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000006781.hg.1 | ENST00000377943.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000006781.hg.1 | ENST00000377943.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000006781.hg.1 | ENST00000377943.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000006781.hg.1 | ENST00000377943.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000006781.hg.1 | ENST00000377943.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000006781.hg.1 | ENST00000377943.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000006781.hg.1 | ENST00000377943.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000006781.hg.1 | ENST00000377943.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000006781.hg.1 | ENST00000377943.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000006781.hg.1 | ENST00000377943.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000006781.hg.1 | ENST00000377943.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000006781.hg.1 | ENST00000478522.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000006781.hg.1 | ENST00000478522.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000006781.hg.1 | ENST00000478522.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000006781.hg.1 | ENST00000478522.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000006781.hg.1 | ENST00000478522.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000006781.hg.1 | ENST00000478522.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000006781.hg.1 | NM_003092.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000006781.hg.1 | NM_003092.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000006781.hg.1 | NM_003092.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000006781.hg.1 | NM_003092.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000006781.hg.1 | NM_003092.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000006781.hg.1 | NM_003092.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000006781.hg.1 | NM_003092.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000006781.hg.1 | NM_003092.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000006781.hg.1 | NM_003092.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000006781.hg.1 | NM_003092.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000006781.hg.1 | NM_003092.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000006781.hg.1 | NM_003092.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000006781.hg.1 | NM_003092.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000006781.hg.1 | NM_198220.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000006781.hg.1 | NM_198220.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000006781.hg.1 | NM_198220.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000006781.hg.1 | NM_198220.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000006781.hg.1 | NM_198220.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000006781.hg.1 | NM_198220.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000006781.hg.1 | NM_198220.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000006781.hg.1 | NM_198220.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000006781.hg.1 | NM_198220.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000006781.hg.1 | NM_198220.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000006781.hg.1 | NM_198220.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000006781.hg.1 | NM_198220.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR20001399.hg.1 | ENST00000246071.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR20001399.hg.1 | ENST00000377943.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR20001399.hg.1 | ENST00000478522.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR20001399.hg.1 | NM_003092.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR20001399.hg.1 | NM_198220.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR20001399.hg.1 | TC20000104.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | PSR2000146432.hg.1 | ENST00000246071.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000146432.hg.1 | ENST00000377943.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000146432.hg.1 | ENST00000478522.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000146432.hg.1 | NM_003092.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2000146432.hg.1 | NM_198220.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2000146432.hg.1 | TC2000006781.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR2000146432.hg.1 | ENST00000246071.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000146432.hg.1 | ENST00000377943.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000146432.hg.1 | ENST00000478522.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000146432.hg.1 | NM_003092.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2000146432.hg.1 | NM_198220.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2000146439.hg.1 | ENST00000246071.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000146439.hg.1 | ENST00000377943.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000146439.hg.1 | ENST00000478522.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000146439.hg.1 | NM_003092.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2000146439.hg.1 | NM_198220.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2000146439.hg.1 | TC2000006781.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR2000146439.hg.1 | ENST00000246071.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000146439.hg.1 | ENST00000377943.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000146439.hg.1 | ENST00000478522.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000146439.hg.1 | NM_003092.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2000146439.hg.1 | NM_198220.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000006781.hg.1 | ENST00000246071.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000006781.hg.1 | ENST00000246071.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000006781.hg.1 | ENST00000246071.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000006781.hg.1 | ENST00000246071.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000006781.hg.1 | ENST00000246071.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000006781.hg.1 | ENST00000246071.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000006781.hg.1 | ENST00000246071.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000006781.hg.1 | ENST00000246071.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000006781.hg.1 | ENST00000246071.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000006781.hg.1 | ENST00000246071.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000006781.hg.1 | ENST00000246071.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000006781.hg.1 | ENST00000246071.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000006781.hg.1 | ENST00000246071.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000006781.hg.1 | ENST00000246071.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000006781.hg.1 | ENST00000377943.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000006781.hg.1 | ENST00000377943.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000006781.hg.1 | ENST00000377943.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000006781.hg.1 | ENST00000377943.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000006781.hg.1 | ENST00000377943.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000006781.hg.1 | ENST00000377943.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000006781.hg.1 | ENST00000377943.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000006781.hg.1 | ENST00000377943.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000006781.hg.1 | ENST00000377943.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000006781.hg.1 | ENST00000377943.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000006781.hg.1 | ENST00000377943.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000006781.hg.1 | ENST00000377943.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000006781.hg.1 | ENST00000478522.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000006781.hg.1 | ENST00000478522.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000006781.hg.1 | ENST00000478522.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000006781.hg.1 | ENST00000478522.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000006781.hg.1 | ENST00000478522.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000006781.hg.1 | ENST00000478522.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000006781.hg.1 | NM_003092.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000006781.hg.1 | NM_003092.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000006781.hg.1 | NM_003092.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000006781.hg.1 | NM_003092.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000006781.hg.1 | NM_003092.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000006781.hg.1 | NM_003092.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000006781.hg.1 | NM_003092.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000006781.hg.1 | NM_003092.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000006781.hg.1 | NM_003092.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000006781.hg.1 | NM_003092.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000006781.hg.1 | NM_003092.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000006781.hg.1 | NM_003092.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000006781.hg.1 | NM_003092.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000006781.hg.1 | NM_198220.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000006781.hg.1 | NM_198220.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000006781.hg.1 | NM_198220.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000006781.hg.1 | NM_198220.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000006781.hg.1 | NM_198220.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000006781.hg.1 | NM_198220.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000006781.hg.1 | NM_198220.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000006781.hg.1 | NM_198220.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000006781.hg.1 | NM_198220.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000006781.hg.1 | NM_198220.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000006781.hg.1 | NM_198220.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000006781.hg.1 | NM_198220.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2000146434.hg.1 | ENST00000246071.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000146434.hg.1 | ENST00000478522.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000146434.hg.1 | NM_003092.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2000146434.hg.1 | TC2000006781.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR2000146434.hg.1 | ENST00000246071.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000146434.hg.1 | ENST00000478522.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000146434.hg.1 | NM_003092.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2000146436.hg.1 | ENST00000246071.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000146436.hg.1 | ENST00000377943.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000146436.hg.1 | ENST00000478522.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000146436.hg.1 | NM_003092.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2000146436.hg.1 | NM_198220.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2000146436.hg.1 | TC2000006781.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR2000146436.hg.1 | ENST00000246071.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000146436.hg.1 | ENST00000377943.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000146436.hg.1 | ENST00000478522.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000146436.hg.1 | NM_003092.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2000146436.hg.1 | NM_198220.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500010814.hg.1 | ENST00000429057.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000146429.hg.1 | ENST00000246071.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000146429.hg.1 | ENST00000377943.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000146429.hg.1 | NM_003092.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2000146429.hg.1 | NM_198220.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2000146429.hg.1 | TC2000006781.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR2000146429.hg.1 | ENST00000246071.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000146429.hg.1 | ENST00000377943.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000146429.hg.1 | NM_003092.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2000146429.hg.1 | NM_198220.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2000146437.hg.1 | ENST00000246071.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000146437.hg.1 | ENST00000377943.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000146437.hg.1 | ENST00000478522.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000146437.hg.1 | NM_003092.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2000146437.hg.1 | NM_198220.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2000146437.hg.1 | TC2000006781.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR2000146437.hg.1 | ENST00000246071.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000146437.hg.1 | ENST00000377943.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000146437.hg.1 | ENST00000478522.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000146437.hg.1 | NM_003092.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2000146437.hg.1 | NM_198220.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600013510.hg.1 | ENST00000407763.2 | Ensembl |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Human Genome Survey Microarray Version 2.0 | 139206 | - | - |

SNRPB2
Genomic Map