Assay Details
| Gene Symbol: HNF4G | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | Z49826.2 | - | 1 - 2 | 129 | 59 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the Z49826.2 transcripts.
Set Membership: |
|
Panther Classification:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0800008002.hg.1 | ENST00000354370.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008002.hg.1 | ENST00000354370.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008002.hg.1 | ENST00000354370.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008002.hg.1 | ENST00000354370.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008002.hg.1 | ENST00000354370.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008002.hg.1 | ENST00000354370.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008002.hg.1 | ENST00000354370.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008002.hg.1 | ENST00000354370.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008002.hg.1 | ENST00000354370.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008002.hg.1 | ENST00000354370.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008002.hg.1 | ENST00000354370.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008002.hg.1 | ENST00000354370.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008002.hg.1 | ENST00000354370.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008002.hg.1 | ENST00000354370.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008002.hg.1 | ENST00000396419.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008002.hg.1 | ENST00000396419.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008002.hg.1 | ENST00000396419.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008002.hg.1 | ENST00000396419.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008002.hg.1 | ENST00000396423.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008002.hg.1 | ENST00000396423.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008002.hg.1 | ENST00000396423.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008002.hg.1 | ENST00000396423.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008002.hg.1 | ENST00000396423.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008002.hg.1 | ENST00000396423.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008002.hg.1 | ENST00000396423.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008002.hg.1 | ENST00000396423.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008002.hg.1 | ENST00000396423.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008002.hg.1 | ENST00000396423.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008002.hg.1 | ENST00000396423.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008002.hg.1 | ENST00000396423.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008002.hg.1 | ENST00000396423.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008002.hg.1 | ENST00000396423.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008002.hg.1 | ENST00000396423.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008002.hg.1 | ENST00000396423.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008002.hg.1 | ENST00000494318.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008002.hg.1 | ENST00000494318.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008002.hg.1 | ENST00000494318.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008002.hg.1 | ENST00000494318.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008002.hg.1 | ENST00000494318.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008002.hg.1 | NM_004133.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008002.hg.1 | NM_004133.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008002.hg.1 | NM_004133.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008002.hg.1 | NM_004133.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008002.hg.1 | NM_004133.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008002.hg.1 | NM_004133.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008002.hg.1 | NM_004133.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008002.hg.1 | NM_004133.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008002.hg.1 | NM_004133.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008002.hg.1 | NM_004133.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008002.hg.1 | NM_004133.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008002.hg.1 | NM_004133.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008002.hg.1 | NM_004133.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008002.hg.1 | NM_004133.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008002.hg.1 | NM_004133.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800008002.hg.1 | ENST00000354370.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800008002.hg.1 | ENST00000354370.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800008002.hg.1 | ENST00000354370.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800008002.hg.1 | ENST00000354370.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800008002.hg.1 | ENST00000354370.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800008002.hg.1 | ENST00000354370.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800008002.hg.1 | ENST00000354370.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800008002.hg.1 | ENST00000354370.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800008002.hg.1 | ENST00000354370.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800008002.hg.1 | ENST00000354370.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800008002.hg.1 | ENST00000354370.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800008002.hg.1 | ENST00000354370.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800008002.hg.1 | ENST00000354370.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800008002.hg.1 | ENST00000354370.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800008002.hg.1 | ENST00000396419.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800008002.hg.1 | ENST00000396419.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800008002.hg.1 | ENST00000396419.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800008002.hg.1 | ENST00000396419.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800008002.hg.1 | ENST00000396423.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800008002.hg.1 | ENST00000396423.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800008002.hg.1 | ENST00000396423.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800008002.hg.1 | ENST00000396423.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800008002.hg.1 | ENST00000396423.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800008002.hg.1 | ENST00000396423.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800008002.hg.1 | ENST00000396423.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800008002.hg.1 | ENST00000396423.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800008002.hg.1 | ENST00000396423.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800008002.hg.1 | ENST00000396423.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800008002.hg.1 | ENST00000396423.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800008002.hg.1 | ENST00000396423.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800008002.hg.1 | ENST00000396423.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800008002.hg.1 | ENST00000396423.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800008002.hg.1 | ENST00000396423.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800008002.hg.1 | ENST00000396423.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800008002.hg.1 | ENST00000494318.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800008002.hg.1 | ENST00000494318.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800008002.hg.1 | ENST00000494318.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800008002.hg.1 | ENST00000494318.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800008002.hg.1 | ENST00000494318.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800008002.hg.1 | NM_004133.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800008002.hg.1 | NM_004133.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800008002.hg.1 | NM_004133.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800008002.hg.1 | NM_004133.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800008002.hg.1 | NM_004133.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800008002.hg.1 | NM_004133.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800008002.hg.1 | NM_004133.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800008002.hg.1 | NM_004133.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800008002.hg.1 | NM_004133.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800008002.hg.1 | NM_004133.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800008002.hg.1 | NM_004133.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800008002.hg.1 | NM_004133.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800008002.hg.1 | NM_004133.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800008002.hg.1 | NM_004133.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800008002.hg.1 | NM_004133.4 | Refseq |

HNF4G
Genomic Map