Assay Details
| Gene Symbol: VDAC3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001135694.2 | NP_001129166.1 | 1 - 2 | 168 | 131 | |
| NM_005662.6 | NP_005653.3 | 1 - 2 | 168 | 131 | ||
| XM_006716394.1 | XP_006716457.1 | 1 - 2 | 305 | 131 | ||
| GenBank mRNA | AF038962.1 | - | 1 - 2 | 57 | 130 | |
| BX643099.1 | - | 1 - 2 | 101 | 131 | ||
| DA930673.1 | - | 1 - 2 | 168 | 131 | ||
| DB484671.1 | - | 1 - 2 | 90 | 131 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AF038962.1, NM_001135694.2, NM_005662.6, XM_006716394.1 transcripts.
Set Membership: |
|

VDAC3
Genomic Map