| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200010842.hg.1 |
ENST00000257905.12 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200010842.hg.1 |
ENST00000257905.12 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200010842.hg.1 |
ENST00000257905.12 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200010842.hg.1 |
ENST00000257905.12 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200010842.hg.1 |
ENST00000257905.12 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200010842.hg.1 |
ENST00000257905.12 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200010842.hg.1 |
ENST00000257905.12 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200010842.hg.1 |
ENST00000257905.12 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200010842.hg.1 |
ENST00000257905.12 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200010842.hg.1 |
ENST00000257905.12 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200010842.hg.1 |
ENST00000257905.12 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200010842.hg.1 |
ENST00000547431.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200010842.hg.1 |
ENST00000547431.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200010842.hg.1 |
ENST00000547431.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200010842.hg.1 |
ENST00000547431.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200010842.hg.1 |
ENST00000547431.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200010842.hg.1 |
ENST00000547431.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200010842.hg.1 |
ENST00000547826.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200010842.hg.1 |
ENST00000547826.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200010842.hg.1 |
ENST00000547826.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200010842.hg.1 |
ENST00000547826.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200010842.hg.1 |
ENST00000547826.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200010842.hg.1 |
ENST00000547826.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200010842.hg.1 |
ENST00000547826.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200010842.hg.1 |
ENST00000553113.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200010842.hg.1 |
ENST00000553113.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200010842.hg.1 |
ENST00000553113.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200010842.hg.1 |
ENST00000553113.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200010842.hg.1 |
ENST00000553113.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200010842.hg.1 |
ENST00000553113.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200010842.hg.1 |
NM_006741.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200010842.hg.1 |
NM_006741.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200010842.hg.1 |
NM_006741.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200010842.hg.1 |
NM_006741.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200010842.hg.1 |
NM_006741.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200010842.hg.1 |
NM_006741.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200010842.hg.1 |
NM_006741.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200010842.hg.1 |
NM_006741.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200010842.hg.1 |
NM_006741.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200010842.hg.1 |
NM_006741.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200010842.hg.1 |
NM_006741.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR12020873.hg.1 |
ENST00000257905.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR12020873.hg.1 |
ENST00000379690.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR12020873.hg.1 |
ENST00000547431.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR12020873.hg.1 |
NM_006741.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR12020873.hg.1 |
TC12001574.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR12020875.hg.1 |
ENST00000257905.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR12020875.hg.1 |
ENST00000379690.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR12020875.hg.1 |
NM_006741.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR12020875.hg.1 |
TC12001574.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100011668.hg.1 |
ENST00000525622.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR12020874.hg.1 |
ENST00000257905.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR12020874.hg.1 |
ENST00000379690.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR12020874.hg.1 |
ENST00000547431.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR12020874.hg.1 |
NM_006741.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR12020874.hg.1 |
TC12001574.hg.1 |
HTA TC |
| Affymetrix Human Clariom D Assay |
PSR1200183348.hg.1 |
ENST00000257905.12 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1200183348.hg.1 |
ENST00000547431.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1200183348.hg.1 |
ENST00000547826.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1200183348.hg.1 |
NM_006741.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1200183348.hg.1 |
ENST00000257905.12 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1200183348.hg.1 |
ENST00000547431.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1200183348.hg.1 |
ENST00000547826.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1200183348.hg.1 |
NM_006741.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1200183348.hg.1 |
TC1200010842.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR1200183345.hg.1 |
ENST00000257905.12 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1200183345.hg.1 |
ENST00000547431.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1200183345.hg.1 |
ENST00000547826.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1200183345.hg.1 |
ENST00000553113.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1200183345.hg.1 |
NM_006741.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1200183345.hg.1 |
ENST00000257905.12 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1200183345.hg.1 |
ENST00000547431.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1200183345.hg.1 |
ENST00000547826.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1200183345.hg.1 |
ENST00000553113.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1200183345.hg.1 |
NM_006741.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1200183345.hg.1 |
TC1200010842.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
TC1200010842.hg.1 |
ENST00000257905.12 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200010842.hg.1 |
ENST00000257905.12 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200010842.hg.1 |
ENST00000257905.12 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200010842.hg.1 |
ENST00000257905.12 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200010842.hg.1 |
ENST00000257905.12 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200010842.hg.1 |
ENST00000257905.12 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200010842.hg.1 |
ENST00000257905.12 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200010842.hg.1 |
ENST00000257905.12 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200010842.hg.1 |
ENST00000257905.12 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200010842.hg.1 |
ENST00000257905.12 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200010842.hg.1 |
ENST00000257905.12 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200010842.hg.1 |
ENST00000547431.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200010842.hg.1 |
ENST00000547431.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200010842.hg.1 |
ENST00000547431.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200010842.hg.1 |
ENST00000547431.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200010842.hg.1 |
ENST00000547431.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200010842.hg.1 |
ENST00000547431.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200010842.hg.1 |
ENST00000547826.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200010842.hg.1 |
ENST00000547826.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200010842.hg.1 |
ENST00000547826.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200010842.hg.1 |
ENST00000547826.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200010842.hg.1 |
ENST00000547826.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200010842.hg.1 |
ENST00000547826.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200010842.hg.1 |
ENST00000547826.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200010842.hg.1 |
ENST00000553113.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200010842.hg.1 |
ENST00000553113.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200010842.hg.1 |
ENST00000553113.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200010842.hg.1 |
ENST00000553113.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200010842.hg.1 |
ENST00000553113.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200010842.hg.1 |
ENST00000553113.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200010842.hg.1 |
NM_006741.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1200010842.hg.1 |
NM_006741.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1200010842.hg.1 |
NM_006741.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1200010842.hg.1 |
NM_006741.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1200010842.hg.1 |
NM_006741.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1200010842.hg.1 |
NM_006741.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1200010842.hg.1 |
NM_006741.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1200010842.hg.1 |
NM_006741.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1200010842.hg.1 |
NM_006741.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1200010842.hg.1 |
NM_006741.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1200010842.hg.1 |
NM_006741.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1100011668.hg.1 |
ENST00000525622.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1200183347.hg.1 |
ENST00000257905.12 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1200183347.hg.1 |
ENST00000547431.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1200183347.hg.1 |
ENST00000547826.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1200183347.hg.1 |
NM_006741.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1200183347.hg.1 |
ENST00000257905.12 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1200183347.hg.1 |
ENST00000547431.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1200183347.hg.1 |
ENST00000547826.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1200183347.hg.1 |
NM_006741.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1200183347.hg.1 |
TC1200010842.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR1200183350.hg.1 |
ENST00000257905.12 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1200183350.hg.1 |
NM_006741.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1200183350.hg.1 |
ENST00000257905.12 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1200183350.hg.1 |
NM_006741.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1200183350.hg.1 |
TC1200010842.hg.1 |
Clariom D TC |