Assay Details
Gene Symbol: TREX2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_080701.3 | NP_542432.2 | 2 | 896 | 69 | |
GenBank mRNA | AF151107.1 | - | 1 | 352 | 69 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ArrayStar
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Human Whole Genome ChIP Tiling Array | A_17_P12033401 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P12033398 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P32770651 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P12033422 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P12033395 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P32770641 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P32770688 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P32770697 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P12033407 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P12033418 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P12033419 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P12033409 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P32770656 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P12033426 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P12033396 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P12033427 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P12033400 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P12033410 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P12033420 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P12033394 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P32770662 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P12033385 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P12033392 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P12033429 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P12033425 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P12033424 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P32770665 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P12033405 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P12033382 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P32770663 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P12033373 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P32770650 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P32770682 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P12033406 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P12033379 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P32770647 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P32770676 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P12033415 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P12033376 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P12033375 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P12033404 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P12033411 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P12033397 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P12033383 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P32770695 | - | - |
Agilent Whole Human Genome Microarray 4x44K v2 | A_33_P3383856 | - | - |
Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k | GT_44k_23_P219223 | - | - |
Agilent Whole Human Genome Oligo Microarray 4x44K | 27490 | - | - |
Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray | 12354 | - | - |
Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray | 43338 | - | - |
Agilent Whole Human Genome Microarray 4x44K v2 | A_33_P3383856 | - | - |
SurePrint G3 Human GE Microarray G4851A | A_33_P3383856 | - | - |
SurePrint G3 Human GE Microarray G4851A | ERCC-00120_69 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P12033422 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P12033405 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P12033419 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P12033407 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P12033429 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P12033404 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32770663 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P12033406 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P12033427 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P12033400 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32770682 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32770662 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P12033418 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32770697 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32770676 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32770665 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P12033409 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32770695 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P12033401 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P12033403 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P12033410 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P12033424 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P12033415 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32770688 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P12033411 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P12033422 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32770682 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P12033404 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P12033411 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32770665 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P12033407 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32770695 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P12033409 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P12033427 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P12033406 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32770662 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P12033415 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P12033403 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P12033400 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32770676 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P12033410 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P12033429 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P12033405 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P12033401 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P12033419 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32770697 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32770688 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32770663 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P12033424 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P12033418 | - | - |
Whole Human Genome Oligo Microarray G4112A | A_23_P219223 | - | - |
Agilent Human CpG Island ChIP-on-Chip Microarray 244K (G4492A) | A_17_P12033407 | - | - |
Agilent Human CpG Island ChIP-on-Chip Microarray 244K (G4492A) | A_17_P12033218 | - | - |
Agilent Human CpG Island ChIP-on-Chip Microarray 244K (G4492A) | A_17_P17414040 | - | - |
Agilent Human CpG Island ChIP-on-Chip Microarray 244K (G4492A) | A_17_P12033217 | - | - |
Agilent Human CpG Island ChIP-on-Chip Microarray 244K (G4492A) | A_17_P17414045 | - | - |
Agilent Human CpG Island ChIP-on-Chip Microarray 244K (G4492A) | A_17_P17414102 | - | - |
Agilent Human CpG Island ChIP-on-Chip Microarray 244K (G4492A) | A_17_P12033405 | - | - |
Agilent Human CpG Island ChIP-on-Chip Microarray 244K (G4492A) | A_17_P17414044 | - | - |
Agilent Human CpG Island ChIP-on-Chip Microarray 244K (G4492A) | A_17_P17414046 | - | - |
Agilent Human CpG Island ChIP-on-Chip Microarray 244K (G4492A) | A_17_P12033406 | - | - |
Agilent Human CpG Island ChIP-on-Chip Microarray 244K (G4492A) | A_17_P17414047 | - | - |
Agilent Human CpG Island ChIP-on-Chip Microarray 244K (G4492A) | A_17_P17414041 | - | - |
Agilent Human CpG Island ChIP-on-Chip Microarray 244K (G4492A) | A_17_P17414104 | - | - |
Agilent Human CpG Island ChIP-on-Chip Microarray 244K (G4492A) | A_17_P17414043 | - | - |
Agilent Human CpG Island ChIP-on-Chip Microarray 244K (G4492A) | A_17_P12033408 | - | - |
Agilent Human CpG Island ChIP-on-Chip Microarray 244K (G4492A) | A_17_P17414042 | - | - |
Agilent Human CpG Island ChIP-on-Chip Microarray 244K (G4492A) | A_17_P17414101 | - | - |
Agilent Human CpG Island ChIP-on-Chip Microarray 244K (G4492A) | A_17_P17414103 | - | - |
Agilent Human CpG Island ChIP-on-Chip Microarray 244K (G4492A) | A_17_P17414105 | - | - |
Agilent Whole Human Genome Microarray 4x44K G4112F | A_23_P219223 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P12033388 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P12033398 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P12033419 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P12033430 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P12033374 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P12033403 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P12033417 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P12033379 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P12033391 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P12033413 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P12033424 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P12033416 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P12033409 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P12033376 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P12033385 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P12033393 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P12033392 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P12033407 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P12033390 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P12033405 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P12033382 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P17414097 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P12033396 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P12033389 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P12033406 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P12033394 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P12033380 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P12033395 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P12033400 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P12033421 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P12033429 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Gene 1.0 ST Array | NM_080701_at | - | - |
Affymetrix Human Gene 1.0 ST Array | 8175810 | - | - |
Affymetrix Human Gene 1.0 ST Array | 8175784 | - | - |
Affymetrix Human Gene 1.0 ST Array | 8175806 | - | - |
Affymetrix Human Gene 1.0 ST Array | 8175804 | - | - |
Affymetrix Human Gene 1.0 ST Array | 8175805 | - | - |
Affymetrix Human Gene 1.0 ST Array | 8175783 | - | - |
Affymetrix Human Gene 1.0 ST Array | 8175809 | - | - |
Affymetrix Human Gene 1.0 ST Array | 8175803 | - | - |
Affymetrix Human Gene 1.0 ST Array | 8175807 | - | - |
Affymetrix Human Gene 1.1 ST Array | 8175782 | - | - |
Affymetrix Human Gene 1.1 ST Array | 8175802 | - | - |
Affymetrix Human Gene 1.1 ST Array | 8175808 | - | - |
Affymetrix HT HG-U133+ PM Array Plate | 211788_PM_s_at | - | - |
Affymetrix Human Genome U219 Array | 11740897_a_at | - | - |
Affymetrix Human Genome U219 Array | 11740897_PM_a_at | - | - |
Human Genome U133 Plus 2.0 Array | 211788_PM_s_at | - | - |
Human Genome U133 Plus 2.0 Array | 213334_PM_x_at | - | - |
Human Genome U133 Plus 2.0 Array | 207891_s_at | - | - |
Human Genome U133 Plus 2.0 Array | 211788_s_at | - | - |
Human Genome U133 Plus 2.0 Array | 213334_x_at | - | - |
Human Genome U133 Plus 2.0 Array | 207891_PM_s_at | - | - |
Human Genome U133A 2.0 Array | 213334_x_at | - | - |
Human Genome U133A 2.0 Array | 207891_s_at | - | - |
Human Genome U133A 2.0 Array | 213334_PM_x_at | - | - |
Human Genome U133A 2.0 Array | 207891_PM_s_at | - | - |
Human Genome U133A 2.0 Array | 211788_s_at | - | - |
Human Genome U133A 2.0 Array | 211788_PM_s_at | - | - |
Affymetrix GeneChip Human Genome U133 Plus 2.0 Array based on a custom CDF (REFSEQ_7) | NM_080701_at | - | - |
AffymetrixPathwork Diagnostics Pathchip | pw577042 | - | - |
Human Genome U95 Set | 41838_at | - | - |
Human Genome U95 Set | 41838_PM_at | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 211788_s_at10 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 211788_s_at6 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 211788_s_at7 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 211788_s_at4 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 211788_s_at9 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 211788_s_at3 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 211788_s_at2 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 211788_s_at11 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 211788_s_at1 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 211788_s_at5 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 211788_s_at8 | - | - |
Human Genome U95 Set | 91995_at | - | - |
Human Genome U95 Set | 91995_PM_at | - | - |
Human Genome U133 Set | 213334_x_at | - | - |
Human Genome U133 Set | 211788_PM_s_at | - | - |
Human Genome U133 Set | 207891_PM_s_at | - | - |
Human Genome U133 Set | 207891_s_at | - | - |
Human Genome U133 Set | 213334_PM_x_at | - | - |
Human Genome U133 Set | 211788_s_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Arraystar Human LncRNA microarray V2.0 | ASHG19A3A040553 | - | - |
Arraystar Human LncRNA microarray V2.0 | ASHG19A3A040553 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Human Genome Survey Microarray Version 2.0 | 181176 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
HumanHT-12 v4 Expression BeadChip | ILMN_1680892 | NM_080701.3 | human_transcriptome |
HumanHT-12 v4 Expression BeadChip | ILMN_1680892 | AF151107.1 | human_transcriptome |
Illumina HumanWG-6 v2.0 expression beadchip | ILMN_1680892 | NM_080701.3 | human_transcriptome |
Illumina HumanWG-6 v2.0 expression beadchip | ILMN_1680892 | AF151107.1 | human_transcriptome |
HumanHT-12 V4.0 expression beadchip | ILMN_1680892 | NM_080701.3 | human_transcriptome |
HumanHT-12 V4.0 expression beadchip | ILMN_1680892 | AF151107.1 | human_transcriptome |
Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_1680892 | NM_080701.3 | human_transcriptome |
Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_1680892 | AF151107.1 | human_transcriptome |
Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip | ILMN_1680892 | NM_080701.3 | human_transcriptome |
Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip | ILMN_1680892 | AF151107.1 | human_transcriptome |
Illumina HumanRef-8 v2 Expression BeadChip | GI_10384-S | - | - |
Illumina human-6 v2.0 expression beadchip | ILMN_1680892 | NM_080701.3 | human_transcriptome |
Illumina human-6 v2.0 expression beadchip | ILMN_1680892 | AF151107.1 | human_transcriptome |
Illumina humanRef-8 v2.0 expression beadchip | ILMN_1680892 | NM_080701.3 | human_transcriptome |
Illumina humanRef-8 v2.0 expression beadchip | ILMN_1680892 | AF151107.1 | human_transcriptome |
Illumina Sentrix Human-6 v2 Expression BeadChip | 5720377 | - | - |
Illumina HumanRef-6 v2.0 expression beadchip | ILMN_10384 | - | - |
Illumina HumanRef-8 v3.0 expression beadchip | ILMN_1680892 | NM_080701.3 | human_transcriptome |
Illumina HumanRef-8 v3.0 expression beadchip | ILMN_1680892 | AF151107.1 | human_transcriptome |
Illumina HumanWG-6 v3.0 expression beadchip | ILMN_1680892 | NM_080701.3 | human_transcriptome |
Illumina HumanWG-6 v3.0 expression beadchip | ILMN_1680892 | AF151107.1 | human_transcriptome |
Illumina HumanHT-12 V3.0 expression beadchip | ILMN_1680892 | NM_080701.3 | human_transcriptome |
Illumina HumanHT-12 V3.0 expression beadchip | ILMN_1680892 | AF151107.1 | human_transcriptome |
Illumina HumanWG-6_V2_0_R2 | ILMN_1680892 | NM_080701.3 | human_transcriptome |
Illumina HumanWG-6_V2_0_R2 | ILMN_1680892 | AF151107.1 | human_transcriptome |
Illumina HumanRef-8 WG-DASL v3.0 | ILMN_1680892 | NM_080701.3 | human_transcriptome |
Illumina HumanRef-8 WG-DASL v3.0 | ILMN_1680892 | AF151107.1 | human_transcriptome |
