Assay Details
Gene Symbol: LRP12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_013437.4 | NP_038465.1 | 1 - 2 | 205 | 96 | |
GenBank mRNA | AF166350.1 | - | 1 - 2 | 120 | 96 | |
AK289752.1 | - | 1 - 2 | 109 | 96 | ||
AL698796.1 | - | 1 - 2 | 173 | 96 | ||
BC032109.2 | - | 1 - 2 | 81 | 4842797 | 96 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC08001511.hg.1
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
ABI
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | PSR08019875.hg.1 | TC08001511.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR08019875.hg.1 | ENST00000276654.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR08019875.hg.1 | ENST00000424843.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR08019875.hg.1 | NM_001135703.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR08019875.hg.1 | NM_013437.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800011395.hg.1 | ENST00000276654.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011395.hg.1 | ENST00000276654.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011395.hg.1 | ENST00000276654.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011395.hg.1 | ENST00000276654.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011395.hg.1 | ENST00000276654.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011395.hg.1 | ENST00000276654.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011395.hg.1 | ENST00000276654.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011395.hg.1 | ENST00000276654.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011395.hg.1 | ENST00000276654.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011395.hg.1 | ENST00000276654.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011395.hg.1 | ENST00000276654.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011395.hg.1 | ENST00000276654.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011395.hg.1 | ENST00000276654.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011395.hg.1 | ENST00000424843.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011395.hg.1 | ENST00000424843.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011395.hg.1 | ENST00000424843.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011395.hg.1 | ENST00000424843.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011395.hg.1 | ENST00000424843.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011395.hg.1 | ENST00000424843.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011395.hg.1 | ENST00000424843.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011395.hg.1 | ENST00000424843.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011395.hg.1 | ENST00000424843.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011395.hg.1 | ENST00000424843.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011395.hg.1 | ENST00000424843.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011395.hg.1 | ENST00000424843.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011395.hg.1 | ENST00000518375.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011395.hg.1 | ENST00000518375.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011395.hg.1 | ENST00000518375.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011395.hg.1 | ENST00000518375.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011395.hg.1 | ENST00000518375.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011395.hg.1 | ENST00000518375.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011395.hg.1 | ENST00000518375.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011395.hg.1 | ENST00000519675.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011395.hg.1 | ENST00000519675.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011395.hg.1 | ENST00000519675.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011395.hg.1 | ENST00000519675.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011395.hg.1 | ENST00000520770.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011395.hg.1 | ENST00000520770.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011395.hg.1 | ENST00000520770.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011395.hg.1 | ENST00000520770.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011395.hg.1 | ENST00000520770.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011395.hg.1 | ENST00000522046.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011395.hg.1 | ENST00000522046.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011395.hg.1 | ENST00000522046.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011395.hg.1 | ENST00000523007.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011395.hg.1 | ENST00000523007.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011395.hg.1 | ENST00000523007.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011395.hg.1 | NM_001135703.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800011395.hg.1 | NM_001135703.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800011395.hg.1 | NM_001135703.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800011395.hg.1 | NM_001135703.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800011395.hg.1 | NM_001135703.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800011395.hg.1 | NM_001135703.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800011395.hg.1 | NM_001135703.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800011395.hg.1 | NM_001135703.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800011395.hg.1 | NM_001135703.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800011395.hg.1 | NM_001135703.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800011395.hg.1 | NM_001135703.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800011395.hg.1 | NM_001135703.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800011395.hg.1 | NM_013437.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800011395.hg.1 | NM_013437.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800011395.hg.1 | NM_013437.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800011395.hg.1 | NM_013437.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800011395.hg.1 | NM_013437.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800011395.hg.1 | NM_013437.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800011395.hg.1 | NM_013437.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800011395.hg.1 | NM_013437.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800011395.hg.1 | NM_013437.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800011395.hg.1 | NM_013437.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800011395.hg.1 | NM_013437.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800011395.hg.1 | NM_013437.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800011395.hg.1 | NM_013437.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR08019876.hg.1 | TC08001511.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR08019876.hg.1 | ENST00000276654.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR08019876.hg.1 | ENST00000424843.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR08019876.hg.1 | NM_001135703.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR08019876.hg.1 | NM_013437.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR08019874.hg.1 | TC08001511.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR08019874.hg.1 | ENST00000276654.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR08019874.hg.1 | NM_013437.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR0800177708.hg.1 | ENST00000276654.9 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0800177708.hg.1 | NM_013437.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR0800177708.hg.1 | ENST00000276654.9 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0800177708.hg.1 | NM_013437.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR0800177708.hg.1 | TC0800011395.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0800177709.hg.1 | ENST00000276654.9 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0800177709.hg.1 | ENST00000424843.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0800177709.hg.1 | ENST00000519675.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0800177709.hg.1 | ENST00000520770.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0800177709.hg.1 | NM_001135703.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0800177709.hg.1 | NM_013437.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR0800177709.hg.1 | ENST00000276654.9 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0800177709.hg.1 | ENST00000424843.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0800177709.hg.1 | ENST00000519675.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0800177709.hg.1 | ENST00000520770.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0800177709.hg.1 | NM_001135703.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0800177709.hg.1 | NM_013437.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR0800177709.hg.1 | TC0800011395.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0800177711.hg.1 | ENST00000276654.9 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0800177711.hg.1 | ENST00000424843.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0800177711.hg.1 | ENST00000519675.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0800177711.hg.1 | ENST00000520770.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0800177711.hg.1 | NM_001135703.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0800177711.hg.1 | NM_013437.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR0800177711.hg.1 | ENST00000276654.9 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0800177711.hg.1 | ENST00000424843.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0800177711.hg.1 | ENST00000519675.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0800177711.hg.1 | ENST00000520770.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0800177711.hg.1 | NM_001135703.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0800177711.hg.1 | NM_013437.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR0800177711.hg.1 | TC0800011395.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | TC0800011395.hg.1 | ENST00000276654.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800011395.hg.1 | ENST00000276654.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800011395.hg.1 | ENST00000276654.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800011395.hg.1 | ENST00000276654.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800011395.hg.1 | ENST00000276654.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800011395.hg.1 | ENST00000276654.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800011395.hg.1 | ENST00000276654.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800011395.hg.1 | ENST00000276654.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800011395.hg.1 | ENST00000276654.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800011395.hg.1 | ENST00000276654.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800011395.hg.1 | ENST00000276654.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800011395.hg.1 | ENST00000276654.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800011395.hg.1 | ENST00000276654.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800011395.hg.1 | ENST00000424843.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800011395.hg.1 | ENST00000424843.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800011395.hg.1 | ENST00000424843.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800011395.hg.1 | ENST00000424843.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800011395.hg.1 | ENST00000424843.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800011395.hg.1 | ENST00000424843.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800011395.hg.1 | ENST00000424843.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800011395.hg.1 | ENST00000424843.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800011395.hg.1 | ENST00000424843.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800011395.hg.1 | ENST00000424843.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800011395.hg.1 | ENST00000424843.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800011395.hg.1 | ENST00000424843.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800011395.hg.1 | ENST00000518375.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800011395.hg.1 | ENST00000518375.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800011395.hg.1 | ENST00000518375.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800011395.hg.1 | ENST00000518375.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800011395.hg.1 | ENST00000518375.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800011395.hg.1 | ENST00000518375.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800011395.hg.1 | ENST00000518375.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800011395.hg.1 | ENST00000519675.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800011395.hg.1 | ENST00000519675.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800011395.hg.1 | ENST00000519675.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800011395.hg.1 | ENST00000519675.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800011395.hg.1 | ENST00000520770.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800011395.hg.1 | ENST00000520770.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800011395.hg.1 | ENST00000520770.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800011395.hg.1 | ENST00000520770.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800011395.hg.1 | ENST00000520770.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800011395.hg.1 | ENST00000522046.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800011395.hg.1 | ENST00000522046.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800011395.hg.1 | ENST00000522046.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800011395.hg.1 | ENST00000523007.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800011395.hg.1 | ENST00000523007.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800011395.hg.1 | ENST00000523007.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800011395.hg.1 | NM_001135703.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0800011395.hg.1 | NM_001135703.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0800011395.hg.1 | NM_001135703.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0800011395.hg.1 | NM_001135703.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0800011395.hg.1 | NM_001135703.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0800011395.hg.1 | NM_001135703.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0800011395.hg.1 | NM_001135703.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0800011395.hg.1 | NM_001135703.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0800011395.hg.1 | NM_001135703.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0800011395.hg.1 | NM_001135703.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0800011395.hg.1 | NM_001135703.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0800011395.hg.1 | NM_001135703.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0800011395.hg.1 | NM_013437.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0800011395.hg.1 | NM_013437.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0800011395.hg.1 | NM_013437.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0800011395.hg.1 | NM_013437.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0800011395.hg.1 | NM_013437.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0800011395.hg.1 | NM_013437.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0800011395.hg.1 | NM_013437.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0800011395.hg.1 | NM_013437.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0800011395.hg.1 | NM_013437.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0800011395.hg.1 | NM_013437.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0800011395.hg.1 | NM_013437.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0800011395.hg.1 | NM_013437.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0800011395.hg.1 | NM_013437.4 | Refseq |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Human Genome Survey Microarray Version 2.0 | 110857 | - | - |
