Assay Details
Gene Symbol: CCDC82 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_017018307.1 | XP_016873796.1 | 1 | 315 | 90 | |
GenBank mRNA | AF161533.1 | - | 1 | 353 | 90 | |
AK313893.1 | - | 1 | 386 | 90 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_017018307.1 transcripts.
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
MicroArray Probes:
Affymetrix
ABI
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC1100012052.hg.1 | ENST00000278520.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012052.hg.1 | ENST00000278520.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012052.hg.1 | ENST00000278520.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012052.hg.1 | ENST00000278520.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012052.hg.1 | ENST00000278520.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012052.hg.1 | ENST00000278520.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012052.hg.1 | ENST00000278520.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012052.hg.1 | ENST00000278520.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012052.hg.1 | ENST00000278520.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012052.hg.1 | ENST00000278520.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012052.hg.1 | ENST00000278520.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012052.hg.1 | ENST00000278520.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012052.hg.1 | ENST00000278520.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012052.hg.1 | ENST00000423339.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012052.hg.1 | ENST00000423339.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012052.hg.1 | ENST00000423339.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012052.hg.1 | ENST00000423339.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012052.hg.1 | ENST00000423339.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012052.hg.1 | ENST00000423339.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012052.hg.1 | ENST00000423339.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012052.hg.1 | ENST00000423339.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012052.hg.1 | ENST00000423339.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012052.hg.1 | ENST00000524836.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012052.hg.1 | ENST00000524836.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012052.hg.1 | ENST00000524836.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012052.hg.1 | ENST00000524836.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012052.hg.1 | ENST00000524836.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012052.hg.1 | ENST00000525786.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012052.hg.1 | ENST00000525786.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012052.hg.1 | ENST00000525786.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012052.hg.1 | ENST00000530106.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012052.hg.1 | ENST00000530106.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012052.hg.1 | ENST00000530106.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012052.hg.1 | ENST00000530106.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012052.hg.1 | ENST00000530106.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012052.hg.1 | ENST00000530203.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012052.hg.1 | ENST00000530203.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012052.hg.1 | ENST00000530203.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012052.hg.1 | ENST00000530203.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012052.hg.1 | ENST00000538597.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012052.hg.1 | ENST00000538597.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012052.hg.1 | ENST00000538597.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012052.hg.1 | ENST00000545264.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012052.hg.1 | ENST00000545264.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012052.hg.1 | ENST00000545264.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012052.hg.1 | ENST00000545264.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012052.hg.1 | NM_024725.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100012052.hg.1 | NM_024725.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100012052.hg.1 | NM_024725.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100012052.hg.1 | NM_024725.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100012052.hg.1 | NM_024725.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100012052.hg.1 | NM_024725.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100012052.hg.1 | NM_024725.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100012052.hg.1 | NM_024725.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100012052.hg.1 | NM_024725.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100012052.hg.1 | NM_024725.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100012052.hg.1 | NM_024725.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100012052.hg.1 | NM_024725.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100012052.hg.1 | NM_024725.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1100012052.hg.1 | ENST00000278520.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012052.hg.1 | ENST00000278520.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012052.hg.1 | ENST00000278520.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012052.hg.1 | ENST00000278520.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012052.hg.1 | ENST00000278520.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012052.hg.1 | ENST00000278520.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012052.hg.1 | ENST00000278520.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012052.hg.1 | ENST00000278520.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012052.hg.1 | ENST00000278520.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012052.hg.1 | ENST00000278520.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012052.hg.1 | ENST00000278520.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012052.hg.1 | ENST00000278520.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012052.hg.1 | ENST00000278520.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012052.hg.1 | ENST00000423339.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012052.hg.1 | ENST00000423339.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012052.hg.1 | ENST00000423339.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012052.hg.1 | ENST00000423339.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012052.hg.1 | ENST00000423339.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012052.hg.1 | ENST00000423339.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012052.hg.1 | ENST00000423339.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012052.hg.1 | ENST00000423339.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012052.hg.1 | ENST00000423339.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012052.hg.1 | ENST00000524836.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012052.hg.1 | ENST00000524836.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012052.hg.1 | ENST00000524836.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012052.hg.1 | ENST00000524836.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012052.hg.1 | ENST00000524836.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012052.hg.1 | ENST00000525786.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012052.hg.1 | ENST00000525786.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012052.hg.1 | ENST00000525786.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012052.hg.1 | ENST00000530106.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012052.hg.1 | ENST00000530106.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012052.hg.1 | ENST00000530106.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012052.hg.1 | ENST00000530106.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012052.hg.1 | ENST00000530106.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012052.hg.1 | ENST00000530203.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012052.hg.1 | ENST00000530203.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012052.hg.1 | ENST00000530203.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012052.hg.1 | ENST00000530203.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012052.hg.1 | ENST00000538597.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012052.hg.1 | ENST00000538597.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012052.hg.1 | ENST00000538597.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012052.hg.1 | ENST00000545264.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012052.hg.1 | ENST00000545264.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012052.hg.1 | ENST00000545264.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012052.hg.1 | ENST00000545264.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012052.hg.1 | NM_024725.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1100012052.hg.1 | NM_024725.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1100012052.hg.1 | NM_024725.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1100012052.hg.1 | NM_024725.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1100012052.hg.1 | NM_024725.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1100012052.hg.1 | NM_024725.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1100012052.hg.1 | NM_024725.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1100012052.hg.1 | NM_024725.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1100012052.hg.1 | NM_024725.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1100012052.hg.1 | NM_024725.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1100012052.hg.1 | NM_024725.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1100012052.hg.1 | NM_024725.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1100012052.hg.1 | NM_024725.3 | Refseq |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Human Genome Survey Microarray Version 2.0 | 149860 | - | - |
