Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009452.hg.1 |
ENST00000261837.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009452.hg.1 |
ENST00000261837.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009452.hg.1 |
ENST00000261837.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009452.hg.1 |
ENST00000261837.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009452.hg.1 |
ENST00000261837.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009452.hg.1 |
ENST00000261837.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009452.hg.1 |
ENST00000261837.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009452.hg.1 |
ENST00000261837.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009452.hg.1 |
ENST00000261837.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009452.hg.1 |
ENST00000261837.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009452.hg.1 |
ENST00000261837.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009452.hg.1 |
ENST00000261837.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009452.hg.1 |
ENST00000261837.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009452.hg.1 |
ENST00000261837.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009452.hg.1 |
ENST00000261837.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009452.hg.1 |
ENST00000261837.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009452.hg.1 |
ENST00000261837.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009452.hg.1 |
ENST00000261837.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009452.hg.1 |
ENST00000261837.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009452.hg.1 |
ENST00000261837.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009452.hg.1 |
ENST00000261837.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009452.hg.1 |
ENST00000261837.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009452.hg.1 |
ENST00000358784.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009452.hg.1 |
ENST00000358784.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009452.hg.1 |
ENST00000358784.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009452.hg.1 |
ENST00000358784.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009452.hg.1 |
ENST00000358784.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009452.hg.1 |
ENST00000358784.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009452.hg.1 |
ENST00000358784.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009452.hg.1 |
ENST00000358784.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009452.hg.1 |
ENST00000358784.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009452.hg.1 |
ENST00000358784.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009452.hg.1 |
ENST00000358784.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009452.hg.1 |
ENST00000358784.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009452.hg.1 |
ENST00000358784.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009452.hg.1 |
ENST00000358784.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009452.hg.1 |
ENST00000358784.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009452.hg.1 |
ENST00000358784.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009452.hg.1 |
ENST00000396335.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009452.hg.1 |
ENST00000396335.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009452.hg.1 |
ENST00000396335.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009452.hg.1 |
ENST00000396335.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009452.hg.1 |
ENST00000396335.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009452.hg.1 |
ENST00000396335.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009452.hg.1 |
ENST00000396335.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009452.hg.1 |
ENST00000396335.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009452.hg.1 |
ENST00000396335.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009452.hg.1 |
ENST00000396335.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009452.hg.1 |
ENST00000396335.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009452.hg.1 |
ENST00000396335.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009452.hg.1 |
ENST00000396335.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009452.hg.1 |
ENST00000557936.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009452.hg.1 |
ENST00000557936.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009452.hg.1 |
ENST00000557936.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009452.hg.1 |
ENST00000557936.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009452.hg.1 |
ENST00000557936.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009452.hg.1 |
ENST00000557936.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009452.hg.1 |
ENST00000557936.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009452.hg.1 |
ENST00000557936.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009452.hg.1 |
ENST00000557936.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009452.hg.1 |
ENST00000557936.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009452.hg.1 |
ENST00000558122.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009452.hg.1 |
ENST00000558122.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009452.hg.1 |
ENST00000558519.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009452.hg.1 |
ENST00000558519.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009452.hg.1 |
ENST00000558519.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009452.hg.1 |
ENST00000558519.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009452.hg.1 |
ENST00000558519.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009452.hg.1 |
ENST00000558519.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009452.hg.1 |
ENST00000558519.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009452.hg.1 |
ENST00000558519.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009452.hg.1 |
ENST00000558519.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009452.hg.1 |
ENST00000559348.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009452.hg.1 |
ENST00000559348.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009452.hg.1 |
ENST00000559348.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009452.hg.1 |
ENST00000559348.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009452.hg.1 |
ENST00000559348.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009452.hg.1 |
ENST00000559348.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009452.hg.1 |
ENST00000559348.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009452.hg.1 |
ENST00000559348.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009452.hg.1 |
ENST00000559348.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009452.hg.1 |
ENST00000559348.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009452.hg.1 |
ENST00000559541.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009452.hg.1 |
ENST00000559541.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009452.hg.1 |
ENST00000559541.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009452.hg.1 |
ENST00000559541.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009452.hg.1 |
ENST00000559541.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009452.hg.1 |
ENST00000559541.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009452.hg.1 |
ENST00000559541.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009452.hg.1 |
ENST00000559541.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009452.hg.1 |
ENST00000560075.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009452.hg.1 |
ENST00000560075.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009452.hg.1 |
ENST00000560075.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009452.hg.1 |
ENST00000560075.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009452.hg.1 |
ENST00000560075.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009452.hg.1 |
ENST00000560075.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009452.hg.1 |
ENST00000560085.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009452.hg.1 |
ENST00000560085.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009452.hg.1 |
ENST00000560085.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009452.hg.1 |
ENST00000560085.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009452.hg.1 |
ENST00000560085.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009452.hg.1 |
ENST00000560116.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009452.hg.1 |
ENST00000560116.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009452.hg.1 |
ENST00000560116.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009452.hg.1 |
ENST00000560116.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009452.hg.1 |
ENST00000561313.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009452.hg.1 |
ENST00000561313.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009452.hg.1 |
ENST00000561313.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009452.hg.1 |
ENST00000561313.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009452.hg.1 |
ENST00000561313.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009452.hg.1 |
ENST00000561313.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009452.hg.1 |
ENST00000561313.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009452.hg.1 |
NM_006578.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009452.hg.1 |
NM_006578.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009452.hg.1 |
NM_006578.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009452.hg.1 |
NM_006578.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009452.hg.1 |
NM_006578.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009452.hg.1 |
NM_006578.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009452.hg.1 |
NM_006578.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009452.hg.1 |
NM_006578.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009452.hg.1 |
NM_006578.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009452.hg.1 |
NM_006578.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009452.hg.1 |
NM_006578.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009452.hg.1 |
NM_006578.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009452.hg.1 |
NM_006578.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009452.hg.1 |
NM_006578.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009452.hg.1 |
NM_006578.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009452.hg.1 |
NM_006578.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009452.hg.1 |
NM_006578.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009452.hg.1 |
NM_016194.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009452.hg.1 |
NM_016194.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009452.hg.1 |
NM_016194.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009452.hg.1 |
NM_016194.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009452.hg.1 |
NM_016194.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009452.hg.1 |
NM_016194.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009452.hg.1 |
NM_016194.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009452.hg.1 |
NM_016194.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009452.hg.1 |
NM_016194.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009452.hg.1 |
NM_016194.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009452.hg.1 |
NM_016194.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009452.hg.1 |
NM_016194.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009452.hg.1 |
NM_016194.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009452.hg.1 |
NM_016194.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009452.hg.1 |
NM_016194.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009452.hg.1 |
NM_016194.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009452.hg.1 |
NM_016194.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009452.hg.1 |
NM_016194.3 |
Refseq |