Assay Details
| Gene Symbol: BCL7A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001024808.2 | NP_001019979.1 | 2 - 3 | 377 | 82 | |
| NM_020993.4 | NP_066273.1 | 2 - 3 | 377 | 82 | ||
| GenBank mRNA | AK296159.1 | - | 2 - 3 | 379 | 82 | |
| AK316486.1 | - | 2 - 3 | 275 | 82 | ||
| BC094723.1 | - | 2 - 3 | 188 | 5300018 | 82 | |
| BC113627.1 | - | 2 - 3 | 201 | 8322679 | 82 | |
| BM801030.1 | - | 336 | 82 | |||
| X89984.1 | - | 2 - 3 | 1124 | 82 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC12000967.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR12012629.hg.1 | ENST00000261822.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR12012629.hg.1 | ENST00000538010.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR12012629.hg.1 | NM_001024808.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR12012629.hg.1 | NM_020993.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR12012629.hg.1 | TC12000967.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR12012628.hg.1 | ENST00000261822.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR12012628.hg.1 | ENST00000538010.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR12012628.hg.1 | NM_001024808.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR12012628.hg.1 | NM_020993.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR12012628.hg.1 | TC12000967.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR12012631.hg.1 | ENST00000261822.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR12012631.hg.1 | ENST00000538010.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR12012631.hg.1 | NM_001024808.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR12012631.hg.1 | NM_020993.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR12012631.hg.1 | TC12000967.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR12012626.hg.1 | ENST00000261822.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR12012626.hg.1 | ENST00000538010.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR12012626.hg.1 | NM_001024808.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR12012626.hg.1 | NM_020993.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR12012626.hg.1 | TC12000967.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009192.hg.1 | ENST00000261822.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009192.hg.1 | ENST00000261822.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009192.hg.1 | ENST00000261822.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009192.hg.1 | ENST00000261822.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009192.hg.1 | ENST00000261822.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009192.hg.1 | ENST00000261822.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009192.hg.1 | ENST00000261822.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009192.hg.1 | ENST00000261822.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009192.hg.1 | ENST00000261822.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009192.hg.1 | ENST00000261822.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009192.hg.1 | ENST00000432926.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009192.hg.1 | ENST00000432926.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009192.hg.1 | ENST00000432926.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009192.hg.1 | ENST00000432926.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009192.hg.1 | ENST00000432926.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009192.hg.1 | ENST00000432926.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009192.hg.1 | ENST00000538010.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009192.hg.1 | ENST00000538010.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009192.hg.1 | ENST00000538010.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009192.hg.1 | ENST00000538010.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009192.hg.1 | ENST00000538010.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009192.hg.1 | ENST00000538010.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009192.hg.1 | ENST00000538010.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009192.hg.1 | ENST00000538010.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009192.hg.1 | ENST00000538010.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009192.hg.1 | ENST00000538010.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009192.hg.1 | ENST00000538010.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009192.hg.1 | ENST00000538010.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009192.hg.1 | ENST00000538010.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009192.hg.1 | NM_001024808.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009192.hg.1 | NM_001024808.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009192.hg.1 | NM_001024808.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009192.hg.1 | NM_001024808.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009192.hg.1 | NM_001024808.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009192.hg.1 | NM_001024808.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009192.hg.1 | NM_001024808.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009192.hg.1 | NM_001024808.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009192.hg.1 | NM_001024808.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009192.hg.1 | NM_001024808.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009192.hg.1 | NM_001024808.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009192.hg.1 | NM_020993.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009192.hg.1 | NM_020993.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009192.hg.1 | NM_020993.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009192.hg.1 | NM_020993.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009192.hg.1 | NM_020993.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009192.hg.1 | NM_020993.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009192.hg.1 | NM_020993.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009192.hg.1 | NM_020993.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009192.hg.1 | NM_020993.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009192.hg.1 | NM_020993.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009192.hg.1 | NM_020993.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009192.hg.1 | NM_020993.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR12012630.hg.1 | ENST00000261822.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR12012630.hg.1 | ENST00000538010.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR12012630.hg.1 | NM_001024808.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR12012630.hg.1 | NM_020993.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR12012630.hg.1 | TC12000967.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | PSR1200168080.hg.1 | ENST00000261822.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200168080.hg.1 | ENST00000538010.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200168080.hg.1 | NM_001024808.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200168080.hg.1 | NM_020993.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200168080.hg.1 | ENST00000261822.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200168080.hg.1 | ENST00000538010.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200168080.hg.1 | NM_001024808.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200168080.hg.1 | NM_020993.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200168080.hg.1 | TC1200009192.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1200168088.hg.1 | ENST00000261822.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200168088.hg.1 | ENST00000432926.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200168088.hg.1 | ENST00000538010.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200168088.hg.1 | NM_001024808.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200168088.hg.1 | NM_020993.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200168088.hg.1 | ENST00000261822.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200168088.hg.1 | ENST00000432926.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200168088.hg.1 | ENST00000538010.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200168088.hg.1 | NM_001024808.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200168088.hg.1 | NM_020993.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200168088.hg.1 | TC1200009192.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1200168081.hg.1 | ENST00000261822.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200168081.hg.1 | ENST00000432926.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200168081.hg.1 | ENST00000538010.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200168081.hg.1 | NM_001024808.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200168081.hg.1 | NM_020993.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200168081.hg.1 | ENST00000261822.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200168081.hg.1 | ENST00000432926.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200168081.hg.1 | ENST00000538010.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200168081.hg.1 | NM_001024808.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200168081.hg.1 | NM_020993.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200168081.hg.1 | TC1200009192.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1200168084.hg.1 | ENST00000261822.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200168084.hg.1 | ENST00000432926.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200168084.hg.1 | ENST00000538010.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200168084.hg.1 | NM_001024808.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200168084.hg.1 | NM_020993.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200168084.hg.1 | ENST00000261822.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200168084.hg.1 | ENST00000432926.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200168084.hg.1 | ENST00000538010.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200168084.hg.1 | NM_001024808.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200168084.hg.1 | NM_020993.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200168084.hg.1 | TC1200009192.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1200168086.hg.1 | ENST00000261822.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200168086.hg.1 | ENST00000432926.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200168086.hg.1 | ENST00000538010.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200168086.hg.1 | NM_001024808.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200168086.hg.1 | NM_020993.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200168086.hg.1 | ENST00000261822.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200168086.hg.1 | ENST00000432926.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200168086.hg.1 | ENST00000538010.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200168086.hg.1 | NM_001024808.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200168086.hg.1 | NM_020993.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200168086.hg.1 | TC1200009192.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC1200009192.hg.1 | ENST00000261822.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009192.hg.1 | ENST00000261822.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009192.hg.1 | ENST00000261822.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009192.hg.1 | ENST00000261822.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009192.hg.1 | ENST00000261822.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009192.hg.1 | ENST00000261822.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009192.hg.1 | ENST00000261822.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009192.hg.1 | ENST00000261822.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009192.hg.1 | ENST00000261822.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009192.hg.1 | ENST00000261822.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009192.hg.1 | ENST00000432926.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009192.hg.1 | ENST00000432926.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009192.hg.1 | ENST00000432926.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009192.hg.1 | ENST00000432926.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009192.hg.1 | ENST00000432926.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009192.hg.1 | ENST00000432926.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009192.hg.1 | ENST00000538010.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009192.hg.1 | ENST00000538010.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009192.hg.1 | ENST00000538010.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009192.hg.1 | ENST00000538010.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009192.hg.1 | ENST00000538010.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009192.hg.1 | ENST00000538010.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009192.hg.1 | ENST00000538010.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009192.hg.1 | ENST00000538010.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009192.hg.1 | ENST00000538010.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009192.hg.1 | ENST00000538010.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009192.hg.1 | ENST00000538010.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009192.hg.1 | ENST00000538010.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009192.hg.1 | ENST00000538010.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009192.hg.1 | NM_001024808.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200009192.hg.1 | NM_001024808.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200009192.hg.1 | NM_001024808.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200009192.hg.1 | NM_001024808.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200009192.hg.1 | NM_001024808.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200009192.hg.1 | NM_001024808.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200009192.hg.1 | NM_001024808.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200009192.hg.1 | NM_001024808.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200009192.hg.1 | NM_001024808.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200009192.hg.1 | NM_001024808.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200009192.hg.1 | NM_001024808.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200009192.hg.1 | NM_020993.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200009192.hg.1 | NM_020993.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200009192.hg.1 | NM_020993.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200009192.hg.1 | NM_020993.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200009192.hg.1 | NM_020993.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200009192.hg.1 | NM_020993.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200009192.hg.1 | NM_020993.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200009192.hg.1 | NM_020993.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200009192.hg.1 | NM_020993.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200009192.hg.1 | NM_020993.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200009192.hg.1 | NM_020993.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200009192.hg.1 | NM_020993.4 | Refseq |

BCL7A
Genomic Map