Assay Details
Gene Symbol: MYBL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001080416.3 | NP_001073885.1 | 15 - 16 | 2539 | 92 | |
XM_011517533.2 | XP_011515835.1 | 15 - 16 | 2595 | 92 | ||
XM_017013455.1 | XP_016868944.1 | 15 - 16 | 2405 | 92 | ||
XM_017013456.1 | XP_016868945.1 | 15 - 16 | 2387 | 92 | ||
GenBank mRNA | AK308490.1 | - | 8 - 9 | 1306 | 92 | |
X13294.1 | - | 15 - 16 | 2232 | 92 | ||
X66087.1 | - | 6 - 7 | 939 | 92 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC08001286.hg.1
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
Function(s)
Process(es)
transcription from RNA polymerase II promoter
cell differentiation
positive regulation of transcription, DNA-templated
positive regulation of transcription from RNA polymerase II promoter
RNA polymerase II core promoter proximal region sequence-specific DNA binding
transcriptional activator activity, RNA polymerase II core promoter proximal region sequence-specific binding
transcription factor activity, RNA polymerase II transcription factor recruiting
transcription from RNA polymerase II promoter
cell differentiation
positive regulation of transcription, DNA-templated
positive regulation of transcription from RNA polymerase II promoter
RNA polymerase II core promoter proximal region sequence-specific DNA binding
transcriptional activator activity, RNA polymerase II core promoter proximal region sequence-specific binding
transcription factor activity, RNA polymerase II transcription factor recruiting
MicroArray Probes:
Affymetrix
ABI
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC0800010685.hg.1 | ENST00000517885.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800010685.hg.1 | ENST00000517885.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800010685.hg.1 | ENST00000517885.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800010685.hg.1 | ENST00000517885.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800010685.hg.1 | ENST00000517885.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800010685.hg.1 | ENST00000517885.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800010685.hg.1 | ENST00000517885.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800010685.hg.1 | ENST00000517885.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800010685.hg.1 | ENST00000517885.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800010685.hg.1 | ENST00000517885.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800010685.hg.1 | ENST00000517885.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800010685.hg.1 | ENST00000517885.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800010685.hg.1 | ENST00000517885.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800010685.hg.1 | ENST00000517885.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800010685.hg.1 | ENST00000517885.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800010685.hg.1 | ENST00000517885.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800010685.hg.1 | ENST00000517885.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800010685.hg.1 | ENST00000518079.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800010685.hg.1 | ENST00000518079.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800010685.hg.1 | ENST00000518079.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800010685.hg.1 | ENST00000518079.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800010685.hg.1 | ENST00000518079.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800010685.hg.1 | ENST00000522419.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800010685.hg.1 | ENST00000522419.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800010685.hg.1 | ENST00000522419.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800010685.hg.1 | ENST00000522677.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800010685.hg.1 | ENST00000522677.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800010685.hg.1 | ENST00000522677.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800010685.hg.1 | ENST00000522677.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800010685.hg.1 | ENST00000522677.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800010685.hg.1 | ENST00000522677.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800010685.hg.1 | ENST00000522677.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800010685.hg.1 | ENST00000522677.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800010685.hg.1 | ENST00000522677.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800010685.hg.1 | ENST00000522677.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800010685.hg.1 | ENST00000522677.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800010685.hg.1 | ENST00000522677.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800010685.hg.1 | ENST00000522677.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800010685.hg.1 | ENST00000522677.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800010685.hg.1 | ENST00000522677.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800010685.hg.1 | ENST00000522677.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800010685.hg.1 | ENST00000522677.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800010685.hg.1 | ENST00000522677.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800010685.hg.1 | ENST00000522677.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800010685.hg.1 | ENST00000522677.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800010685.hg.1 | ENST00000522677.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800010685.hg.1 | ENST00000522677.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800010685.hg.1 | ENST00000522677.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800010685.hg.1 | ENST00000523304.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800010685.hg.1 | ENST00000523304.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800010685.hg.1 | ENST00000523304.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800010685.hg.1 | ENST00000523304.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800010685.hg.1 | ENST00000523304.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800010685.hg.1 | ENST00000523304.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800010685.hg.1 | ENST00000523304.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800010685.hg.1 | ENST00000523304.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800010685.hg.1 | ENST00000523304.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800010685.hg.1 | ENST00000524176.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800010685.hg.1 | ENST00000524176.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800010685.hg.1 | ENST00000524176.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800010685.hg.1 | ENST00000524176.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800010685.hg.1 | ENST00000524176.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800010685.hg.1 | ENST00000524176.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800010685.hg.1 | ENST00000524176.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800010685.hg.1 | ENST00000524176.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800010685.hg.1 | ENST00000524176.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800010685.hg.1 | ENST00000524176.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800010685.hg.1 | ENST00000524176.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800010685.hg.1 | ENST00000524176.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800010685.hg.1 | ENST00000524176.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800010685.hg.1 | ENST00000524176.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800010685.hg.1 | ENST00000524176.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800010685.hg.1 | ENST00000524176.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800010685.hg.1 | ENST00000524176.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800010685.hg.1 | ENST00000524176.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800010685.hg.1 | NM_001080416.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800010685.hg.1 | NM_001080416.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800010685.hg.1 | NM_001080416.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800010685.hg.1 | NM_001080416.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800010685.hg.1 | NM_001080416.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800010685.hg.1 | NM_001080416.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800010685.hg.1 | NM_001080416.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800010685.hg.1 | NM_001080416.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800010685.hg.1 | NM_001080416.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800010685.hg.1 | NM_001080416.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800010685.hg.1 | NM_001080416.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800010685.hg.1 | NM_001080416.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800010685.hg.1 | NM_001080416.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800010685.hg.1 | NM_001080416.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800010685.hg.1 | NM_001080416.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800010685.hg.1 | NM_001080416.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800010685.hg.1 | NM_001080416.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800010685.hg.1 | NM_001080416.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800010685.hg.1 | NM_001080416.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800010685.hg.1 | NM_001080416.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800010685.hg.1 | NM_001080416.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800010685.hg.1 | NM_001080416.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800010685.hg.1 | NM_001080416.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800010685.hg.1 | NM_001144755.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800010685.hg.1 | NM_001144755.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800010685.hg.1 | NM_001144755.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800010685.hg.1 | NM_001144755.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800010685.hg.1 | NM_001144755.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800010685.hg.1 | NM_001144755.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800010685.hg.1 | NM_001144755.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800010685.hg.1 | NM_001144755.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800010685.hg.1 | NM_001144755.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800010685.hg.1 | NM_001144755.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800010685.hg.1 | NM_001144755.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800010685.hg.1 | NM_001144755.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800010685.hg.1 | NM_001144755.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800010685.hg.1 | NM_001144755.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800010685.hg.1 | NM_001144755.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800010685.hg.1 | NM_001144755.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800010685.hg.1 | NM_001144755.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800010685.hg.1 | NM_001144755.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800010685.hg.1 | NM_001144755.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800010685.hg.1 | NM_001144755.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800010685.hg.1 | NM_001144755.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800010685.hg.1 | NM_001144755.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800010685.hg.1 | NM_001294282.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800010685.hg.1 | NM_001294282.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800010685.hg.1 | NM_001294282.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800010685.hg.1 | NM_001294282.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800010685.hg.1 | NM_001294282.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800010685.hg.1 | NM_001294282.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800010685.hg.1 | NM_001294282.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800010685.hg.1 | NM_001294282.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800010685.hg.1 | NM_001294282.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800010685.hg.1 | NM_001294282.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800010685.hg.1 | NM_001294282.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800010685.hg.1 | NM_001294282.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800010685.hg.1 | NM_001294282.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800010685.hg.1 | NM_001294282.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800010685.hg.1 | NM_001294282.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800010685.hg.1 | NM_001294282.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800010685.hg.1 | NM_001294282.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800010685.hg.1 | NM_001294282.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800010685.hg.1 | NM_001294282.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800010685.hg.1 | NM_001294282.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800010685.hg.1 | NM_001294282.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800010685.hg.1 | NM_001294282.1 | Refseq |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Human Genome Survey Microarray Version 2.0 | 207803 | - | - |
