Assay Details
| Gene Symbol: LVRN | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK055381.1 | - | 1 | 1951 | 63 | |
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Transcript Location | Amplicon Length | |
|---|---|---|---|---|---|---|---|
| GenBankmRNA | CB270410.1 | - | 81 | Chr. - - [-] on Build | 63 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | adaptor related protein complex 3 sigma 1 subunit | AP3S1 |
CLAPS3, Sigma3A |
-40.8kb | Sense | Chr. 5 - 115841606 - 115914081 on NCBI Build GRCh38 |
| Downstream | ADP ribosylation factor like GTPase 14 effector protein like | ARL14EPL | 92.9kb | Sense | Chr. 5 - 116051140 - 116060023 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

LVRN
Genomic Map