| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR08017087.hg.1 |
TC08001325.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR08017087.hg.1 |
ENST00000297354.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR08017087.hg.1 |
NM_153225.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800010800.hg.1 |
ENST00000297354.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800010800.hg.1 |
ENST00000297354.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800010800.hg.1 |
ENST00000297354.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800010800.hg.1 |
ENST00000297354.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800010800.hg.1 |
ENST00000297354.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800010800.hg.1 |
ENST00000297354.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800010800.hg.1 |
ENST00000297354.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800010800.hg.1 |
ENST00000297354.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800010800.hg.1 |
ENST00000297354.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800010800.hg.1 |
ENST00000519697.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800010800.hg.1 |
ENST00000519697.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800010800.hg.1 |
ENST00000519697.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800010800.hg.1 |
ENST00000519697.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800010800.hg.1 |
ENST00000519697.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800010800.hg.1 |
ENST00000519697.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800010800.hg.1 |
ENST00000519697.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800010800.hg.1 |
NM_153225.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800010800.hg.1 |
NM_153225.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800010800.hg.1 |
NM_153225.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800010800.hg.1 |
NM_153225.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800010800.hg.1 |
NM_153225.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800010800.hg.1 |
NM_153225.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800010800.hg.1 |
NM_153225.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800010800.hg.1 |
NM_153225.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800010800.hg.1 |
NM_153225.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR08017086.hg.1 |
TC08001325.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR08017086.hg.1 |
ENST00000297354.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR08017086.hg.1 |
ENST00000519697.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR08017086.hg.1 |
NM_153225.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0800173334.hg.1 |
ENST00000297354.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0800173334.hg.1 |
ENST00000519697.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0800173334.hg.1 |
NM_153225.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0800173334.hg.1 |
ENST00000297354.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0800173334.hg.1 |
ENST00000519697.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0800173334.hg.1 |
NM_153225.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0800173334.hg.1 |
TC0800010800.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0800173336.hg.1 |
ENST00000297354.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0800173336.hg.1 |
NM_153225.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0800173336.hg.1 |
ENST00000297354.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0800173336.hg.1 |
NM_153225.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0800173336.hg.1 |
TC0800010800.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
TC0800010800.hg.1 |
ENST00000297354.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800010800.hg.1 |
ENST00000297354.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800010800.hg.1 |
ENST00000297354.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800010800.hg.1 |
ENST00000297354.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800010800.hg.1 |
ENST00000297354.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800010800.hg.1 |
ENST00000297354.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800010800.hg.1 |
ENST00000297354.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800010800.hg.1 |
ENST00000297354.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800010800.hg.1 |
ENST00000297354.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800010800.hg.1 |
ENST00000519697.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800010800.hg.1 |
ENST00000519697.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800010800.hg.1 |
ENST00000519697.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800010800.hg.1 |
ENST00000519697.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800010800.hg.1 |
ENST00000519697.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800010800.hg.1 |
ENST00000519697.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800010800.hg.1 |
ENST00000519697.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800010800.hg.1 |
NM_153225.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0800010800.hg.1 |
NM_153225.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0800010800.hg.1 |
NM_153225.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0800010800.hg.1 |
NM_153225.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0800010800.hg.1 |
NM_153225.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0800010800.hg.1 |
NM_153225.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0800010800.hg.1 |
NM_153225.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0800010800.hg.1 |
NM_153225.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0800010800.hg.1 |
NM_153225.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0800173335.hg.1 |
ENST00000297354.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0800173335.hg.1 |
NM_153225.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0800173335.hg.1 |
ENST00000297354.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0800173335.hg.1 |
NM_153225.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0800173335.hg.1 |
TC0800010800.hg.1 |
Clariom D TC |