Assay Details
| Gene Symbol: SHROOM4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_020717.3 | NP_065768.2 | 3 - 4 | 437 | 66 | |
| NR_027121.1 | 3 - 4 | 437 | 66 | |||
| XM_017029682.1 | XP_016885171.1 | 3 - 4 | 543 | 66 | ||
| XM_017029684.1 | XP_016885173.1 | 2 - 3 | 560 | 66 | ||
| XM_017029685.1 | XP_016885174.1 | 3 - 4 | 560 | 66 | ||
| XM_017029687.1 | XP_016885176.1 | 3 - 4 | 547 | 66 | ||
| GenBank mRNA | AB033028.1 | - | 3 - 4 | 437 | 66 | |
| AY044234.1 | - | 2 - 3 | 518 | 66 | ||
| BC151240.1 | - | 3 - 4 | 448 | 8860513 | 66 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC0X001039.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR0X013996.hg.1 | TC0X001039.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X013996.hg.1 | ENST00000289292.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X013996.hg.1 | ENST00000376020.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X013996.hg.1 | ENST00000460112.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X013996.hg.1 | NM_020717.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X013996.hg.1 | NR_027121.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009684.hg.1 | ENST00000289292.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009684.hg.1 | ENST00000289292.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009684.hg.1 | ENST00000289292.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009684.hg.1 | ENST00000289292.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009684.hg.1 | ENST00000289292.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009684.hg.1 | ENST00000289292.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009684.hg.1 | ENST00000289292.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009684.hg.1 | ENST00000289292.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009684.hg.1 | ENST00000289292.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009684.hg.1 | ENST00000289292.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009684.hg.1 | ENST00000289292.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009684.hg.1 | ENST00000289292.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009684.hg.1 | ENST00000289292.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009684.hg.1 | ENST00000289292.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009684.hg.1 | ENST00000289292.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009684.hg.1 | ENST00000289292.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009684.hg.1 | ENST00000289292.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009684.hg.1 | ENST00000376020.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009684.hg.1 | ENST00000376020.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009684.hg.1 | ENST00000376020.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009684.hg.1 | ENST00000376020.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009684.hg.1 | ENST00000376020.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009684.hg.1 | ENST00000376020.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009684.hg.1 | ENST00000376020.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009684.hg.1 | ENST00000376020.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009684.hg.1 | ENST00000376020.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009684.hg.1 | ENST00000376020.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009684.hg.1 | ENST00000376020.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009684.hg.1 | ENST00000376020.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009684.hg.1 | ENST00000376020.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009684.hg.1 | ENST00000376020.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009684.hg.1 | ENST00000376020.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009684.hg.1 | ENST00000376020.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009684.hg.1 | ENST00000376020.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009684.hg.1 | ENST00000376020.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009684.hg.1 | ENST00000460112.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009684.hg.1 | ENST00000460112.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009684.hg.1 | ENST00000460112.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009684.hg.1 | ENST00000460112.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009684.hg.1 | ENST00000460112.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009684.hg.1 | ENST00000460112.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009684.hg.1 | ENST00000460112.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009684.hg.1 | ENST00000460112.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009684.hg.1 | ENST00000460112.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009684.hg.1 | ENST00000460112.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009684.hg.1 | ENST00000460112.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009684.hg.1 | ENST00000460112.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009684.hg.1 | ENST00000460112.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009684.hg.1 | ENST00000483955.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009684.hg.1 | ENST00000483955.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009684.hg.1 | ENST00000483955.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009684.hg.1 | ENST00000484922.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009684.hg.1 | ENST00000484922.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009684.hg.1 | NM_020717.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009684.hg.1 | NM_020717.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009684.hg.1 | NM_020717.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009684.hg.1 | NM_020717.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009684.hg.1 | NM_020717.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009684.hg.1 | NM_020717.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009684.hg.1 | NM_020717.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009684.hg.1 | NM_020717.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009684.hg.1 | NM_020717.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009684.hg.1 | NM_020717.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009684.hg.1 | NM_020717.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009684.hg.1 | NM_020717.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009684.hg.1 | NM_020717.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009684.hg.1 | NM_020717.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009684.hg.1 | NM_020717.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009684.hg.1 | NM_020717.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009684.hg.1 | NM_020717.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009684.hg.1 | NM_020717.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009684.hg.1 | NR_027121.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009684.hg.1 | NR_027121.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009684.hg.1 | NR_027121.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009684.hg.1 | NR_027121.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009684.hg.1 | NR_027121.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009684.hg.1 | NR_027121.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009684.hg.1 | NR_027121.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009684.hg.1 | NR_027121.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009684.hg.1 | NR_027121.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009684.hg.1 | NR_027121.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009684.hg.1 | NR_027121.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009684.hg.1 | NR_027121.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009684.hg.1 | NR_027121.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009684.hg.1 | NR_027121.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009684.hg.1 | NR_027121.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009684.hg.1 | NR_027121.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00009684.hg.1 | ENST00000289292.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009684.hg.1 | ENST00000289292.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009684.hg.1 | ENST00000289292.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009684.hg.1 | ENST00000289292.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009684.hg.1 | ENST00000289292.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009684.hg.1 | ENST00000289292.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009684.hg.1 | ENST00000289292.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009684.hg.1 | ENST00000289292.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009684.hg.1 | ENST00000289292.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009684.hg.1 | ENST00000289292.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009684.hg.1 | ENST00000289292.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009684.hg.1 | ENST00000289292.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009684.hg.1 | ENST00000289292.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009684.hg.1 | ENST00000289292.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009684.hg.1 | ENST00000289292.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009684.hg.1 | ENST00000289292.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009684.hg.1 | ENST00000289292.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009684.hg.1 | ENST00000376020.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009684.hg.1 | ENST00000376020.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009684.hg.1 | ENST00000376020.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009684.hg.1 | ENST00000376020.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009684.hg.1 | ENST00000376020.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009684.hg.1 | ENST00000376020.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009684.hg.1 | ENST00000376020.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009684.hg.1 | ENST00000376020.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009684.hg.1 | ENST00000376020.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009684.hg.1 | ENST00000376020.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009684.hg.1 | ENST00000376020.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009684.hg.1 | ENST00000376020.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009684.hg.1 | ENST00000376020.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009684.hg.1 | ENST00000376020.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009684.hg.1 | ENST00000376020.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009684.hg.1 | ENST00000376020.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009684.hg.1 | ENST00000376020.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009684.hg.1 | ENST00000376020.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009684.hg.1 | ENST00000460112.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009684.hg.1 | ENST00000460112.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009684.hg.1 | ENST00000460112.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009684.hg.1 | ENST00000460112.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009684.hg.1 | ENST00000460112.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009684.hg.1 | ENST00000460112.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009684.hg.1 | ENST00000460112.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009684.hg.1 | ENST00000460112.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009684.hg.1 | ENST00000460112.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009684.hg.1 | ENST00000460112.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009684.hg.1 | ENST00000460112.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009684.hg.1 | ENST00000460112.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009684.hg.1 | ENST00000460112.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009684.hg.1 | ENST00000483955.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009684.hg.1 | ENST00000483955.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009684.hg.1 | ENST00000483955.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009684.hg.1 | ENST00000484922.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009684.hg.1 | ENST00000484922.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009684.hg.1 | NM_020717.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00009684.hg.1 | NM_020717.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00009684.hg.1 | NM_020717.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00009684.hg.1 | NM_020717.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00009684.hg.1 | NM_020717.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00009684.hg.1 | NM_020717.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00009684.hg.1 | NM_020717.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00009684.hg.1 | NM_020717.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00009684.hg.1 | NM_020717.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00009684.hg.1 | NM_020717.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00009684.hg.1 | NM_020717.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00009684.hg.1 | NM_020717.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00009684.hg.1 | NM_020717.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00009684.hg.1 | NM_020717.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00009684.hg.1 | NM_020717.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00009684.hg.1 | NM_020717.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00009684.hg.1 | NM_020717.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00009684.hg.1 | NM_020717.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00009684.hg.1 | NR_027121.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00009684.hg.1 | NR_027121.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00009684.hg.1 | NR_027121.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00009684.hg.1 | NR_027121.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00009684.hg.1 | NR_027121.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00009684.hg.1 | NR_027121.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00009684.hg.1 | NR_027121.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00009684.hg.1 | NR_027121.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00009684.hg.1 | NR_027121.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00009684.hg.1 | NR_027121.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00009684.hg.1 | NR_027121.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00009684.hg.1 | NR_027121.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00009684.hg.1 | NR_027121.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00009684.hg.1 | NR_027121.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00009684.hg.1 | NR_027121.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00009684.hg.1 | NR_027121.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00166194.hg.1 | ENST00000289292.11 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00166194.hg.1 | ENST00000376020.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00166194.hg.1 | ENST00000460112.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00166194.hg.1 | NM_020717.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00166194.hg.1 | NR_027121.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00166194.hg.1 | ENST00000289292.11 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00166194.hg.1 | ENST00000376020.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00166194.hg.1 | ENST00000460112.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00166194.hg.1 | NM_020717.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00166194.hg.1 | NR_027121.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00166194.hg.1 | TC0X00009684.hg.1 | Clariom D TC |

SHROOM4
Genomic Map