Assay Details
| Gene Symbol: DYNLRB1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001281729.1 | NP_001268658.1 | 2 - 3 | 156 | 109 | |
| GenBank mRNA | BC007223.2 | - | 1 - 2 | 78 | 109 | |
| BM561321.1 | - | 2 - 3 | 126 | 109 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001281729.1 transcripts.
Set Membership: |
|
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
ABI
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC2000007191.hg.1 | ENST00000300469.13 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007191.hg.1 | ENST00000300469.13 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007191.hg.1 | ENST00000300469.13 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007191.hg.1 | ENST00000357156.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007191.hg.1 | ENST00000357156.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007191.hg.1 | ENST00000357156.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007191.hg.1 | ENST00000357156.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007191.hg.1 | ENST00000357156.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007191.hg.1 | ENST00000374846.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007191.hg.1 | ENST00000374846.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007191.hg.1 | ENST00000374846.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007191.hg.1 | ENST00000374846.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007191.hg.1 | ENST00000374846.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007191.hg.1 | ENST00000480759.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007191.hg.1 | ENST00000480759.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007191.hg.1 | ENST00000480759.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007191.hg.1 | ENST00000480759.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007191.hg.1 | ENST00000480759.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007191.hg.1 | ENST00000480759.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007191.hg.1 | NM_001281727.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007191.hg.1 | NM_001281727.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007191.hg.1 | NM_001281727.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007191.hg.1 | NM_001281727.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007191.hg.1 | NM_001281727.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007191.hg.1 | NM_001281728.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007191.hg.1 | NM_001281728.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007191.hg.1 | NM_001281728.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007191.hg.1 | NM_001281728.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007191.hg.1 | NM_001281728.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007191.hg.1 | NM_001281728.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007191.hg.1 | NM_001281729.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007191.hg.1 | NM_001281729.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007191.hg.1 | NM_001281729.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007191.hg.1 | NM_001281729.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007191.hg.1 | NM_001281729.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007191.hg.1 | NM_001281729.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007191.hg.1 | NM_014183.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007191.hg.1 | NM_014183.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007191.hg.1 | NM_014183.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007191.hg.1 | NM_014183.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007191.hg.1 | NM_014183.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007191.hg.1 | NM_177953.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007191.hg.1 | NM_177953.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007191.hg.1 | NM_177953.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007191.hg.1 | NM_177953.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007191.hg.1 | NM_177953.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007191.hg.1 | NM_177953.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007191.hg.1 | NR_104032.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007191.hg.1 | NR_104032.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007191.hg.1 | NR_104032.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007191.hg.1 | NR_104032.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007191.hg.1 | NR_104032.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007191.hg.1 | NR_104032.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2000149354.hg.1 | ENST00000300469.13 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000149354.hg.1 | ENST00000480759.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000149354.hg.1 | NM_001281729.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2000149354.hg.1 | TC2000007191.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR2000149354.hg.1 | ENST00000300469.13 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000149354.hg.1 | ENST00000480759.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000149354.hg.1 | NM_001281729.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007191.hg.1 | ENST00000300469.13 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007191.hg.1 | ENST00000300469.13 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007191.hg.1 | ENST00000300469.13 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007191.hg.1 | ENST00000357156.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007191.hg.1 | ENST00000357156.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007191.hg.1 | ENST00000357156.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007191.hg.1 | ENST00000357156.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007191.hg.1 | ENST00000357156.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007191.hg.1 | ENST00000374846.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007191.hg.1 | ENST00000374846.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007191.hg.1 | ENST00000374846.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007191.hg.1 | ENST00000374846.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007191.hg.1 | ENST00000374846.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007191.hg.1 | ENST00000480759.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007191.hg.1 | ENST00000480759.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007191.hg.1 | ENST00000480759.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007191.hg.1 | ENST00000480759.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007191.hg.1 | ENST00000480759.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007191.hg.1 | ENST00000480759.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007191.hg.1 | NM_001281727.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007191.hg.1 | NM_001281727.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007191.hg.1 | NM_001281727.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007191.hg.1 | NM_001281727.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007191.hg.1 | NM_001281727.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007191.hg.1 | NM_001281728.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007191.hg.1 | NM_001281728.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007191.hg.1 | NM_001281728.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007191.hg.1 | NM_001281728.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007191.hg.1 | NM_001281728.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007191.hg.1 | NM_001281728.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007191.hg.1 | NM_001281729.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007191.hg.1 | NM_001281729.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007191.hg.1 | NM_001281729.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007191.hg.1 | NM_001281729.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007191.hg.1 | NM_001281729.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007191.hg.1 | NM_001281729.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007191.hg.1 | NM_014183.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007191.hg.1 | NM_014183.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007191.hg.1 | NM_014183.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007191.hg.1 | NM_014183.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007191.hg.1 | NM_014183.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007191.hg.1 | NM_177953.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007191.hg.1 | NM_177953.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007191.hg.1 | NM_177953.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007191.hg.1 | NM_177953.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007191.hg.1 | NM_177953.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007191.hg.1 | NM_177953.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007191.hg.1 | NR_104032.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007191.hg.1 | NR_104032.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007191.hg.1 | NR_104032.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007191.hg.1 | NR_104032.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007191.hg.1 | NR_104032.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007191.hg.1 | NR_104032.1 | Refseq |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Human Genome Survey Microarray Version 2.0 | 184693 | - | - |

DYNLRB1
Genomic Map