Assay Details
Gene Symbol: FUBP3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_003934.1 | NP_003925.1 | 1 - 2 | 193 | 85 | |
XM_005272232.1 | XP_005272289.1 | 1 - 2 | 199 | 85 | ||
XM_011519172.2 | XP_011517474.1 | 1 - 2 | 199 | 85 | ||
GenBank mRNA | AK295155.1 | - | 1 - 2 | 193 | 85 | |
AL537190.3 | - | 1 - 2 | 164 | 85 | ||
BC007874.2 | - | 1 - 2 | 138 | 85 | ||
BC137338.1 | - | 1 - 2 | 190 | 9021336 | 85 | |
BC137340.1 | - | 1 - 2 | 190 | 9021338 | 85 | |
BI461266.1 | - | 1 - 2 | 198 | 85 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC09000738.hg.1
Additional Information:
This assay is in the 5'UTR region of the BC007874.2 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
transcription, DNA-templated
transcription from RNA polymerase II promoter
positive regulation of gene expression
positive regulation of transcription, DNA-templated
positive regulation of transcription from RNA polymerase II promoter
RNA polymerase II core promoter proximal region sequence-specific DNA binding
transcriptional activator activity, RNA polymerase II core promoter proximal region sequence-specific binding
protein binding
poly(A) RNA binding
transcription, DNA-templated
transcription from RNA polymerase II promoter
positive regulation of gene expression
positive regulation of transcription, DNA-templated
positive regulation of transcription from RNA polymerase II promoter
RNA polymerase II core promoter proximal region sequence-specific DNA binding
transcriptional activator activity, RNA polymerase II core promoter proximal region sequence-specific binding
protein binding
poly(A) RNA binding
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | PSR09009322.hg.1 | TC09000738.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR09009322.hg.1 | ENST00000358721.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR09009322.hg.1 | ENST00000467100.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR09009322.hg.1 | NM_003934.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR09009324.hg.1 | TC09000738.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR09009324.hg.1 | ENST00000319725.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR09009324.hg.1 | ENST00000358721.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR09009324.hg.1 | ENST00000372376.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR09009324.hg.1 | ENST00000467100.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR09009324.hg.1 | NM_003934.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR09009329.hg.1 | TC09000738.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR09009329.hg.1 | ENST00000319725.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR09009329.hg.1 | ENST00000358721.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR09009329.hg.1 | ENST00000372376.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR09009329.hg.1 | NM_003934.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900008959.hg.1 | ENST00000319725.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900008959.hg.1 | ENST00000319725.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900008959.hg.1 | ENST00000319725.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900008959.hg.1 | ENST00000319725.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900008959.hg.1 | ENST00000319725.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900008959.hg.1 | ENST00000319725.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900008959.hg.1 | ENST00000319725.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900008959.hg.1 | ENST00000319725.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900008959.hg.1 | ENST00000319725.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900008959.hg.1 | ENST00000319725.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900008959.hg.1 | ENST00000319725.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900008959.hg.1 | ENST00000319725.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900008959.hg.1 | ENST00000319725.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900008959.hg.1 | ENST00000319725.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900008959.hg.1 | ENST00000319725.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900008959.hg.1 | ENST00000319725.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900008959.hg.1 | ENST00000319725.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900008959.hg.1 | ENST00000319725.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900008959.hg.1 | ENST00000319725.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900008959.hg.1 | ENST00000319725.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900008959.hg.1 | ENST00000319725.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900008959.hg.1 | ENST00000319725.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900008959.hg.1 | ENST00000319725.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900008959.hg.1 | ENST00000319725.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900008959.hg.1 | ENST00000465949.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900008959.hg.1 | ENST00000465949.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900008959.hg.1 | ENST00000465949.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900008959.hg.1 | ENST00000465949.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900008959.hg.1 | ENST00000465949.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900008959.hg.1 | ENST00000467100.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900008959.hg.1 | ENST00000467100.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900008959.hg.1 | ENST00000467100.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900008959.hg.1 | ENST00000467100.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900008959.hg.1 | ENST00000467100.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900008959.hg.1 | ENST00000467100.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900008959.hg.1 | ENST00000467100.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900008959.hg.1 | ENST00000472006.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900008959.hg.1 | ENST00000472006.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900008959.hg.1 | ENST00000472006.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900008959.hg.1 | ENST00000472006.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900008959.hg.1 | ENST00000487406.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900008959.hg.1 | ENST00000487406.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900008959.hg.1 | ENST00000487406.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900008959.hg.1 | ENST00000487406.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900008959.hg.1 | ENST00000487406.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900008959.hg.1 | ENST00000487406.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900008959.hg.1 | ENST00000487406.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900008959.hg.1 | ENST00000487406.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900008959.hg.1 | ENST00000492199.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900008959.hg.1 | ENST00000492199.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900008959.hg.1 | ENST00000492199.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900008959.hg.1 | ENST00000492199.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900008959.hg.1 | NM_003934.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900008959.hg.1 | NM_003934.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900008959.hg.1 | NM_003934.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900008959.hg.1 | NM_003934.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900008959.hg.1 | NM_003934.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900008959.hg.1 | NM_003934.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900008959.hg.1 | NM_003934.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900008959.hg.1 | NM_003934.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900008959.hg.1 | NM_003934.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900008959.hg.1 | NM_003934.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900008959.hg.1 | NM_003934.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900008959.hg.1 | NM_003934.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900008959.hg.1 | NM_003934.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900008959.hg.1 | NM_003934.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900008959.hg.1 | NM_003934.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900008959.hg.1 | NM_003934.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900008959.hg.1 | NM_003934.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900008959.hg.1 | NM_003934.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900008959.hg.1 | NM_003934.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900008959.hg.1 | NM_003934.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900008959.hg.1 | NM_003934.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900008959.hg.1 | NM_003934.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900008959.hg.1 | NM_003934.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900008959.hg.1 | NM_003934.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900008959.hg.1 | NM_003934.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0900160893.hg.1 | ENST00000467100.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0900160893.hg.1 | NM_003934.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0900160893.hg.1 | ENST00000467100.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0900160893.hg.1 | NM_003934.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0900160893.hg.1 | TC0900008959.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0900160899.hg.1 | ENST00000319725.9 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0900160899.hg.1 | ENST00000465949.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0900160899.hg.1 | NM_003934.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0900160899.hg.1 | ENST00000319725.9 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0900160899.hg.1 | ENST00000465949.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0900160899.hg.1 | NM_003934.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0900160899.hg.1 | TC0900008959.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0900160895.hg.1 | ENST00000319725.9 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0900160895.hg.1 | ENST00000467100.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0900160895.hg.1 | NM_003934.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0900160895.hg.1 | ENST00000319725.9 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0900160895.hg.1 | ENST00000467100.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0900160895.hg.1 | NM_003934.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0900160895.hg.1 | TC0900008959.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | TC0900008959.hg.1 | ENST00000319725.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900008959.hg.1 | ENST00000319725.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900008959.hg.1 | ENST00000319725.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900008959.hg.1 | ENST00000319725.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900008959.hg.1 | ENST00000319725.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900008959.hg.1 | ENST00000319725.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900008959.hg.1 | ENST00000319725.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900008959.hg.1 | ENST00000319725.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900008959.hg.1 | ENST00000319725.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900008959.hg.1 | ENST00000319725.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900008959.hg.1 | ENST00000319725.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900008959.hg.1 | ENST00000319725.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900008959.hg.1 | ENST00000319725.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900008959.hg.1 | ENST00000319725.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900008959.hg.1 | ENST00000319725.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900008959.hg.1 | ENST00000319725.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900008959.hg.1 | ENST00000319725.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900008959.hg.1 | ENST00000319725.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900008959.hg.1 | ENST00000319725.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900008959.hg.1 | ENST00000319725.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900008959.hg.1 | ENST00000319725.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900008959.hg.1 | ENST00000319725.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900008959.hg.1 | ENST00000319725.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900008959.hg.1 | ENST00000319725.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900008959.hg.1 | ENST00000465949.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900008959.hg.1 | ENST00000465949.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900008959.hg.1 | ENST00000465949.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900008959.hg.1 | ENST00000465949.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900008959.hg.1 | ENST00000465949.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900008959.hg.1 | ENST00000467100.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900008959.hg.1 | ENST00000467100.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900008959.hg.1 | ENST00000467100.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900008959.hg.1 | ENST00000467100.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900008959.hg.1 | ENST00000467100.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900008959.hg.1 | ENST00000467100.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900008959.hg.1 | ENST00000467100.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900008959.hg.1 | ENST00000472006.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900008959.hg.1 | ENST00000472006.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900008959.hg.1 | ENST00000472006.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900008959.hg.1 | ENST00000472006.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900008959.hg.1 | ENST00000487406.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900008959.hg.1 | ENST00000487406.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900008959.hg.1 | ENST00000487406.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900008959.hg.1 | ENST00000487406.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900008959.hg.1 | ENST00000487406.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900008959.hg.1 | ENST00000487406.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900008959.hg.1 | ENST00000487406.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900008959.hg.1 | ENST00000487406.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900008959.hg.1 | ENST00000492199.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900008959.hg.1 | ENST00000492199.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900008959.hg.1 | ENST00000492199.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900008959.hg.1 | ENST00000492199.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900008959.hg.1 | NM_003934.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0900008959.hg.1 | NM_003934.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0900008959.hg.1 | NM_003934.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0900008959.hg.1 | NM_003934.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0900008959.hg.1 | NM_003934.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0900008959.hg.1 | NM_003934.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0900008959.hg.1 | NM_003934.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0900008959.hg.1 | NM_003934.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0900008959.hg.1 | NM_003934.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0900008959.hg.1 | NM_003934.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0900008959.hg.1 | NM_003934.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0900008959.hg.1 | NM_003934.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0900008959.hg.1 | NM_003934.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0900008959.hg.1 | NM_003934.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0900008959.hg.1 | NM_003934.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0900008959.hg.1 | NM_003934.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0900008959.hg.1 | NM_003934.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0900008959.hg.1 | NM_003934.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0900008959.hg.1 | NM_003934.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0900008959.hg.1 | NM_003934.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0900008959.hg.1 | NM_003934.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0900008959.hg.1 | NM_003934.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0900008959.hg.1 | NM_003934.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0900008959.hg.1 | NM_003934.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0900008959.hg.1 | NM_003934.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0900160894.hg.1 | ENST00000319725.9 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0900160894.hg.1 | ENST00000467100.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0900160894.hg.1 | NM_003934.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0900160894.hg.1 | ENST00000319725.9 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0900160894.hg.1 | ENST00000467100.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0900160894.hg.1 | NM_003934.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0900160894.hg.1 | TC0900008959.hg.1 | Clariom D TC |
