Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR21005352.hg.1 |
NM_033656.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR21005352.hg.1 |
TC21000459.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR21005353.hg.1 |
ENST00000342449.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR21005353.hg.1 |
NM_033656.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR21005353.hg.1 |
TC21000459.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR2100155052.hg.1 |
NM_033656.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2100155052.hg.1 |
TC2100008165.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR2100155052.hg.1 |
NM_033656.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2100155053.hg.1 |
ENST00000342449.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2100155053.hg.1 |
NM_033656.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2100155053.hg.1 |
TC2100008165.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR2100155053.hg.1 |
ENST00000342449.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2100155053.hg.1 |
NM_033656.3 |
Refseq |