Assay Details
| Gene Symbol: LOC283588 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK055958.1 | - | 1 | 556 | 101 | |
| AK095276.1 | - | 1 | 1303 | 101 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P29930734 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P29930753 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P29930741 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P09470079 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P29930707 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P29930798 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P29930739 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P09470028 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P29930737 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P29930757 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P09470013 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P29930761 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P29930786 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P29930698 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P09470059 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P09470008 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P29930780 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P29930787 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P09470002 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P29930765 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P09470063 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P29930781 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P09470017 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P09470019 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P29930783 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P09470003 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P16726981 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P29930770 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P09469996 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P09470004 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P09470024 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P09470046 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P29930768 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P29930752 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P29930796 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P16726985 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P29930708 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P29930751 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P29930700 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P29930778 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P29930742 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P09470055 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P09469995 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P09470022 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P29930789 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P09470011 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P09470021 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P29930716 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P29930736 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P09470050 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P29930740 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P09470048 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P29930779 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P29930774 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P29930728 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P29930735 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P09470018 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P29930797 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P29930764 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P09470045 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P09470090 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P09470000 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P29930790 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P29930785 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P09470009 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P29930772 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P29930738 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P29930773 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P29930788 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P29930794 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P09470010 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P29930733 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P29930713 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P29930727 | - | - |
| Agilent SurePrint G3 Human CGH Microarray 4x180K | 15895 | - | - |
| Agilent SurePrint G3 Human CGH Microarray 8x60K | 19996 | - | - |
| Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k | GT_44k_23_P334795 | - | - |
| Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k | GT_44k_32_P88946 | - | - |
| Agilent Whole Human Genome Oligo Microarray 4x44K | 19869 | - | - |
| Agilent Whole Human Genome Oligo Microarray 4x44K | 40857 | - | - |
| Agilent Human Genome CGH Microarray 44B | A_14_P108191 | - | - |
| Agilent Human Genome CGH Microarray 44A | A_14_P108191 | - | - |
| Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray | 43237 | - | - |
| Agilent Human Genome CGH + SNP Microarray | A_14_P108191 | - | - |
| Agilent Whole Human Genome Microarray 4x44K v2 | A_33_P3305203 | - | - |
| Whole Human Genome Oligo Microarray G4112A | A_23_P334795 | - | - |
| Whole Human Genome Oligo Microarray G4112A | A_32_P88946 | - | - |
| Agilent Human Genome CGH Microarray 105A (G4412A) | A_14_P108191 | - | - |
| Agilent Human Genome 244K CGH Microarray (Alpha Test) | A_14_P108191 | - | - |
| Agilent Human Genome CGH Microarray 4x44K | A_14_P108191 | - | - |
| Agilent SurePrint G3 CGH array (4X180K) CCA_VUMC design AMADID#022522 | 148856 | - | - |
| Agilent HD CGH Microarray (2X105K) Oxford design AMADID#019015 | 48735 | - | - |
| Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_14_P108191 | - | - |
| Agilent Human 1B Microarray G4111A | A_32_P744434 | - | - |
| Agilent SurePrint G3 Human CGH Microarray 2x400K | A_14_P108191 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Gene 1.0 ST Array | 7980743 | - | - |
| Affymetrix Human Gene 1.1 ST Array | 7980742 | - | - |
| Affymetrix HT HG-U133+ PM Array Plate | 1557113_PM_at | - | - |
| Human Genome U133 Plus 2.0 Array | 1557113_at | - | - |
| Human Genome U133 Plus 2.0 Array | 1557113_PM_at | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1557113_at1 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1557113_at6 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1557113_at11 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1557113_at7 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1557113_at3 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1557113_at5 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1557113_at2 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1557113_at4 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1557113_at10 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1557113_at9 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1557113_at8 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Human Genome Survey Microarray Version 2.0 | 281755 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| HumanHT-12 v4 Expression BeadChip | ILMN_3295651 | AK095276.1 | human_transcriptome |
| HumanHT-12 v4 Expression BeadChip | ILMN_3295651 | AK055958.1 | human_transcriptome |
| HumanHT-12 v4 Expression BeadChip | ILMN_3207230 | AK095276.1 | human_transcriptome |
| HumanHT-12 v4 Expression BeadChip | ILMN_3207230 | AK055958.1 | human_transcriptome |

LOC283588
Genomic Map