| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Gene 1.0 ST Array |
8105990 |
- |
- |
| Affymetrix Human Gene 1.0 ST Array |
8112477 |
- |
- |
| Affymetrix Human Gene 1.0 ST Array |
8112520 |
- |
- |
| Affymetrix Human Gene 1.0 ST Array |
8105988 |
- |
- |
| Affymetrix Human Gene 1.0 ST Array |
8105936 |
- |
- |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500007687.hg.1 |
ENST00000514492.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500011016.hg.1 |
ENST00000505635.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500011016.hg.1 |
ENST00000505635.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500013299.hg.1 |
ENST00000507274.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500013299.hg.1 |
ENST00000507274.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500013299.hg.1 |
ENST00000507274.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500013299.hg.1 |
ENST00000507274.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500013299.hg.1 |
ENST00000507274.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500013299.hg.1 |
ENST00000507274.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500013299.hg.1 |
ENST00000508395.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500013299.hg.1 |
ENST00000508395.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500013299.hg.1 |
ENST00000508395.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500013299.hg.1 |
ENST00000508395.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500013299.hg.1 |
ENST00000508395.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500013299.hg.1 |
ENST00000508395.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500013299.hg.1 |
ENST00000508395.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500013299.hg.1 |
ENST00000508395.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500013299.hg.1 |
ENST00000512782.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500013299.hg.1 |
ENST00000512782.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500013299.hg.1 |
ENST00000512782.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500013299.hg.1 |
ENST00000512782.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500013299.hg.1 |
ENST00000512782.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500013299.hg.1 |
ENST00000512782.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500013299.hg.1 |
ENST00000514048.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500013299.hg.1 |
ENST00000514048.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500013299.hg.1 |
ENST00000514048.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500013299.hg.1 |
ENST00000514048.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500013299.hg.1 |
ENST00000514048.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500013299.hg.1 |
ENST00000514048.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500013299.hg.1 |
ENST00000514048.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500013299.hg.1 |
ENST00000514048.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500013299.hg.1 |
ENST00000514048.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500013299.hg.1 |
ENST00000514048.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500013299.hg.1 |
ENST00000514048.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500013299.hg.1 |
ENST00000623525.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500010279.hg.1 |
ENST00000502532.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500007686.hg.1 |
ENST00000515156.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500007686.hg.1 |
ENST00000515156.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0500152364.hg.1 |
NR_073404.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0500152364.hg.1 |
NR_073404.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0500152364.hg.1 |
TC0500007695.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
TC0500013299.hg.1 |
ENST00000507274.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500013299.hg.1 |
ENST00000507274.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500013299.hg.1 |
ENST00000507274.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500013299.hg.1 |
ENST00000507274.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500013299.hg.1 |
ENST00000507274.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500013299.hg.1 |
ENST00000507274.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500013299.hg.1 |
ENST00000508395.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500013299.hg.1 |
ENST00000508395.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500013299.hg.1 |
ENST00000508395.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500013299.hg.1 |
ENST00000508395.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500013299.hg.1 |
ENST00000508395.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500013299.hg.1 |
ENST00000508395.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500013299.hg.1 |
ENST00000508395.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500013299.hg.1 |
ENST00000508395.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500013299.hg.1 |
ENST00000512782.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500013299.hg.1 |
ENST00000512782.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500013299.hg.1 |
ENST00000512782.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500013299.hg.1 |
ENST00000512782.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500013299.hg.1 |
ENST00000512782.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500013299.hg.1 |
ENST00000512782.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500013299.hg.1 |
ENST00000514048.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500013299.hg.1 |
ENST00000514048.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500013299.hg.1 |
ENST00000514048.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500013299.hg.1 |
ENST00000514048.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500013299.hg.1 |
ENST00000514048.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500013299.hg.1 |
ENST00000514048.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500013299.hg.1 |
ENST00000514048.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500013299.hg.1 |
ENST00000514048.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500013299.hg.1 |
ENST00000514048.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500013299.hg.1 |
ENST00000514048.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500013299.hg.1 |
ENST00000514048.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500013299.hg.1 |
ENST00000623525.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500007686.hg.1 |
ENST00000515156.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500007686.hg.1 |
ENST00000515156.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500010279.hg.1 |
ENST00000502532.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500007695.hg.1 |
NR_073404.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0500007687.hg.1 |
ENST00000514492.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500011016.hg.1 |
ENST00000505635.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500011016.hg.1 |
ENST00000505635.1 |
Ensembl |