| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000008293.hg.1 |
ENST00000441457.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000008293.hg.1 |
ENST00000441457.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000008293.hg.1 |
ENST00000441457.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000008293.hg.1 |
ENST00000441457.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000008293.hg.1 |
ENST00000441457.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000008293.hg.1 |
ENST00000441457.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000008293.hg.1 |
ENST00000441457.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000008293.hg.1 |
ENST00000441457.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000008293.hg.1 |
NR_120671.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000008293.hg.1 |
NR_120671.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000008293.hg.1 |
NR_120671.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000008293.hg.1 |
NR_120671.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000008293.hg.1 |
NR_120671.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000008293.hg.1 |
NR_120671.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000008293.hg.1 |
NR_120671.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000008293.hg.1 |
NR_120671.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1000156231.hg.1 |
ENST00000441457.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1000156231.hg.1 |
NR_120671.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1000156231.hg.1 |
ENST00000441457.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1000156231.hg.1 |
NR_120671.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1000156231.hg.1 |
TC1000008293.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR1000156233.hg.1 |
ENST00000441457.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1000156233.hg.1 |
NR_120671.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1000156233.hg.1 |
ENST00000441457.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1000156233.hg.1 |
NR_120671.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1000156233.hg.1 |
TC1000008293.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
TC1000008293.hg.1 |
ENST00000441457.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000008293.hg.1 |
ENST00000441457.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000008293.hg.1 |
ENST00000441457.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000008293.hg.1 |
ENST00000441457.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000008293.hg.1 |
ENST00000441457.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000008293.hg.1 |
ENST00000441457.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000008293.hg.1 |
ENST00000441457.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000008293.hg.1 |
ENST00000441457.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000008293.hg.1 |
NR_120671.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000008293.hg.1 |
NR_120671.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000008293.hg.1 |
NR_120671.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000008293.hg.1 |
NR_120671.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000008293.hg.1 |
NR_120671.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000008293.hg.1 |
NR_120671.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000008293.hg.1 |
NR_120671.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000008293.hg.1 |
NR_120671.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1000156228.hg.1 |
ENST00000441457.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1000156228.hg.1 |
NR_120671.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1000156228.hg.1 |
ENST00000441457.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1000156228.hg.1 |
NR_120671.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1000156228.hg.1 |
TC1000008293.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR1000156229.hg.1 |
ENST00000441457.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1000156229.hg.1 |
NR_120671.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1000156229.hg.1 |
ENST00000441457.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1000156229.hg.1 |
NR_120671.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1000156229.hg.1 |
TC1000008293.hg.1 |
Clariom D TC |