Assay Details
Gene Symbol: LINC01257 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_026670.2 | 1 - 2 | 222 | 79 | ||
GenBank mRNA | AL137487.1 | - | 1 - 2 | 222 | 79 | |
BC112214.1 | - | 1 - 2 | 197 | 79 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC12001042.hg.1
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | PSR12013696.hg.1 | ENST00000376678.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR12013696.hg.1 | NR_026670.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR12013696.hg.1 | TC12001042.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR12013697.hg.1 | ENST00000376678.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR12013697.hg.1 | NR_026670.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR12013697.hg.1 | TC12001042.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | TC1200009425.hg.1 | ENST00000376678.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009425.hg.1 | ENST00000376678.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009425.hg.1 | ENST00000376678.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009425.hg.1 | ENST00000376678.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009425.hg.1 | ENST00000376678.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009425.hg.1 | ENST00000376678.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009425.hg.1 | ENST00000376678.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009425.hg.1 | ENST00000376678.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009425.hg.1 | ENST00000376678.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009425.hg.1 | ENST00000376678.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009425.hg.1 | ENST00000376678.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009425.hg.1 | ENST00000376678.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009425.hg.1 | ENST00000376678.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009425.hg.1 | ENST00000376678.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009425.hg.1 | ENST00000376678.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009425.hg.1 | ENST00000376678.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009425.hg.1 | ENST00000536191.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009425.hg.1 | ENST00000536191.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009425.hg.1 | ENST00000536191.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009425.hg.1 | ENST00000536191.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009425.hg.1 | NR_026670.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200009425.hg.1 | NR_026670.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200009425.hg.1 | NR_026670.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200009425.hg.1 | NR_026670.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200009425.hg.1 | NR_026670.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200009425.hg.1 | NR_026670.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200009425.hg.1 | NR_026670.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200009425.hg.1 | NR_026670.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200012475.hg.1 | ENST00000535133.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200012475.hg.1 | ENST00000535133.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200012475.hg.1 | ENST00000535133.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1200170184.hg.1 | ENST00000376678.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1200170184.hg.1 | ENST00000376678.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1200170184.hg.1 | NR_026670.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR1200170184.hg.1 | ENST00000376678.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1200170184.hg.1 | ENST00000376678.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1200170184.hg.1 | NR_026670.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR1200170184.hg.1 | TC1200009425.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | TC1200009425.hg.1 | ENST00000376678.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009425.hg.1 | ENST00000376678.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009425.hg.1 | ENST00000376678.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009425.hg.1 | ENST00000376678.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009425.hg.1 | ENST00000376678.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009425.hg.1 | ENST00000376678.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009425.hg.1 | ENST00000376678.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009425.hg.1 | ENST00000376678.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009425.hg.1 | ENST00000376678.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009425.hg.1 | ENST00000376678.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009425.hg.1 | ENST00000376678.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009425.hg.1 | ENST00000376678.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009425.hg.1 | ENST00000376678.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009425.hg.1 | ENST00000376678.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009425.hg.1 | ENST00000376678.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009425.hg.1 | ENST00000376678.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009425.hg.1 | ENST00000536191.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009425.hg.1 | ENST00000536191.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009425.hg.1 | ENST00000536191.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009425.hg.1 | ENST00000536191.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009425.hg.1 | NR_026670.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1200009425.hg.1 | NR_026670.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1200009425.hg.1 | NR_026670.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1200009425.hg.1 | NR_026670.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1200009425.hg.1 | NR_026670.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1200009425.hg.1 | NR_026670.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1200009425.hg.1 | NR_026670.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1200009425.hg.1 | NR_026670.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1200012475.hg.1 | ENST00000535133.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200012475.hg.1 | ENST00000535133.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200012475.hg.1 | ENST00000535133.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1200170183.hg.1 | ENST00000376678.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1200170183.hg.1 | ENST00000376678.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1200170183.hg.1 | NR_026670.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR1200170183.hg.1 | ENST00000376678.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1200170183.hg.1 | ENST00000376678.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1200170183.hg.1 | NR_026670.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR1200170183.hg.1 | TC1200009425.hg.1 | Clariom D TC |
