Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007677.hg.1 |
ENST00000295896.12 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007677.hg.1 |
ENST00000295896.12 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007677.hg.1 |
ENST00000295896.12 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007677.hg.1 |
ENST00000295896.12 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007677.hg.1 |
ENST00000295896.12 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007677.hg.1 |
ENST00000295896.12 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007677.hg.1 |
ENST00000295896.12 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007677.hg.1 |
ENST00000295896.12 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007677.hg.1 |
ENST00000295896.12 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007677.hg.1 |
ENST00000491896.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007677.hg.1 |
ENST00000491896.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007677.hg.1 |
ENST00000491896.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007677.hg.1 |
ENST00000491896.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007677.hg.1 |
ENST00000491896.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007677.hg.1 |
ENST00000616659.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007677.hg.1 |
ENST00000616659.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007677.hg.1 |
ENST00000616659.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007677.hg.1 |
ENST00000616659.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007677.hg.1 |
ENST00000616659.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007677.hg.1 |
ENST00000616659.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007677.hg.1 |
NR_026866.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007677.hg.1 |
NR_026866.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007677.hg.1 |
NR_026866.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007677.hg.1 |
NR_026866.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007677.hg.1 |
NR_026866.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007677.hg.1 |
NR_026866.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007677.hg.1 |
NR_026866.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007677.hg.1 |
NR_026866.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007677.hg.1 |
NR_026866.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR03008462.hg.1 |
TC03000392.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR03008462.hg.1 |
ENST00000295896.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR03008462.hg.1 |
NR_026866.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300007677.hg.1 |
ENST00000295896.12 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007677.hg.1 |
ENST00000295896.12 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007677.hg.1 |
ENST00000295896.12 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007677.hg.1 |
ENST00000295896.12 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007677.hg.1 |
ENST00000295896.12 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007677.hg.1 |
ENST00000295896.12 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007677.hg.1 |
ENST00000295896.12 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007677.hg.1 |
ENST00000295896.12 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007677.hg.1 |
ENST00000295896.12 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007677.hg.1 |
ENST00000491896.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007677.hg.1 |
ENST00000491896.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007677.hg.1 |
ENST00000491896.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007677.hg.1 |
ENST00000491896.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007677.hg.1 |
ENST00000491896.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007677.hg.1 |
ENST00000616659.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007677.hg.1 |
ENST00000616659.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007677.hg.1 |
ENST00000616659.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007677.hg.1 |
ENST00000616659.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007677.hg.1 |
ENST00000616659.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007677.hg.1 |
ENST00000616659.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007677.hg.1 |
NR_026866.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300007677.hg.1 |
NR_026866.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300007677.hg.1 |
NR_026866.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300007677.hg.1 |
NR_026866.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300007677.hg.1 |
NR_026866.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300007677.hg.1 |
NR_026866.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300007677.hg.1 |
NR_026866.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300007677.hg.1 |
NR_026866.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300007677.hg.1 |
NR_026866.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300155483.hg.1 |
ENST00000295896.12 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300155483.hg.1 |
ENST00000491896.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300155483.hg.1 |
ENST00000616659.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300155483.hg.1 |
NR_026866.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300155483.hg.1 |
ENST00000295896.12 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300155483.hg.1 |
ENST00000491896.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300155483.hg.1 |
ENST00000616659.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300155483.hg.1 |
NR_026866.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300155483.hg.1 |
TC0300007677.hg.1 |
Clariom D TC |