Assay Details
| Gene Symbol: CMTR1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_015050.2 | NP_055865.1 | 8 - 9 | 1032 | 66 | |
| XM_005248955.3 | XP_005249012.1 | 8 - 9 | 962 | 66 | ||
| GenBank mRNA | AK292564.1 | - | 8 - 9 | 886 | 66 | |
| BC031890.1 | - | 8 - 9 | 926 | 4944457 | 66 | |
| D43949.2 | - | 8 - 9 | 922 | 66 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC06000545.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR06006002.hg.1 | TC06000545.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR06006002.hg.1 | ENST00000373427.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR06006002.hg.1 | ENST00000373451.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR06006002.hg.1 | NM_015050.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR06006003.hg.1 | TC06000545.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR06006003.hg.1 | ENST00000373427.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR06006003.hg.1 | ENST00000373451.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR06006003.hg.1 | NM_015050.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007869.hg.1 | ENST00000373451.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007869.hg.1 | ENST00000373451.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007869.hg.1 | ENST00000373451.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007869.hg.1 | ENST00000373451.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007869.hg.1 | ENST00000373451.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007869.hg.1 | ENST00000373451.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007869.hg.1 | ENST00000373451.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007869.hg.1 | ENST00000373451.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007869.hg.1 | ENST00000373451.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007869.hg.1 | ENST00000373451.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007869.hg.1 | ENST00000373451.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007869.hg.1 | ENST00000373451.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007869.hg.1 | ENST00000373451.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007869.hg.1 | ENST00000373451.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007869.hg.1 | ENST00000373451.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007869.hg.1 | ENST00000373451.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007869.hg.1 | ENST00000373451.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007869.hg.1 | ENST00000373451.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007869.hg.1 | ENST00000373451.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007869.hg.1 | ENST00000373451.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007869.hg.1 | ENST00000373451.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007869.hg.1 | ENST00000373451.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007869.hg.1 | ENST00000373451.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007869.hg.1 | ENST00000373451.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007869.hg.1 | ENST00000373451.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007869.hg.1 | ENST00000373451.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007869.hg.1 | ENST00000373451.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007869.hg.1 | ENST00000373451.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007869.hg.1 | ENST00000373451.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007869.hg.1 | ENST00000373451.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007869.hg.1 | ENST00000455891.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007869.hg.1 | ENST00000455891.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007869.hg.1 | ENST00000455891.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007869.hg.1 | ENST00000455891.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007869.hg.1 | ENST00000455891.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007869.hg.1 | ENST00000455891.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007869.hg.1 | ENST00000455891.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007869.hg.1 | ENST00000455891.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007869.hg.1 | ENST00000455891.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007869.hg.1 | ENST00000455891.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007869.hg.1 | ENST00000455891.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007869.hg.1 | ENST00000455891.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007869.hg.1 | ENST00000457419.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007869.hg.1 | ENST00000457419.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007869.hg.1 | ENST00000457419.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007869.hg.1 | ENST00000457419.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007869.hg.1 | ENST00000457419.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007869.hg.1 | ENST00000457419.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007869.hg.1 | ENST00000457419.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007869.hg.1 | ENST00000471097.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007869.hg.1 | ENST00000471097.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007869.hg.1 | ENST00000471097.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007869.hg.1 | ENST00000471097.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007869.hg.1 | ENST00000475364.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007869.hg.1 | ENST00000475364.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007869.hg.1 | ENST00000475364.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007869.hg.1 | ENST00000475364.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007869.hg.1 | ENST00000475364.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007869.hg.1 | ENST00000475364.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007869.hg.1 | ENST00000475364.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007869.hg.1 | ENST00000475364.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007869.hg.1 | ENST00000493656.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007869.hg.1 | ENST00000493656.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007869.hg.1 | ENST00000493656.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007869.hg.1 | ENST00000493656.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007869.hg.1 | NM_015050.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007869.hg.1 | NM_015050.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007869.hg.1 | NM_015050.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007869.hg.1 | NM_015050.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007869.hg.1 | NM_015050.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007869.hg.1 | NM_015050.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007869.hg.1 | NM_015050.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007869.hg.1 | NM_015050.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007869.hg.1 | NM_015050.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007869.hg.1 | NM_015050.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007869.hg.1 | NM_015050.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007869.hg.1 | NM_015050.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007869.hg.1 | NM_015050.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007869.hg.1 | NM_015050.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007869.hg.1 | NM_015050.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007869.hg.1 | NM_015050.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007869.hg.1 | NM_015050.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007869.hg.1 | NM_015050.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007869.hg.1 | NM_015050.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007869.hg.1 | NM_015050.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007869.hg.1 | NM_015050.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007869.hg.1 | NM_015050.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007869.hg.1 | NM_015050.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007869.hg.1 | NM_015050.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007869.hg.1 | NM_015050.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007869.hg.1 | NM_015050.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007869.hg.1 | NM_015050.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007869.hg.1 | NM_015050.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007869.hg.1 | NM_015050.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007869.hg.1 | NM_015050.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007869.hg.1 | NM_015050.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600154705.hg.1 | ENST00000373451.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600154705.hg.1 | ENST00000455891.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600154705.hg.1 | NM_015050.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600154705.hg.1 | ENST00000373451.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600154705.hg.1 | ENST00000455891.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600154705.hg.1 | NM_015050.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600154705.hg.1 | TC0600007869.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0600007869.hg.1 | ENST00000373451.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007869.hg.1 | ENST00000373451.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007869.hg.1 | ENST00000373451.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007869.hg.1 | ENST00000373451.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007869.hg.1 | ENST00000373451.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007869.hg.1 | ENST00000373451.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007869.hg.1 | ENST00000373451.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007869.hg.1 | ENST00000373451.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007869.hg.1 | ENST00000373451.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007869.hg.1 | ENST00000373451.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007869.hg.1 | ENST00000373451.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007869.hg.1 | ENST00000373451.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007869.hg.1 | ENST00000373451.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007869.hg.1 | ENST00000373451.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007869.hg.1 | ENST00000373451.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007869.hg.1 | ENST00000373451.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007869.hg.1 | ENST00000373451.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007869.hg.1 | ENST00000373451.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007869.hg.1 | ENST00000373451.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007869.hg.1 | ENST00000373451.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007869.hg.1 | ENST00000373451.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007869.hg.1 | ENST00000373451.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007869.hg.1 | ENST00000373451.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007869.hg.1 | ENST00000373451.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007869.hg.1 | ENST00000373451.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007869.hg.1 | ENST00000373451.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007869.hg.1 | ENST00000373451.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007869.hg.1 | ENST00000373451.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007869.hg.1 | ENST00000373451.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007869.hg.1 | ENST00000373451.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007869.hg.1 | ENST00000455891.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007869.hg.1 | ENST00000455891.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007869.hg.1 | ENST00000455891.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007869.hg.1 | ENST00000455891.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007869.hg.1 | ENST00000455891.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007869.hg.1 | ENST00000455891.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007869.hg.1 | ENST00000455891.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007869.hg.1 | ENST00000455891.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007869.hg.1 | ENST00000455891.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007869.hg.1 | ENST00000455891.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007869.hg.1 | ENST00000455891.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007869.hg.1 | ENST00000455891.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007869.hg.1 | ENST00000457419.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007869.hg.1 | ENST00000457419.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007869.hg.1 | ENST00000457419.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007869.hg.1 | ENST00000457419.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007869.hg.1 | ENST00000457419.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007869.hg.1 | ENST00000457419.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007869.hg.1 | ENST00000457419.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007869.hg.1 | ENST00000471097.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007869.hg.1 | ENST00000471097.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007869.hg.1 | ENST00000471097.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007869.hg.1 | ENST00000471097.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007869.hg.1 | ENST00000475364.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007869.hg.1 | ENST00000475364.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007869.hg.1 | ENST00000475364.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007869.hg.1 | ENST00000475364.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007869.hg.1 | ENST00000475364.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007869.hg.1 | ENST00000475364.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007869.hg.1 | ENST00000475364.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007869.hg.1 | ENST00000475364.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007869.hg.1 | ENST00000493656.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007869.hg.1 | ENST00000493656.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007869.hg.1 | ENST00000493656.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007869.hg.1 | ENST00000493656.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007869.hg.1 | NM_015050.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600007869.hg.1 | NM_015050.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600007869.hg.1 | NM_015050.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600007869.hg.1 | NM_015050.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600007869.hg.1 | NM_015050.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600007869.hg.1 | NM_015050.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600007869.hg.1 | NM_015050.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600007869.hg.1 | NM_015050.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600007869.hg.1 | NM_015050.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600007869.hg.1 | NM_015050.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600007869.hg.1 | NM_015050.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600007869.hg.1 | NM_015050.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600007869.hg.1 | NM_015050.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600007869.hg.1 | NM_015050.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600007869.hg.1 | NM_015050.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600007869.hg.1 | NM_015050.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600007869.hg.1 | NM_015050.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600007869.hg.1 | NM_015050.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600007869.hg.1 | NM_015050.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600007869.hg.1 | NM_015050.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600007869.hg.1 | NM_015050.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600007869.hg.1 | NM_015050.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600007869.hg.1 | NM_015050.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600007869.hg.1 | NM_015050.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600007869.hg.1 | NM_015050.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600007869.hg.1 | NM_015050.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600007869.hg.1 | NM_015050.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600007869.hg.1 | NM_015050.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600007869.hg.1 | NM_015050.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600007869.hg.1 | NM_015050.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600007869.hg.1 | NM_015050.2 | Refseq |

CMTR1
Genomic Map