Assay Details
| Gene Symbol: SETD1A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_014712.2 | NP_055527.1 | 3 - 4 | 934 | 82 | |
| XM_005255723.1 | XP_005255780.1 | 3 - 4 | 418 | 82 | ||
| XM_006721106.3 | XP_006721169.1 | 3 - 4 | 339 | 82 | ||
| XM_017023909.1 | XP_016879398.1 | 3 - 4 | 1571 | 82 | ||
| GenBank mRNA | AB002337.2 | - | 3 - 4 | 934 | 82 | |
| BC035795.2 | - | 3 - 4 | 434 | 82 | ||
More Information
Set Membership: |
|

SETD1A
Genomic Map