Assay Details
| Gene Symbol: NWD2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001144990.1 | NP_001138462.1 | 7 | 2528 | 64 | |
| GenBank mRNA | AB033065.2 | - | 1 | 796 | 64 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

NWD2
Genomic Map