Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR21006814.hg.1 |
ENST00000291691.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR21006814.hg.1 |
ENST00000397682.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR21006814.hg.1 |
ENST00000397685.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR21006814.hg.1 |
NM_058180.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR21006814.hg.1 |
TC21000556.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR21006815.hg.1 |
ENST00000291691.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR21006815.hg.1 |
ENST00000397680.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR21006815.hg.1 |
ENST00000397682.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR21006815.hg.1 |
ENST00000397685.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR21006815.hg.1 |
NM_058180.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR21006815.hg.1 |
TC21000556.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR21006812.hg.1 |
ENST00000291691.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR21006812.hg.1 |
ENST00000397682.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR21006812.hg.1 |
ENST00000397685.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR21006812.hg.1 |
NM_058180.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR21006812.hg.1 |
TC21000556.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR21006806.hg.1 |
ENST00000291691.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR21006806.hg.1 |
ENST00000397679.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR21006806.hg.1 |
ENST00000397680.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR21006806.hg.1 |
ENST00000397682.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR21006806.hg.1 |
ENST00000397683.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR21006806.hg.1 |
ENST00000397685.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR21006806.hg.1 |
NM_058180.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR21006806.hg.1 |
TC21000556.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR2100157479.hg.1 |
TC2100008467.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR2100157479.hg.1 |
ENST00000291691.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2100157479.hg.1 |
ENST00000397679.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2100157479.hg.1 |
ENST00000397680.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2100157479.hg.1 |
ENST00000397682.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2100157479.hg.1 |
ENST00000397683.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2100157479.hg.1 |
ENST00000417060.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2100157479.hg.1 |
ENST00000445935.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2100157479.hg.1 |
ENST00000475776.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2100157479.hg.1 |
ENST00000491666.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2100157479.hg.1 |
NM_001286462.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2100157479.hg.1 |
NM_001286463.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2100157479.hg.1 |
NM_001286476.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2100157479.hg.1 |
NM_001286477.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2100157479.hg.1 |
NM_058180.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2100157479.hg.1 |
ENST00000291691.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2100157479.hg.1 |
ENST00000397679.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2100157479.hg.1 |
ENST00000397680.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2100157479.hg.1 |
ENST00000397682.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2100157479.hg.1 |
ENST00000397683.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2100157479.hg.1 |
ENST00000417060.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2100157479.hg.1 |
ENST00000445935.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2100157479.hg.1 |
ENST00000475776.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2100157479.hg.1 |
ENST00000491666.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2100157479.hg.1 |
NM_001286462.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2100157479.hg.1 |
NM_001286463.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2100157479.hg.1 |
NM_001286476.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2100157479.hg.1 |
NM_001286477.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2100157479.hg.1 |
NM_058180.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2100157486.hg.1 |
TC2100008467.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR2100157486.hg.1 |
ENST00000291691.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2100157486.hg.1 |
ENST00000397682.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2100157486.hg.1 |
ENST00000445935.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2100157486.hg.1 |
ENST00000475776.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2100157486.hg.1 |
NM_001286476.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2100157486.hg.1 |
NM_058180.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2100157486.hg.1 |
ENST00000291691.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2100157486.hg.1 |
ENST00000397682.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2100157486.hg.1 |
ENST00000445935.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2100157486.hg.1 |
ENST00000475776.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2100157486.hg.1 |
NM_001286476.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2100157486.hg.1 |
NM_058180.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2100157487.hg.1 |
TC2100008467.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR2100157487.hg.1 |
ENST00000291691.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2100157487.hg.1 |
ENST00000397682.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2100157487.hg.1 |
ENST00000445935.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2100157487.hg.1 |
ENST00000475776.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2100157487.hg.1 |
NM_001286476.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2100157487.hg.1 |
NM_058180.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2100157487.hg.1 |
ENST00000291691.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2100157487.hg.1 |
ENST00000397682.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2100157487.hg.1 |
ENST00000445935.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2100157487.hg.1 |
ENST00000475776.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2100157487.hg.1 |
NM_001286476.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2100157487.hg.1 |
NM_058180.4 |
Refseq |