Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X006880.hg.1 |
TC0X000514.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X006880.hg.1 |
ENST00000337685.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X006880.hg.1 |
ENST00000357175.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X006880.hg.1 |
ENST00000372552.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X006880.hg.1 |
NM_001171020.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X006880.hg.1 |
NM_152423.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X006878.hg.1 |
TC0X000514.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X006878.hg.1 |
ENST00000337685.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X006878.hg.1 |
ENST00000357175.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X006878.hg.1 |
ENST00000372552.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X006878.hg.1 |
NM_001171020.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X006878.hg.1 |
NM_152423.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008040.hg.1 |
ENST00000337685.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008040.hg.1 |
ENST00000337685.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008040.hg.1 |
ENST00000337685.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008040.hg.1 |
ENST00000337685.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008040.hg.1 |
ENST00000337685.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008040.hg.1 |
ENST00000337685.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008040.hg.1 |
ENST00000337685.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008040.hg.1 |
ENST00000337685.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008040.hg.1 |
ENST00000357175.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008040.hg.1 |
ENST00000357175.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008040.hg.1 |
ENST00000357175.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008040.hg.1 |
ENST00000357175.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008040.hg.1 |
ENST00000357175.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008040.hg.1 |
ENST00000357175.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008040.hg.1 |
ENST00000357175.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008040.hg.1 |
ENST00000372552.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008040.hg.1 |
ENST00000372552.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008040.hg.1 |
ENST00000372552.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008040.hg.1 |
ENST00000372552.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008040.hg.1 |
ENST00000372552.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008040.hg.1 |
ENST00000372552.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008040.hg.1 |
ENST00000619455.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008040.hg.1 |
ENST00000619455.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008040.hg.1 |
ENST00000619455.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008040.hg.1 |
ENST00000619455.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008040.hg.1 |
ENST00000619455.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008040.hg.1 |
NM_001171020.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008040.hg.1 |
NM_001171020.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008040.hg.1 |
NM_001171020.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008040.hg.1 |
NM_001171020.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008040.hg.1 |
NM_001171020.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008040.hg.1 |
NM_001171020.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008040.hg.1 |
NM_001171020.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008040.hg.1 |
NM_152423.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008040.hg.1 |
NM_152423.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008040.hg.1 |
NM_152423.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008040.hg.1 |
NM_152423.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008040.hg.1 |
NM_152423.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008040.hg.1 |
NM_152423.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008040.hg.1 |
NM_152423.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008040.hg.1 |
NM_152423.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X006877.hg.1 |
TC0X000514.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X006877.hg.1 |
ENST00000337685.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X006877.hg.1 |
ENST00000357175.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X006877.hg.1 |
ENST00000372552.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X006877.hg.1 |
NM_001171020.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X006877.hg.1 |
NM_152423.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00154981.hg.1 |
ENST00000337685.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00154981.hg.1 |
ENST00000357175.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00154981.hg.1 |
ENST00000372552.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00154981.hg.1 |
ENST00000619455.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00154981.hg.1 |
NM_001171020.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00154981.hg.1 |
NM_152423.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00154981.hg.1 |
ENST00000337685.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00154981.hg.1 |
ENST00000357175.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00154981.hg.1 |
ENST00000372552.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00154981.hg.1 |
ENST00000619455.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00154981.hg.1 |
NM_001171020.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00154981.hg.1 |
NM_152423.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00154981.hg.1 |
TC0X00008040.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0X00154983.hg.1 |
ENST00000337685.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00154983.hg.1 |
ENST00000357175.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00154983.hg.1 |
ENST00000372552.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00154983.hg.1 |
ENST00000619455.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00154983.hg.1 |
NM_001171020.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00154983.hg.1 |
NM_152423.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00154983.hg.1 |
ENST00000337685.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00154983.hg.1 |
ENST00000357175.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00154983.hg.1 |
ENST00000372552.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00154983.hg.1 |
ENST00000619455.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00154983.hg.1 |
NM_001171020.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00154983.hg.1 |
NM_152423.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00154983.hg.1 |
TC0X00008040.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0X00008040.hg.1 |
ENST00000337685.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008040.hg.1 |
ENST00000337685.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008040.hg.1 |
ENST00000337685.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008040.hg.1 |
ENST00000337685.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008040.hg.1 |
ENST00000337685.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008040.hg.1 |
ENST00000337685.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008040.hg.1 |
ENST00000337685.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008040.hg.1 |
ENST00000337685.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008040.hg.1 |
ENST00000357175.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008040.hg.1 |
ENST00000357175.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008040.hg.1 |
ENST00000357175.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008040.hg.1 |
ENST00000357175.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008040.hg.1 |
ENST00000357175.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008040.hg.1 |
ENST00000357175.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008040.hg.1 |
ENST00000357175.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008040.hg.1 |
ENST00000372552.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008040.hg.1 |
ENST00000372552.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008040.hg.1 |
ENST00000372552.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008040.hg.1 |
ENST00000372552.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008040.hg.1 |
ENST00000372552.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008040.hg.1 |
ENST00000372552.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008040.hg.1 |
ENST00000619455.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008040.hg.1 |
ENST00000619455.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008040.hg.1 |
ENST00000619455.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008040.hg.1 |
ENST00000619455.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008040.hg.1 |
ENST00000619455.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008040.hg.1 |
NM_001171020.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00008040.hg.1 |
NM_001171020.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00008040.hg.1 |
NM_001171020.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00008040.hg.1 |
NM_001171020.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00008040.hg.1 |
NM_001171020.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00008040.hg.1 |
NM_001171020.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00008040.hg.1 |
NM_001171020.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00008040.hg.1 |
NM_152423.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00008040.hg.1 |
NM_152423.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00008040.hg.1 |
NM_152423.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00008040.hg.1 |
NM_152423.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00008040.hg.1 |
NM_152423.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00008040.hg.1 |
NM_152423.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00008040.hg.1 |
NM_152423.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00008040.hg.1 |
NM_152423.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00154982.hg.1 |
ENST00000337685.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00154982.hg.1 |
ENST00000357175.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00154982.hg.1 |
ENST00000372552.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00154982.hg.1 |
ENST00000619455.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00154982.hg.1 |
NM_001171020.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00154982.hg.1 |
NM_152423.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00154982.hg.1 |
ENST00000337685.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00154982.hg.1 |
ENST00000357175.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00154982.hg.1 |
ENST00000372552.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00154982.hg.1 |
ENST00000619455.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00154982.hg.1 |
NM_001171020.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00154982.hg.1 |
NM_152423.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00154982.hg.1 |
TC0X00008040.hg.1 |
Clariom D TC |