Assay Details
| Gene Symbol: CEP83-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_027035.1 | 1 | 250 | 71 | ||
| GenBank mRNA | AK057269.1 | - | 1 | 250 | 71 | |
| BC035086.2 | - | 1 | 233 | 71 | ||
| BC044616.1 | - | 1 | 234 | 71 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC12000731.hg.1
Set Membership: |
|
MicroArray Probes:
Agilent
Affymetrix
ArrayStar
ABI
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P08544550 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P08544557 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P28977956 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P28977955 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P08544554 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P28977960 | - | - |
| Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M | 831139 | - | - |
| Agilent Whole Human Genome Microarray 4x44K v2 | A_33_P3374997 | - | - |
| Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k | GT_44k_32_P209148 | - | - |
| Agilent Whole Human Genome Oligo Microarray 4x44K | 15723 | - | - |
| Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray | 20490 | - | - |
| Agilent Human Genome CGH + SNP Microarray | A_16_P02679623 | - | - |
| Agilent Whole Human Genome Microarray 4x44K v2 | A_33_P3374997 | - | - |
| SurePrint G3 Human GE Microarray G4851A | A_33_P3374997 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P08544544 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28977944 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P08544536 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P08544549 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28977945 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28977949 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P16582415 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P08544535 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28977946 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28977950 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28977948 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28977946 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P08544536 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P08544549 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28977948 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28977950 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28977949 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P08544544 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28977944 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P08544535 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P16582415 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28977945 | - | - |
| Whole Human Genome Oligo Microarray G4112A | A_32_P209148 | - | - |
| Agilent Whole Human Genome Microarray 4x44K G4112F | A_32_P209148 | - | - |
| Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_18_P11562909 | - | - |
| Agilent Human 1B Microarray G4111A | A_32_P803811 | - | - |
| Agilent Human 1B Microarray G4111A | A_32_P209148 | - | - |
| Agilent Human 1B Microarray G4111A | A_32_P195553 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P08544545 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P08544549 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P08544543 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P16582413 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P08544552 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P08544537 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P08544547 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P08544535 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P08544551 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P08544539 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P08544554 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P08544556 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P08544558 | - | - |
| Agilent SurePrint G3 Human CGH Microarray 2x400K | A_16_P02679623 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix HT HG-U133+ PM Array Plate | 237021_PM_at | - | - |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008491.hg.1 | ENST00000623122.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008491.hg.1 | NR_027035.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008491.hg.1 | NR_027035.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR12009308.hg.1 | NR_027035.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR12009308.hg.1 | TC12000731.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | TC1200008491.hg.1 | ENST00000623122.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008491.hg.1 | NR_027035.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200008491.hg.1 | NR_027035.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200162236.hg.1 | ENST00000623122.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200162236.hg.1 | NR_027035.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200162236.hg.1 | ENST00000623122.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200162236.hg.1 | NR_027035.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200162236.hg.1 | TC1200008491.hg.1 | Clariom D TC |
| Human Genome U133 Plus 2.0 Array | 237021_at | - | - |
| Human Genome U133 Plus 2.0 Array | 237021_PM_at | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 237021_at6 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 237021_at10 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 237021_at3 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 237021_at2 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 237021_at9 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 237021_at4 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 237021_at5 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 237021_at11 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 237021_at8 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 237021_at7 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 237021_at1 | - | - |
| Human Genome U95 Set | 66051_PM_at | - | - |
| Human Genome U95 Set | 66051_at | - | - |
| Human Genome U95 Set | 75379_PM_at | - | - |
| Human Genome U95 Set | 75379_at | - | - |
| Human Genome U133 Set | 237021_at | - | - |
| Human Genome U133 Set | 237021_PM_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Arraystar Human LncRNA microarray V2.0 | ASHG19A3A048926 | - | - |
| Arraystar Human LncRNA microarray V2.0 | ASHG19A3A048926 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Human Genome Survey Microarray Version 2.0 | 232947 | - | - |

CEP83-AS1
Genomic Map