Assay Details
Gene Symbol: ZNF565 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001042474.2 | NP_001035939.1 | 2 - 3 | 272 | 73 | |
NM_152477.4 | NP_689690.3 | 2 - 3 | 127 | 73 | ||
XM_011526512.2 | XP_011524814.1 | 2 - 3 | 340 | 73 | ||
XM_011526514.2 | XP_011524816.1 | 2 - 3 | 206 | 73 | ||
XM_017026341.1 | XP_016881830.1 | 3 - 4 | 815 | 73 | ||
GenBank mRNA | AK057290.1 | - | 2 - 3 | 859 | 73 | |
AK094310.1 | - | 2 - 3 | 269 | 73 | ||
BC068453.1 | - | 2 - 3 | 109 | 30343899 | 73 | |
CB960081.1 | - | 2 - 3 | 136 | 73 | ||
DA473136.1 | - | 2 - 3 | 127 | 73 | ||
DA766324.1 | - | 2 - 3 | 272 | 73 | ||
DB473200.1 | - | 2 - 3 | 120 | 73 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC19001464.hg.1
Additional Information:
This assay is in the 5'UTR region of the AK094310.1,BC068453.1,NM_001042474.2,NM_152477.4,XM_011526512.2,XM_011526514.2,XM_017026341.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC1900010517.hg.1 | ENST00000304116.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900010517.hg.1 | ENST00000304116.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900010517.hg.1 | ENST00000304116.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900010517.hg.1 | ENST00000304116.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900010517.hg.1 | ENST00000304116.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900010517.hg.1 | ENST00000304116.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900010517.hg.1 | ENST00000304116.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900010517.hg.1 | ENST00000304116.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900010517.hg.1 | ENST00000304116.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900010517.hg.1 | ENST00000304116.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900010517.hg.1 | ENST00000355114.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900010517.hg.1 | ENST00000355114.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900010517.hg.1 | ENST00000355114.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900010517.hg.1 | ENST00000355114.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900010517.hg.1 | ENST00000355114.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900010517.hg.1 | ENST00000355114.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900010517.hg.1 | ENST00000355114.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900010517.hg.1 | ENST00000355114.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900010517.hg.1 | ENST00000363309.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900010517.hg.1 | ENST00000392173.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900010517.hg.1 | ENST00000392173.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900010517.hg.1 | ENST00000392173.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900010517.hg.1 | ENST00000392173.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900010517.hg.1 | ENST00000392173.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900010517.hg.1 | ENST00000392173.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900010517.hg.1 | ENST00000392173.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900010517.hg.1 | ENST00000392173.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900010517.hg.1 | ENST00000585994.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900010517.hg.1 | ENST00000585994.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900010517.hg.1 | ENST00000585994.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900010517.hg.1 | ENST00000585994.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900010517.hg.1 | ENST00000585994.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900010517.hg.1 | ENST00000585994.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900010517.hg.1 | ENST00000586648.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900010517.hg.1 | ENST00000586648.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900010517.hg.1 | ENST00000586648.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900010517.hg.1 | ENST00000586648.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900010517.hg.1 | ENST00000587803.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900010517.hg.1 | ENST00000587803.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900010517.hg.1 | ENST00000587803.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900010517.hg.1 | ENST00000587803.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900010517.hg.1 | ENST00000591473.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900010517.hg.1 | ENST00000591473.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900010517.hg.1 | ENST00000591473.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900010517.hg.1 | ENST00000591473.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900010517.hg.1 | ENST00000591473.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900010517.hg.1 | ENST00000591473.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900010517.hg.1 | NM_001042474.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900010517.hg.1 | NM_001042474.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900010517.hg.1 | NM_001042474.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900010517.hg.1 | NM_001042474.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900010517.hg.1 | NM_001042474.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900010517.hg.1 | NM_001042474.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900010517.hg.1 | NM_001042474.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900010517.hg.1 | NM_001042474.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900010517.hg.1 | NM_001042474.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900010517.hg.1 | NM_001042474.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900010517.hg.1 | NM_152477.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900010517.hg.1 | NM_152477.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900010517.hg.1 | NM_152477.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900010517.hg.1 | NM_152477.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900010517.hg.1 | NM_152477.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900010517.hg.1 | NM_152477.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900010517.hg.1 | NM_152477.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900010517.hg.1 | NM_152477.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900010517.hg.1 | NM_152477.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900010517.hg.1 | NM_152477.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR19019886.hg.1 | ENST00000304116.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR19019886.hg.1 | NM_152477.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR19019886.hg.1 | TC19001464.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR19019884.hg.1 | ENST00000304116.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR19019884.hg.1 | ENST00000355114.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR19019884.hg.1 | ENST00000392173.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR19019884.hg.1 | NM_001042474.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR19019884.hg.1 | NM_152477.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR19019884.hg.1 | TC19001464.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR19019888.hg.1 | ENST00000392173.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR19019888.hg.1 | NM_001042474.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR19019888.hg.1 | TC19001464.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR19019885.hg.1 | ENST00000304116.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR19019885.hg.1 | ENST00000355114.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR19019885.hg.1 | ENST00000392173.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR19019885.hg.1 | NM_001042474.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR19019885.hg.1 | NM_152477.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR19019885.hg.1 | TC19001464.hg.1 | HTA TC |
Affymetrix Human Clariom D Assay | PSR1900186272.hg.1 | ENST00000304116.9 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1900186272.hg.1 | ENST00000586648.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1900186272.hg.1 | ENST00000587803.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1900186272.hg.1 | ENST00000591473.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1900186272.hg.1 | NM_152477.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR1900186272.hg.1 | TC1900010517.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR1900186272.hg.1 | ENST00000304116.9 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1900186272.hg.1 | ENST00000586648.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1900186272.hg.1 | ENST00000587803.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1900186272.hg.1 | ENST00000591473.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1900186272.hg.1 | NM_152477.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR1900186276.hg.1 | ENST00000392173.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1900186276.hg.1 | NM_001042474.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR1900186276.hg.1 | TC1900010517.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR1900186276.hg.1 | ENST00000392173.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1900186276.hg.1 | NM_001042474.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1900010517.hg.1 | ENST00000304116.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900010517.hg.1 | ENST00000304116.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900010517.hg.1 | ENST00000304116.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900010517.hg.1 | ENST00000304116.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900010517.hg.1 | ENST00000304116.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900010517.hg.1 | ENST00000304116.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900010517.hg.1 | ENST00000304116.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900010517.hg.1 | ENST00000304116.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900010517.hg.1 | ENST00000304116.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900010517.hg.1 | ENST00000304116.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900010517.hg.1 | ENST00000355114.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900010517.hg.1 | ENST00000355114.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900010517.hg.1 | ENST00000355114.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900010517.hg.1 | ENST00000355114.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900010517.hg.1 | ENST00000355114.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900010517.hg.1 | ENST00000355114.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900010517.hg.1 | ENST00000355114.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900010517.hg.1 | ENST00000355114.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900010517.hg.1 | ENST00000363309.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900010517.hg.1 | ENST00000392173.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900010517.hg.1 | ENST00000392173.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900010517.hg.1 | ENST00000392173.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900010517.hg.1 | ENST00000392173.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900010517.hg.1 | ENST00000392173.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900010517.hg.1 | ENST00000392173.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900010517.hg.1 | ENST00000392173.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900010517.hg.1 | ENST00000392173.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900010517.hg.1 | ENST00000585994.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900010517.hg.1 | ENST00000585994.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900010517.hg.1 | ENST00000585994.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900010517.hg.1 | ENST00000585994.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900010517.hg.1 | ENST00000585994.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900010517.hg.1 | ENST00000585994.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900010517.hg.1 | ENST00000586648.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900010517.hg.1 | ENST00000586648.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900010517.hg.1 | ENST00000586648.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900010517.hg.1 | ENST00000586648.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900010517.hg.1 | ENST00000587803.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900010517.hg.1 | ENST00000587803.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900010517.hg.1 | ENST00000587803.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900010517.hg.1 | ENST00000587803.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900010517.hg.1 | ENST00000591473.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900010517.hg.1 | ENST00000591473.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900010517.hg.1 | ENST00000591473.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900010517.hg.1 | ENST00000591473.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900010517.hg.1 | ENST00000591473.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900010517.hg.1 | ENST00000591473.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900010517.hg.1 | NM_001042474.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1900010517.hg.1 | NM_001042474.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1900010517.hg.1 | NM_001042474.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1900010517.hg.1 | NM_001042474.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1900010517.hg.1 | NM_001042474.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1900010517.hg.1 | NM_001042474.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1900010517.hg.1 | NM_001042474.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1900010517.hg.1 | NM_001042474.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1900010517.hg.1 | NM_001042474.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1900010517.hg.1 | NM_001042474.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1900010517.hg.1 | NM_152477.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1900010517.hg.1 | NM_152477.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1900010517.hg.1 | NM_152477.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1900010517.hg.1 | NM_152477.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1900010517.hg.1 | NM_152477.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1900010517.hg.1 | NM_152477.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1900010517.hg.1 | NM_152477.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1900010517.hg.1 | NM_152477.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1900010517.hg.1 | NM_152477.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1900010517.hg.1 | NM_152477.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR1900186264.hg.1 | ENST00000304116.9 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1900186264.hg.1 | ENST00000355114.9 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1900186264.hg.1 | ENST00000392173.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1900186264.hg.1 | ENST00000585994.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1900186264.hg.1 | ENST00000586648.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1900186264.hg.1 | ENST00000587803.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1900186264.hg.1 | ENST00000591473.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1900186264.hg.1 | NM_001042474.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR1900186264.hg.1 | NM_152477.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR1900186264.hg.1 | TC1900010517.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR1900186264.hg.1 | ENST00000304116.9 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1900186264.hg.1 | ENST00000355114.9 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1900186264.hg.1 | ENST00000392173.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1900186264.hg.1 | ENST00000585994.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1900186264.hg.1 | ENST00000586648.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1900186264.hg.1 | ENST00000587803.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1900186264.hg.1 | ENST00000591473.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1900186264.hg.1 | NM_001042474.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR1900186264.hg.1 | NM_152477.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR1900186266.hg.1 | ENST00000304116.9 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1900186266.hg.1 | ENST00000355114.9 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1900186266.hg.1 | ENST00000392173.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1900186266.hg.1 | ENST00000585994.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1900186266.hg.1 | ENST00000586648.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1900186266.hg.1 | NM_001042474.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR1900186266.hg.1 | NM_152477.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR1900186266.hg.1 | TC1900010517.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR1900186266.hg.1 | ENST00000304116.9 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1900186266.hg.1 | ENST00000355114.9 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1900186266.hg.1 | ENST00000392173.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1900186266.hg.1 | ENST00000585994.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1900186266.hg.1 | ENST00000586648.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1900186266.hg.1 | NM_001042474.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR1900186266.hg.1 | NM_152477.4 | Refseq |
