Assay Details
Gene Symbol: PPM1K | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | BC041350.1 | - | 1 | 173 | 86 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
This assay is in the 5'UTR region of the BC041350.1 transcripts.
This assay is in the 5'UTR region of the BC041350.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC0300010508.hg.1 | ENST00000390967.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000009147.hg.1 | ENST00000365375.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000008751.hg.1 | ENST00000517185.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400011283.hg.1 | ENST00000295908.11 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400011283.hg.1 | ENST00000295908.11 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400011283.hg.1 | ENST00000295908.11 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400011283.hg.1 | ENST00000295908.11 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400011283.hg.1 | ENST00000295908.11 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400011283.hg.1 | ENST00000295908.11 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400011283.hg.1 | ENST00000295908.11 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400011283.hg.1 | ENST00000295908.11 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400011283.hg.1 | ENST00000295908.11 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400011283.hg.1 | ENST00000295908.11 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400011283.hg.1 | ENST00000363599.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400011283.hg.1 | ENST00000505022.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400011283.hg.1 | ENST00000505022.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400011283.hg.1 | ENST00000505022.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400011283.hg.1 | ENST00000505022.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400011283.hg.1 | ENST00000506423.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400011283.hg.1 | ENST00000506423.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400011283.hg.1 | ENST00000506423.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400011283.hg.1 | ENST00000506423.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400011283.hg.1 | ENST00000506423.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400011283.hg.1 | ENST00000506423.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400011283.hg.1 | ENST00000506423.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400011283.hg.1 | ENST00000506423.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400011283.hg.1 | ENST00000508256.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400011283.hg.1 | ENST00000508256.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400011283.hg.1 | ENST00000508256.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400011283.hg.1 | ENST00000508256.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400011283.hg.1 | ENST00000508256.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400011283.hg.1 | ENST00000508256.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400011283.hg.1 | ENST00000508256.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400011283.hg.1 | ENST00000509340.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400011283.hg.1 | ENST00000509340.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400011283.hg.1 | ENST00000509340.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400011283.hg.1 | ENST00000510548.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400011283.hg.1 | ENST00000510548.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400011283.hg.1 | ENST00000510548.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400011283.hg.1 | ENST00000510548.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400011283.hg.1 | ENST00000510548.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400011283.hg.1 | ENST00000511506.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400011283.hg.1 | ENST00000511506.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400011283.hg.1 | ENST00000511506.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400011283.hg.1 | ENST00000513546.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400011283.hg.1 | ENST00000513546.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400011283.hg.1 | ENST00000513546.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400011283.hg.1 | ENST00000513546.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400011283.hg.1 | ENST00000513546.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400011283.hg.1 | ENST00000513546.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400011283.hg.1 | ENST00000513546.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400011283.hg.1 | ENST00000513546.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400011283.hg.1 | ENST00000513546.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400011283.hg.1 | ENST00000513546.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400011283.hg.1 | ENST00000513546.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400011283.hg.1 | ENST00000513546.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400011283.hg.1 | ENST00000514204.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400011283.hg.1 | ENST00000514204.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400011283.hg.1 | ENST00000514204.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400011283.hg.1 | ENST00000514204.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400011283.hg.1 | ENST00000514204.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400011283.hg.1 | ENST00000514204.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400011283.hg.1 | ENST00000514204.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400011283.hg.1 | ENST00000514204.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400011283.hg.1 | ENST00000514204.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400011283.hg.1 | ENST00000514204.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400011283.hg.1 | ENST00000514204.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400011283.hg.1 | ENST00000514204.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400011283.hg.1 | ENST00000608933.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400011283.hg.1 | ENST00000608933.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400011283.hg.1 | ENST00000608933.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400011283.hg.1 | ENST00000608933.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400011283.hg.1 | ENST00000608933.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400011283.hg.1 | ENST00000608933.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400011283.hg.1 | ENST00000608933.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400011283.hg.1 | ENST00000608933.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400011283.hg.1 | ENST00000608933.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400011283.hg.1 | ENST00000608933.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400011283.hg.1 | ENST00000608933.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400011283.hg.1 | ENST00000608933.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400011283.hg.1 | ENST00000608933.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400011283.hg.1 | ENST00000608933.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400011283.hg.1 | ENST00000608933.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400011283.hg.1 | ENST00000608933.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400011283.hg.1 | NM_152542.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400011283.hg.1 | NM_152542.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400011283.hg.1 | NM_152542.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400011283.hg.1 | NM_152542.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400011283.hg.1 | NM_152542.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400011283.hg.1 | NM_152542.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400011283.hg.1 | NM_152542.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400011283.hg.1 | NM_152542.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400011283.hg.1 | NM_152542.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400011283.hg.1 | NM_152542.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400011283.hg.1 | NM_152542.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400011283.hg.1 | NM_152542.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400011283.hg.1 | NM_152542.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400011283.hg.1 | NM_152542.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400011283.hg.1 | NM_152542.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400011283.hg.1 | NM_152542.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400011283.hg.1 | NM_152542.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400011283.hg.1 | NM_152542.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100008759.hg.1 | ENST00000384272.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008759.hg.1 | ENST00000384272.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000008751.hg.1 | ENST00000517185.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000009147.hg.1 | ENST00000365375.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400011283.hg.1 | ENST00000295908.11 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400011283.hg.1 | ENST00000295908.11 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400011283.hg.1 | ENST00000295908.11 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400011283.hg.1 | ENST00000295908.11 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400011283.hg.1 | ENST00000295908.11 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400011283.hg.1 | ENST00000295908.11 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400011283.hg.1 | ENST00000295908.11 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400011283.hg.1 | ENST00000295908.11 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400011283.hg.1 | ENST00000295908.11 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400011283.hg.1 | ENST00000295908.11 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400011283.hg.1 | ENST00000363599.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400011283.hg.1 | ENST00000505022.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400011283.hg.1 | ENST00000505022.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400011283.hg.1 | ENST00000505022.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400011283.hg.1 | ENST00000505022.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400011283.hg.1 | ENST00000506423.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400011283.hg.1 | ENST00000506423.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400011283.hg.1 | ENST00000506423.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400011283.hg.1 | ENST00000506423.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400011283.hg.1 | ENST00000506423.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400011283.hg.1 | ENST00000506423.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400011283.hg.1 | ENST00000506423.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400011283.hg.1 | ENST00000506423.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400011283.hg.1 | ENST00000508256.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400011283.hg.1 | ENST00000508256.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400011283.hg.1 | ENST00000508256.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400011283.hg.1 | ENST00000508256.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400011283.hg.1 | ENST00000508256.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400011283.hg.1 | ENST00000508256.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400011283.hg.1 | ENST00000508256.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400011283.hg.1 | ENST00000509340.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400011283.hg.1 | ENST00000509340.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400011283.hg.1 | ENST00000509340.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400011283.hg.1 | ENST00000510548.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400011283.hg.1 | ENST00000510548.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400011283.hg.1 | ENST00000510548.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400011283.hg.1 | ENST00000510548.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400011283.hg.1 | ENST00000510548.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400011283.hg.1 | ENST00000511506.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400011283.hg.1 | ENST00000511506.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400011283.hg.1 | ENST00000511506.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400011283.hg.1 | ENST00000513546.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400011283.hg.1 | ENST00000513546.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400011283.hg.1 | ENST00000513546.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400011283.hg.1 | ENST00000513546.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400011283.hg.1 | ENST00000513546.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400011283.hg.1 | ENST00000513546.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400011283.hg.1 | ENST00000513546.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400011283.hg.1 | ENST00000513546.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400011283.hg.1 | ENST00000513546.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400011283.hg.1 | ENST00000513546.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400011283.hg.1 | ENST00000513546.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400011283.hg.1 | ENST00000513546.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400011283.hg.1 | ENST00000514204.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400011283.hg.1 | ENST00000514204.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400011283.hg.1 | ENST00000514204.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400011283.hg.1 | ENST00000514204.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400011283.hg.1 | ENST00000514204.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400011283.hg.1 | ENST00000514204.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400011283.hg.1 | ENST00000514204.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400011283.hg.1 | ENST00000514204.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400011283.hg.1 | ENST00000514204.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400011283.hg.1 | ENST00000514204.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400011283.hg.1 | ENST00000514204.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400011283.hg.1 | ENST00000514204.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400011283.hg.1 | ENST00000608933.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400011283.hg.1 | ENST00000608933.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400011283.hg.1 | ENST00000608933.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400011283.hg.1 | ENST00000608933.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400011283.hg.1 | ENST00000608933.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400011283.hg.1 | ENST00000608933.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400011283.hg.1 | ENST00000608933.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400011283.hg.1 | ENST00000608933.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400011283.hg.1 | ENST00000608933.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400011283.hg.1 | ENST00000608933.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400011283.hg.1 | ENST00000608933.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400011283.hg.1 | ENST00000608933.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400011283.hg.1 | ENST00000608933.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400011283.hg.1 | ENST00000608933.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400011283.hg.1 | ENST00000608933.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400011283.hg.1 | ENST00000608933.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400011283.hg.1 | NM_152542.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0400011283.hg.1 | NM_152542.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0400011283.hg.1 | NM_152542.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0400011283.hg.1 | NM_152542.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0400011283.hg.1 | NM_152542.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0400011283.hg.1 | NM_152542.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0400011283.hg.1 | NM_152542.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0400011283.hg.1 | NM_152542.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0400011283.hg.1 | NM_152542.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0400011283.hg.1 | NM_152542.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0400011283.hg.1 | NM_152542.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0400011283.hg.1 | NM_152542.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0400011283.hg.1 | NM_152542.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0400011283.hg.1 | NM_152542.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0400011283.hg.1 | NM_152542.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0400011283.hg.1 | NM_152542.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0400011283.hg.1 | NM_152542.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0400011283.hg.1 | NM_152542.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0300010508.hg.1 | ENST00000390967.1 | Ensembl |
