Assay Details
| Gene Symbol: LINC01096 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK058104.1 | - | 1 - 2 | 224 | 58 | |
More Information
Set Membership: |
|
MicroArray Probes:
Agilent
Affymetrix
ArrayStar
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Human Genome Microarray 4x44K v2 | A_24_P892494 | - | - |
| Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k | GT_44k_24_P892494 | - | - |
| Agilent Whole Human Genome Oligo Microarray 4x44K | 4242 | - | - |
| Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray | 53091 | - | - |
| Agilent Whole Human Genome Microarray 4x44K v2 | A_24_P892494 | - | - |
| SurePrint G3 Human GE Microarray G4851A | A_24_P892494 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P15547118 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02936346 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P23088750 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P23088758 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P23088733 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P23088759 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P23088757 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P23088756 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P23088751 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P15547121 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P23088742 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02936342 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P23088762 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02936343 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P15547123 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P15547119 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02936348 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P23088758 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P23088750 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P15547121 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P23088757 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P23088762 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P15547119 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P23088759 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02936346 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P15547123 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P23088756 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02936348 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02936343 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02936342 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P23088733 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P15547118 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P23088751 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P23088742 | - | - |
| Whole Human Genome Oligo Microarray G4112A | A_24_P892494 | - | - |
| Agilent Whole Human Genome Microarray 4x44K G4112F | A_24_P892494 | - | - |
| Agilent Human 1B Microarray G4111A | A_32_P527973 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P15547088 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P15547125 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P02936317 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P02936330 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P15547105 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P02936297 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P02936267 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P15547097 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P02936295 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P02936302 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P02936291 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P02936284 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P02936307 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P15547118 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P15547111 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P02936300 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P02936312 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P02936272 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P15547099 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P15547117 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P02936286 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P15547128 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P02936327 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P02936305 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P02936321 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P15547091 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P15547103 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P02936345 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P02936326 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P02936348 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P02936283 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P02936292 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P15547119 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P02936268 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P02936333 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P02936350 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P02936279 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P15547093 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P02936332 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P02936331 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P15547123 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P02936288 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P02936323 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P02936310 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P02936282 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P15547131 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P15547121 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P02936276 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P02936274 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P02936281 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P15547109 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P02936265 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P02936290 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P15547112 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix HT HG-U133+ PM Array Plate | 236112_PM_at | - | - |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010084.hg.1 | ENST00000501050.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010084.hg.1 | ENST00000501050.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010084.hg.1 | ENST00000501050.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010084.hg.1 | ENST00000501050.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010084.hg.1 | ENST00000501050.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010084.hg.1 | ENST00000503938.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010084.hg.1 | ENST00000503938.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010084.hg.1 | NR_015450.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010084.hg.1 | NR_015450.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010084.hg.1 | NR_015450.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010084.hg.1 | NR_015450.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010084.hg.1 | NR_015450.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010084.hg.1 | NR_015450.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400010084.hg.1 | ENST00000501050.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400010084.hg.1 | ENST00000501050.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400010084.hg.1 | ENST00000501050.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400010084.hg.1 | ENST00000501050.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400010084.hg.1 | ENST00000501050.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400010084.hg.1 | ENST00000503938.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400010084.hg.1 | ENST00000503938.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400010084.hg.1 | NR_015450.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400010084.hg.1 | NR_015450.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400010084.hg.1 | NR_015450.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400010084.hg.1 | NR_015450.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400010084.hg.1 | NR_015450.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400010084.hg.1 | NR_015450.1 | Refseq |
| Human Genome U133 Plus 2.0 Array | 236112_PM_at | - | - |
| Human Genome U133 Plus 2.0 Array | 236112_at | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 236112_at6 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 236112_at5 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 236112_at2 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 236112_at10 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 236112_at3 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 236112_at4 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 236112_at7 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 236112_at8 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 236112_at11 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 236112_at1 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 236112_at9 | - | - |
| Human Genome U95 Set | 54460_PM_at | - | - |
| Human Genome U95 Set | 54460_at | - | - |
| Human Genome U133 Set | 236112_PM_at | - | - |
| Human Genome U133 Set | 236112_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Arraystar Human LncRNA microarray V2.0 | ASHG19A3A023965 | - | - |
| Arraystar Human LncRNA microarray V2.0 | ASHG19A3A023965 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Human Genome Survey Microarray Version 2.0 | 504106 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| HumanHT-12 v4 Expression BeadChip | ILMN_3237884 | BC048110.1 | human_transcriptome |
| HumanHT-12 v4 Expression BeadChip | ILMN_3237884 | BC053955.1 | human_transcriptome |
| HumanHT-12 v4 Expression BeadChip | ILMN_3237884 | NR_015450.1 | human_transcriptome |
| HumanHT-12 v4 Expression BeadChip | ILMN_3237884 | AK058104.1 | human_transcriptome |
| HumanHT-12 v4 Expression BeadChip | ILMN_3237884 | BC035174.2 | human_transcriptome |
| HumanHT-12 v4 Expression BeadChip | ILMN_1849228 | BC048110.1 | human_transcriptome |
| HumanHT-12 v4 Expression BeadChip | ILMN_1849228 | BC053955.1 | human_transcriptome |
| HumanHT-12 v4 Expression BeadChip | ILMN_1849228 | NR_015450.1 | human_transcriptome |
| HumanHT-12 v4 Expression BeadChip | ILMN_1849228 | AK058104.1 | human_transcriptome |
| HumanHT-12 v4 Expression BeadChip | ILMN_1849228 | BC035174.2 | human_transcriptome |
| HumanHT-12 V4.0 expression beadchip | ILMN_1849228 | BC048110.1 | human_transcriptome |
| HumanHT-12 V4.0 expression beadchip | ILMN_1849228 | BC053955.1 | human_transcriptome |
| HumanHT-12 V4.0 expression beadchip | ILMN_1849228 | NR_015450.1 | human_transcriptome |
| HumanHT-12 V4.0 expression beadchip | ILMN_1849228 | AK058104.1 | human_transcriptome |
| HumanHT-12 V4.0 expression beadchip | ILMN_1849228 | BC035174.2 | human_transcriptome |
| HumanHT-12 V4.0 expression beadchip | ILMN_3237884 | BC048110.1 | human_transcriptome |
| HumanHT-12 V4.0 expression beadchip | ILMN_3237884 | BC053955.1 | human_transcriptome |
| HumanHT-12 V4.0 expression beadchip | ILMN_3237884 | NR_015450.1 | human_transcriptome |
| HumanHT-12 V4.0 expression beadchip | ILMN_3237884 | AK058104.1 | human_transcriptome |
| HumanHT-12 V4.0 expression beadchip | ILMN_3237884 | BC035174.2 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_3237884 | BC048110.1 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_3237884 | BC053955.1 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_3237884 | NR_015450.1 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_3237884 | AK058104.1 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_3237884 | BC035174.2 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip | ILMN_3237884 | BC048110.1 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip | ILMN_3237884 | BC053955.1 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip | ILMN_3237884 | NR_015450.1 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip | ILMN_3237884 | AK058104.1 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip | ILMN_3237884 | BC035174.2 | human_transcriptome |
| Illumina human-6 v2.0 expression beadchip | ILMN_1849228 | BC048110.1 | human_transcriptome |
| Illumina human-6 v2.0 expression beadchip | ILMN_1849228 | BC053955.1 | human_transcriptome |
| Illumina human-6 v2.0 expression beadchip | ILMN_1849228 | NR_015450.1 | human_transcriptome |
| Illumina human-6 v2.0 expression beadchip | ILMN_1849228 | AK058104.1 | human_transcriptome |
| Illumina human-6 v2.0 expression beadchip | ILMN_1849228 | BC035174.2 | human_transcriptome |
| Illumina Sentrix Human-6 v2 Expression BeadChip | 6660376 | - | - |
| Illumina HumanRef-6 v2.0 expression beadchip | ILMN_101255 | - | - |
| Illumina HumanWG-6 v3.0 expression beadchip | ILMN_1849228 | BC048110.1 | human_transcriptome |
| Illumina HumanWG-6 v3.0 expression beadchip | ILMN_1849228 | BC053955.1 | human_transcriptome |
| Illumina HumanWG-6 v3.0 expression beadchip | ILMN_1849228 | NR_015450.1 | human_transcriptome |
| Illumina HumanWG-6 v3.0 expression beadchip | ILMN_1849228 | AK058104.1 | human_transcriptome |
| Illumina HumanWG-6 v3.0 expression beadchip | ILMN_1849228 | BC035174.2 | human_transcriptome |
| Illumina HumanHT-12 V3.0 expression beadchip | ILMN_1849228 | BC048110.1 | human_transcriptome |
| Illumina HumanHT-12 V3.0 expression beadchip | ILMN_1849228 | BC053955.1 | human_transcriptome |
| Illumina HumanHT-12 V3.0 expression beadchip | ILMN_1849228 | NR_015450.1 | human_transcriptome |
| Illumina HumanHT-12 V3.0 expression beadchip | ILMN_1849228 | AK058104.1 | human_transcriptome |
| Illumina HumanHT-12 V3.0 expression beadchip | ILMN_1849228 | BC035174.2 | human_transcriptome |
| Illumina HumanWG-6_V2_0_R2 | ILMN_1849228 | BC048110.1 | human_transcriptome |
| Illumina HumanWG-6_V2_0_R2 | ILMN_1849228 | BC053955.1 | human_transcriptome |
| Illumina HumanWG-6_V2_0_R2 | ILMN_1849228 | NR_015450.1 | human_transcriptome |
| Illumina HumanWG-6_V2_0_R2 | ILMN_1849228 | AK058104.1 | human_transcriptome |
| Illumina HumanWG-6_V2_0_R2 | ILMN_1849228 | BC035174.2 | human_transcriptome |

LINC01096
Genomic Map