Assay Details
Gene Symbol: LOC101928794 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_111908.1 | 1 - 2 | 318 | 82 | ||
GenBank mRNA | AW003767.1 | - | 1 - 2 | 317 | 82 | |
BQ011784.1 | - | 1 - 2 | 444 | 82 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
