| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR12000371.hg.1 |
ENST00000322367.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR12000371.hg.1 |
ENST00000327702.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR12000371.hg.1 |
ENST00000335762.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR12000371.hg.1 |
ENST00000344100.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR12000371.hg.1 |
ENST00000347598.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR12000371.hg.1 |
ENST00000399591.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR12000371.hg.1 |
ENST00000399595.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR12000371.hg.1 |
ENST00000399597.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR12000371.hg.1 |
ENST00000399601.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR12000371.hg.1 |
ENST00000399603.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR12000371.hg.1 |
ENST00000399606.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR12000371.hg.1 |
ENST00000399617.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR12000371.hg.1 |
ENST00000399621.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR12000371.hg.1 |
ENST00000399629.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR12000371.hg.1 |
ENST00000399634.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR12000371.hg.1 |
ENST00000399637.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR12000371.hg.1 |
ENST00000399638.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR12000371.hg.1 |
ENST00000399641.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR12000371.hg.1 |
ENST00000399644.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR12000371.hg.1 |
ENST00000399649.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR12000371.hg.1 |
ENST00000399655.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR12000371.hg.1 |
ENST00000402845.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR12000371.hg.1 |
ENST00000406454.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR12000371.hg.1 |
NM_000719.6 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR12000371.hg.1 |
NM_001129827.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR12000371.hg.1 |
NM_001129829.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR12000371.hg.1 |
NM_001129830.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR12000371.hg.1 |
NM_001129831.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR12000371.hg.1 |
NM_001129832.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR12000371.hg.1 |
NM_001129833.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR12000371.hg.1 |
NM_001129834.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR12000371.hg.1 |
NM_001129835.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR12000371.hg.1 |
NM_001129836.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR12000371.hg.1 |
NM_001129837.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR12000371.hg.1 |
NM_001129838.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR12000371.hg.1 |
NM_001129839.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR12000371.hg.1 |
NM_001129840.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR12000371.hg.1 |
NM_001129841.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR12000371.hg.1 |
NM_001129842.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR12000371.hg.1 |
NM_001129843.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR12000371.hg.1 |
NM_001129844.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR12000371.hg.1 |
NM_001129846.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR12000371.hg.1 |
NM_001167623.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR12000371.hg.1 |
NM_001167624.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR12000371.hg.1 |
NM_001167625.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR12000371.hg.1 |
NM_199460.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR12000371.hg.1 |
TC12000025.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200006500.hg.1 |
ENST00000613818.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200006500.hg.1 |
ENST00000613818.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200006500.hg.1 |
ENST00000613818.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200006500.hg.1 |
ENST00000613818.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1200198559.hg.1 |
ENST00000327702.11 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1200198559.hg.1 |
ENST00000335762.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1200198559.hg.1 |
ENST00000344100.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1200198559.hg.1 |
ENST00000347598.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1200198559.hg.1 |
ENST00000399591.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1200198559.hg.1 |
ENST00000399595.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1200198559.hg.1 |
ENST00000399597.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1200198559.hg.1 |
ENST00000399601.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1200198559.hg.1 |
ENST00000399603.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1200198559.hg.1 |
ENST00000399606.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1200198559.hg.1 |
ENST00000399617.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1200198559.hg.1 |
ENST00000399621.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1200198559.hg.1 |
ENST00000399629.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1200198559.hg.1 |
ENST00000399634.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1200198559.hg.1 |
ENST00000399637.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1200198559.hg.1 |
ENST00000399638.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1200198559.hg.1 |
ENST00000399641.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1200198559.hg.1 |
ENST00000399644.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1200198559.hg.1 |
ENST00000399649.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1200198559.hg.1 |
ENST00000399655.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1200198559.hg.1 |
ENST00000402845.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1200198559.hg.1 |
ENST00000406454.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1200198559.hg.1 |
ENST00000613940.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1200198559.hg.1 |
ENST00000615303.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1200198559.hg.1 |
ENST00000615400.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1200198559.hg.1 |
ENST00000616390.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1200198559.hg.1 |
ENST00000620878.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1200198559.hg.1 |
ENST00000621625.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1200198559.hg.1 |
NM_000719.6 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1200198559.hg.1 |
NM_001129827.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1200198559.hg.1 |
NM_001129829.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1200198559.hg.1 |
NM_001129830.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1200198559.hg.1 |
NM_001129831.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1200198559.hg.1 |
NM_001129832.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1200198559.hg.1 |
NM_001129833.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1200198559.hg.1 |
NM_001129834.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1200198559.hg.1 |
NM_001129835.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1200198559.hg.1 |
NM_001129836.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1200198559.hg.1 |
NM_001129837.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1200198559.hg.1 |
NM_001129838.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1200198559.hg.1 |
NM_001129839.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1200198559.hg.1 |
NM_001129840.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1200198559.hg.1 |
NM_001129841.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1200198559.hg.1 |
NM_001129842.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1200198559.hg.1 |
NM_001129843.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1200198559.hg.1 |
NM_001129844.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1200198559.hg.1 |
NM_001129846.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1200198559.hg.1 |
NM_001167623.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1200198559.hg.1 |
NM_001167624.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1200198559.hg.1 |
NM_001167625.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1200198559.hg.1 |
NM_199460.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1200198559.hg.1 |
ENST00000327702.11 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1200198559.hg.1 |
ENST00000335762.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1200198559.hg.1 |
ENST00000344100.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1200198559.hg.1 |
ENST00000347598.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1200198559.hg.1 |
ENST00000399591.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1200198559.hg.1 |
ENST00000399595.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1200198559.hg.1 |
ENST00000399597.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1200198559.hg.1 |
ENST00000399601.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1200198559.hg.1 |
ENST00000399603.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1200198559.hg.1 |
ENST00000399606.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1200198559.hg.1 |
ENST00000399617.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1200198559.hg.1 |
ENST00000399621.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1200198559.hg.1 |
ENST00000399629.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1200198559.hg.1 |
ENST00000399634.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1200198559.hg.1 |
ENST00000399637.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1200198559.hg.1 |
ENST00000399638.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1200198559.hg.1 |
ENST00000399641.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1200198559.hg.1 |
ENST00000399644.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1200198559.hg.1 |
ENST00000399649.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1200198559.hg.1 |
ENST00000399655.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1200198559.hg.1 |
ENST00000402845.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1200198559.hg.1 |
ENST00000406454.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1200198559.hg.1 |
ENST00000613940.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1200198559.hg.1 |
ENST00000615303.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1200198559.hg.1 |
ENST00000615400.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1200198559.hg.1 |
ENST00000616390.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1200198559.hg.1 |
ENST00000620878.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1200198559.hg.1 |
ENST00000621625.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1200198559.hg.1 |
NM_000719.6 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1200198559.hg.1 |
NM_001129827.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1200198559.hg.1 |
NM_001129829.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1200198559.hg.1 |
NM_001129830.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1200198559.hg.1 |
NM_001129831.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1200198559.hg.1 |
NM_001129832.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1200198559.hg.1 |
NM_001129833.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1200198559.hg.1 |
NM_001129834.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1200198559.hg.1 |
NM_001129835.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1200198559.hg.1 |
NM_001129836.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1200198559.hg.1 |
NM_001129837.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1200198559.hg.1 |
NM_001129838.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1200198559.hg.1 |
NM_001129839.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1200198559.hg.1 |
NM_001129840.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1200198559.hg.1 |
NM_001129841.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1200198559.hg.1 |
NM_001129842.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1200198559.hg.1 |
NM_001129843.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1200198559.hg.1 |
NM_001129844.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1200198559.hg.1 |
NM_001129846.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1200198559.hg.1 |
NM_001167623.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1200198559.hg.1 |
NM_001167624.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1200198559.hg.1 |
NM_001167625.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1200198559.hg.1 |
NM_199460.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1200198559.hg.1 |
TC1200012570.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
TC1200006500.hg.1 |
ENST00000613818.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200006500.hg.1 |
ENST00000613818.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200006500.hg.1 |
ENST00000613818.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200006500.hg.1 |
ENST00000613818.1 |
Ensembl |