Assay Details
| Gene Symbol: CMKLR1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001142343.1 | NP_001135815.1 | 1 | 266 | 75 | |
| NM_004072.2 | NP_004063.1 | 1 | 266 | 75 | ||
| GenBank mRNA | BC106927.2 | - | 1 | 80 | 40033186 | 75 |
| BC106928.1 | - | 1 | 80 | 40033187 | 75 | |
| U79527.1 | - | 1 | 272 | 75 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC12001933.hg.1
Additional Information:
This assay is in the 5'UTR region of the BC106927.2,BC106928.1,NM_001142343.1,NM_004072.2,U79527.1 transcripts.
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
skeletal system development
chemotaxis
immune response
G-protein coupled receptor signaling pathway
positive regulation of macrophage chemotaxis
negative regulation of NF-kappaB transcription factor activity
negative regulation of interleukin-12 production
positive regulation of fat cell differentiation
regulation of calcium-mediated signaling
chemokine-mediated signaling pathway
receptor activity
G-protein coupled receptor activity
chemokine receptor activity
protein binding
skeletal system development
chemotaxis
immune response
G-protein coupled receptor signaling pathway
positive regulation of macrophage chemotaxis
negative regulation of NF-kappaB transcription factor activity
negative regulation of interleukin-12 production
positive regulation of fat cell differentiation
regulation of calcium-mediated signaling
chemokine-mediated signaling pathway
receptor activity
G-protein coupled receptor activity
chemokine receptor activity
protein binding
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR12025146.hg.1 | ENST00000312143.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR12025146.hg.1 | ENST00000397688.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR12025146.hg.1 | ENST00000550402.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR12025146.hg.1 | ENST00000552995.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR12025146.hg.1 | NM_001142343.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR12025146.hg.1 | NM_001142344.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR12025146.hg.1 | NM_004072.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR12025146.hg.1 | TC12001933.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR12025145.hg.1 | ENST00000312143.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR12025145.hg.1 | ENST00000397688.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR12025145.hg.1 | ENST00000550402.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR12025145.hg.1 | ENST00000552995.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR12025145.hg.1 | NM_001142343.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR12025145.hg.1 | NM_001142344.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR12025145.hg.1 | NM_004072.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR12025145.hg.1 | TC12001933.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011838.hg.1 | ENST00000312143.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011838.hg.1 | ENST00000312143.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011838.hg.1 | ENST00000312143.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011838.hg.1 | ENST00000312143.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011838.hg.1 | ENST00000312143.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011838.hg.1 | ENST00000312143.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011838.hg.1 | ENST00000312143.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011838.hg.1 | ENST00000312143.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011838.hg.1 | ENST00000312143.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011838.hg.1 | ENST00000412676.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011838.hg.1 | ENST00000412676.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011838.hg.1 | ENST00000412676.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011838.hg.1 | ENST00000412676.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011838.hg.1 | ENST00000412676.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011838.hg.1 | ENST00000412676.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011838.hg.1 | ENST00000412676.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011838.hg.1 | ENST00000412676.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011838.hg.1 | ENST00000549466.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011838.hg.1 | ENST00000549466.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011838.hg.1 | ENST00000549466.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011838.hg.1 | ENST00000550402.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011838.hg.1 | ENST00000550402.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011838.hg.1 | ENST00000550402.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011838.hg.1 | ENST00000550402.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011838.hg.1 | ENST00000550402.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011838.hg.1 | ENST00000550402.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011838.hg.1 | ENST00000550402.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011838.hg.1 | ENST00000550573.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011838.hg.1 | ENST00000550573.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011838.hg.1 | ENST00000550573.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011838.hg.1 | ENST00000552995.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011838.hg.1 | ENST00000552995.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011838.hg.1 | ENST00000552995.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011838.hg.1 | ENST00000552995.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011838.hg.1 | ENST00000552995.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011838.hg.1 | ENST00000552995.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011838.hg.1 | ENST00000552995.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011838.hg.1 | NM_001142343.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011838.hg.1 | NM_001142343.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011838.hg.1 | NM_001142343.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011838.hg.1 | NM_001142343.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011838.hg.1 | NM_001142343.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011838.hg.1 | NM_001142343.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011838.hg.1 | NM_001142343.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011838.hg.1 | NM_001142343.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011838.hg.1 | NM_001142343.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011838.hg.1 | NM_001142343.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011838.hg.1 | NM_001142344.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011838.hg.1 | NM_001142344.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011838.hg.1 | NM_001142344.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011838.hg.1 | NM_001142344.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011838.hg.1 | NM_001142344.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011838.hg.1 | NM_001142344.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011838.hg.1 | NM_001142344.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011838.hg.1 | NM_001142344.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011838.hg.1 | NM_001142344.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011838.hg.1 | NM_001142345.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011838.hg.1 | NM_001142345.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011838.hg.1 | NM_001142345.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011838.hg.1 | NM_001142345.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011838.hg.1 | NM_001142345.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011838.hg.1 | NM_001142345.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011838.hg.1 | NM_001142345.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011838.hg.1 | NM_001142345.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011838.hg.1 | NM_004072.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011838.hg.1 | NM_004072.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011838.hg.1 | NM_004072.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011838.hg.1 | NM_004072.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011838.hg.1 | NM_004072.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011838.hg.1 | NM_004072.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011838.hg.1 | NM_004072.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011838.hg.1 | NM_004072.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011838.hg.1 | NM_004072.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200191684.hg.1 | ENST00000312143.11 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200191684.hg.1 | ENST00000550402.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200191684.hg.1 | ENST00000552995.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200191684.hg.1 | NM_001142343.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200191684.hg.1 | NM_001142344.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200191684.hg.1 | NM_004072.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200191684.hg.1 | ENST00000312143.11 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200191684.hg.1 | ENST00000550402.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200191684.hg.1 | ENST00000552995.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200191684.hg.1 | NM_001142343.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200191684.hg.1 | NM_001142344.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200191684.hg.1 | NM_004072.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200191684.hg.1 | TC1200011838.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1200191685.hg.1 | ENST00000312143.11 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200191685.hg.1 | ENST00000550402.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200191685.hg.1 | ENST00000552995.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200191685.hg.1 | NM_001142343.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200191685.hg.1 | NM_001142344.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200191685.hg.1 | NM_004072.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200191685.hg.1 | ENST00000312143.11 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200191685.hg.1 | ENST00000550402.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200191685.hg.1 | ENST00000552995.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200191685.hg.1 | NM_001142343.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200191685.hg.1 | NM_001142344.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200191685.hg.1 | NM_004072.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200191685.hg.1 | TC1200011838.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC1200011838.hg.1 | ENST00000312143.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011838.hg.1 | ENST00000312143.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011838.hg.1 | ENST00000312143.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011838.hg.1 | ENST00000312143.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011838.hg.1 | ENST00000312143.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011838.hg.1 | ENST00000312143.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011838.hg.1 | ENST00000312143.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011838.hg.1 | ENST00000312143.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011838.hg.1 | ENST00000312143.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011838.hg.1 | ENST00000412676.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011838.hg.1 | ENST00000412676.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011838.hg.1 | ENST00000412676.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011838.hg.1 | ENST00000412676.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011838.hg.1 | ENST00000412676.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011838.hg.1 | ENST00000412676.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011838.hg.1 | ENST00000412676.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011838.hg.1 | ENST00000412676.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011838.hg.1 | ENST00000549466.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011838.hg.1 | ENST00000549466.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011838.hg.1 | ENST00000549466.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011838.hg.1 | ENST00000550402.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011838.hg.1 | ENST00000550402.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011838.hg.1 | ENST00000550402.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011838.hg.1 | ENST00000550402.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011838.hg.1 | ENST00000550402.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011838.hg.1 | ENST00000550402.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011838.hg.1 | ENST00000550402.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011838.hg.1 | ENST00000550573.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011838.hg.1 | ENST00000550573.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011838.hg.1 | ENST00000550573.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011838.hg.1 | ENST00000552995.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011838.hg.1 | ENST00000552995.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011838.hg.1 | ENST00000552995.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011838.hg.1 | ENST00000552995.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011838.hg.1 | ENST00000552995.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011838.hg.1 | ENST00000552995.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011838.hg.1 | ENST00000552995.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011838.hg.1 | NM_001142343.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200011838.hg.1 | NM_001142343.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200011838.hg.1 | NM_001142343.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200011838.hg.1 | NM_001142343.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200011838.hg.1 | NM_001142343.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200011838.hg.1 | NM_001142343.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200011838.hg.1 | NM_001142343.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200011838.hg.1 | NM_001142343.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200011838.hg.1 | NM_001142343.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200011838.hg.1 | NM_001142343.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200011838.hg.1 | NM_001142344.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200011838.hg.1 | NM_001142344.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200011838.hg.1 | NM_001142344.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200011838.hg.1 | NM_001142344.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200011838.hg.1 | NM_001142344.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200011838.hg.1 | NM_001142344.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200011838.hg.1 | NM_001142344.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200011838.hg.1 | NM_001142344.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200011838.hg.1 | NM_001142344.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200011838.hg.1 | NM_001142345.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200011838.hg.1 | NM_001142345.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200011838.hg.1 | NM_001142345.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200011838.hg.1 | NM_001142345.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200011838.hg.1 | NM_001142345.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200011838.hg.1 | NM_001142345.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200011838.hg.1 | NM_001142345.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200011838.hg.1 | NM_001142345.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200011838.hg.1 | NM_004072.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200011838.hg.1 | NM_004072.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200011838.hg.1 | NM_004072.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200011838.hg.1 | NM_004072.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200011838.hg.1 | NM_004072.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200011838.hg.1 | NM_004072.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200011838.hg.1 | NM_004072.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200011838.hg.1 | NM_004072.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200011838.hg.1 | NM_004072.2 | Refseq |

CMKLR1
Genomic Map