Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR19014960.hg.1 |
ENST00000245810.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR19014960.hg.1 |
ENST00000545374.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR19014960.hg.1 |
NM_004158.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR19014960.hg.1 |
TC19001093.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009424.hg.1 |
ENST00000245810.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009424.hg.1 |
ENST00000245810.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009424.hg.1 |
ENST00000597721.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009424.hg.1 |
ENST00000597721.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009424.hg.1 |
ENST00000597721.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009424.hg.1 |
ENST00000597721.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009424.hg.1 |
ENST00000597721.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009424.hg.1 |
NM_004158.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009424.hg.1 |
NM_004158.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR19014958.hg.1 |
ENST00000245810.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR19014958.hg.1 |
ENST00000545374.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR19014958.hg.1 |
NM_004158.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR19014958.hg.1 |
TC19001093.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR1900175580.hg.1 |
ENST00000245810.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900175580.hg.1 |
ENST00000597721.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900175580.hg.1 |
NM_004158.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900175580.hg.1 |
TC1900009424.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1900175580.hg.1 |
ENST00000245810.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900175580.hg.1 |
ENST00000597721.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900175580.hg.1 |
NM_004158.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900009424.hg.1 |
ENST00000245810.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900009424.hg.1 |
ENST00000245810.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900009424.hg.1 |
ENST00000597721.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900009424.hg.1 |
ENST00000597721.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900009424.hg.1 |
ENST00000597721.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900009424.hg.1 |
ENST00000597721.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900009424.hg.1 |
ENST00000597721.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900009424.hg.1 |
NM_004158.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900009424.hg.1 |
NM_004158.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900175578.hg.1 |
ENST00000245810.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900175578.hg.1 |
ENST00000597721.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900175578.hg.1 |
NM_004158.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900175578.hg.1 |
TC1900009424.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1900175578.hg.1 |
ENST00000245810.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900175578.hg.1 |
ENST00000597721.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900175578.hg.1 |
NM_004158.3 |
Refseq |