Assay Details
Gene Symbol: DDX10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_004398.3 | NP_004389.2 | 12 - 13 | 1624 | 72 | |
GenBank mRNA | AB040537.1 | - | 12 - 13 | 1560 | 72 | |
AB209884.1 | - | 12 - 13 | 1527 | 72 | ||
AK296752.1 | - | 10 - 11 | 1298 | 72 | ||
AK315216.1 | - | 12 - 13 | 1555 | 72 | ||
BC049217.1 | - | 12 - 13 | 1520 | 72 | ||
BC091521.1 | - | 12 - 13 | 1572 | 30388370 | 72 | |
BC093654.1 | - | 12 - 13 | 1528 | 7939499 | 72 | |
BC093656.1 | - | 12 - 13 | 1528 | 7939501 | 72 | |
U28042.1 | - | 12 - 13 | 1560 | 72 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC11000981.hg.1
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC1100008991.hg.1 | ENST00000322536.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100008991.hg.1 | ENST00000322536.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100008991.hg.1 | ENST00000322536.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100008991.hg.1 | ENST00000322536.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100008991.hg.1 | ENST00000322536.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100008991.hg.1 | ENST00000322536.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100008991.hg.1 | ENST00000322536.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100008991.hg.1 | ENST00000322536.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100008991.hg.1 | ENST00000322536.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100008991.hg.1 | ENST00000322536.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100008991.hg.1 | ENST00000322536.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100008991.hg.1 | ENST00000322536.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100008991.hg.1 | ENST00000322536.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100008991.hg.1 | ENST00000322536.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100008991.hg.1 | ENST00000322536.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100008991.hg.1 | ENST00000322536.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100008991.hg.1 | ENST00000322536.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100008991.hg.1 | ENST00000322536.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100008991.hg.1 | ENST00000322536.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100008991.hg.1 | ENST00000322536.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100008991.hg.1 | ENST00000322536.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100008991.hg.1 | ENST00000322536.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100008991.hg.1 | ENST00000322536.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100008991.hg.1 | ENST00000322536.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100008991.hg.1 | ENST00000524979.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100008991.hg.1 | ENST00000524979.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100008991.hg.1 | ENST00000524979.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100008991.hg.1 | ENST00000524979.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100008991.hg.1 | ENST00000524979.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100008991.hg.1 | ENST00000526794.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100008991.hg.1 | ENST00000526794.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100008991.hg.1 | ENST00000526794.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100008991.hg.1 | ENST00000526794.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100008991.hg.1 | ENST00000526794.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100008991.hg.1 | ENST00000526794.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100008991.hg.1 | ENST00000526794.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100008991.hg.1 | ENST00000526794.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100008991.hg.1 | ENST00000526794.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100008991.hg.1 | ENST00000526794.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100008991.hg.1 | ENST00000526794.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100008991.hg.1 | ENST00000526794.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100008991.hg.1 | ENST00000526794.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100008991.hg.1 | ENST00000526794.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100008991.hg.1 | ENST00000526794.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100008991.hg.1 | ENST00000526794.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100008991.hg.1 | ENST00000526794.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100008991.hg.1 | ENST00000526794.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100008991.hg.1 | ENST00000526794.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100008991.hg.1 | ENST00000526794.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100008991.hg.1 | ENST00000530116.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100008991.hg.1 | ENST00000530116.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100008991.hg.1 | ENST00000533198.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100008991.hg.1 | ENST00000533198.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100008991.hg.1 | ENST00000533198.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100008991.hg.1 | ENST00000534221.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100008991.hg.1 | ENST00000534221.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100008991.hg.1 | ENST00000534221.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100008991.hg.1 | ENST00000534221.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100008991.hg.1 | ENST00000534439.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100008991.hg.1 | ENST00000534439.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100008991.hg.1 | ENST00000534439.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100008991.hg.1 | ENST00000534439.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100008991.hg.1 | ENST00000534439.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100008991.hg.1 | ENST00000534774.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100008991.hg.1 | ENST00000534774.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100008991.hg.1 | ENST00000534774.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100008991.hg.1 | ENST00000534774.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100008991.hg.1 | ENST00000534774.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100008991.hg.1 | ENST00000534774.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100008991.hg.1 | NM_004398.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100008991.hg.1 | NM_004398.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100008991.hg.1 | NM_004398.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100008991.hg.1 | NM_004398.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100008991.hg.1 | NM_004398.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100008991.hg.1 | NM_004398.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100008991.hg.1 | NM_004398.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100008991.hg.1 | NM_004398.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100008991.hg.1 | NM_004398.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100008991.hg.1 | NM_004398.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100008991.hg.1 | NM_004398.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100008991.hg.1 | NM_004398.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100008991.hg.1 | NM_004398.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100008991.hg.1 | NM_004398.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100008991.hg.1 | NM_004398.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100008991.hg.1 | NM_004398.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100008991.hg.1 | NM_004398.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100008991.hg.1 | NM_004398.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100008991.hg.1 | NM_004398.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100008991.hg.1 | NM_004398.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100008991.hg.1 | NM_004398.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100008991.hg.1 | NM_004398.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100008991.hg.1 | NM_004398.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100008991.hg.1 | NM_004398.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR11012272.hg.1 | ENST00000322536.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR11012272.hg.1 | ENST00000456020.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR11012272.hg.1 | ENST00000526794.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR11012272.hg.1 | NM_004398.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR11012272.hg.1 | TC11000981.hg.1 | HTA TC |
Affymetrix Human Clariom D Assay | PSR1100166488.hg.1 | ENST00000322536.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100166488.hg.1 | ENST00000526794.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100166488.hg.1 | NM_004398.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR1100166488.hg.1 | ENST00000322536.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100166488.hg.1 | ENST00000526794.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100166488.hg.1 | NM_004398.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR1100166488.hg.1 | TC1100008991.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR1100166490.hg.1 | ENST00000322536.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100166490.hg.1 | ENST00000526794.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100166490.hg.1 | NM_004398.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR1100166490.hg.1 | ENST00000322536.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100166490.hg.1 | ENST00000526794.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100166490.hg.1 | NM_004398.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR1100166490.hg.1 | TC1100008991.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | TC1100008991.hg.1 | ENST00000322536.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100008991.hg.1 | ENST00000322536.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100008991.hg.1 | ENST00000322536.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100008991.hg.1 | ENST00000322536.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100008991.hg.1 | ENST00000322536.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100008991.hg.1 | ENST00000322536.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100008991.hg.1 | ENST00000322536.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100008991.hg.1 | ENST00000322536.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100008991.hg.1 | ENST00000322536.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100008991.hg.1 | ENST00000322536.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100008991.hg.1 | ENST00000322536.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100008991.hg.1 | ENST00000322536.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100008991.hg.1 | ENST00000322536.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100008991.hg.1 | ENST00000322536.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100008991.hg.1 | ENST00000322536.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100008991.hg.1 | ENST00000322536.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100008991.hg.1 | ENST00000322536.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100008991.hg.1 | ENST00000322536.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100008991.hg.1 | ENST00000322536.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100008991.hg.1 | ENST00000322536.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100008991.hg.1 | ENST00000322536.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100008991.hg.1 | ENST00000322536.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100008991.hg.1 | ENST00000322536.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100008991.hg.1 | ENST00000322536.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100008991.hg.1 | ENST00000524979.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100008991.hg.1 | ENST00000524979.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100008991.hg.1 | ENST00000524979.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100008991.hg.1 | ENST00000524979.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100008991.hg.1 | ENST00000524979.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100008991.hg.1 | ENST00000526794.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100008991.hg.1 | ENST00000526794.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100008991.hg.1 | ENST00000526794.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100008991.hg.1 | ENST00000526794.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100008991.hg.1 | ENST00000526794.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100008991.hg.1 | ENST00000526794.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100008991.hg.1 | ENST00000526794.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100008991.hg.1 | ENST00000526794.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100008991.hg.1 | ENST00000526794.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100008991.hg.1 | ENST00000526794.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100008991.hg.1 | ENST00000526794.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100008991.hg.1 | ENST00000526794.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100008991.hg.1 | ENST00000526794.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100008991.hg.1 | ENST00000526794.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100008991.hg.1 | ENST00000526794.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100008991.hg.1 | ENST00000526794.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100008991.hg.1 | ENST00000526794.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100008991.hg.1 | ENST00000526794.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100008991.hg.1 | ENST00000526794.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100008991.hg.1 | ENST00000526794.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100008991.hg.1 | ENST00000530116.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100008991.hg.1 | ENST00000530116.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100008991.hg.1 | ENST00000533198.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100008991.hg.1 | ENST00000533198.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100008991.hg.1 | ENST00000533198.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100008991.hg.1 | ENST00000534221.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100008991.hg.1 | ENST00000534221.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100008991.hg.1 | ENST00000534221.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100008991.hg.1 | ENST00000534221.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100008991.hg.1 | ENST00000534439.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100008991.hg.1 | ENST00000534439.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100008991.hg.1 | ENST00000534439.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100008991.hg.1 | ENST00000534439.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100008991.hg.1 | ENST00000534439.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100008991.hg.1 | ENST00000534774.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100008991.hg.1 | ENST00000534774.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100008991.hg.1 | ENST00000534774.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100008991.hg.1 | ENST00000534774.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100008991.hg.1 | ENST00000534774.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100008991.hg.1 | ENST00000534774.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100008991.hg.1 | NM_004398.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1100008991.hg.1 | NM_004398.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1100008991.hg.1 | NM_004398.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1100008991.hg.1 | NM_004398.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1100008991.hg.1 | NM_004398.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1100008991.hg.1 | NM_004398.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1100008991.hg.1 | NM_004398.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1100008991.hg.1 | NM_004398.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1100008991.hg.1 | NM_004398.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1100008991.hg.1 | NM_004398.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1100008991.hg.1 | NM_004398.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1100008991.hg.1 | NM_004398.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1100008991.hg.1 | NM_004398.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1100008991.hg.1 | NM_004398.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1100008991.hg.1 | NM_004398.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1100008991.hg.1 | NM_004398.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1100008991.hg.1 | NM_004398.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1100008991.hg.1 | NM_004398.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1100008991.hg.1 | NM_004398.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1100008991.hg.1 | NM_004398.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1100008991.hg.1 | NM_004398.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1100008991.hg.1 | NM_004398.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1100008991.hg.1 | NM_004398.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1100008991.hg.1 | NM_004398.3 | Refseq |
