Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR09014225.hg.1 |
TC09001063.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR09014225.hg.1 |
ENST00000340291.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR09014225.hg.1 |
ENST00000396638.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR09014225.hg.1 |
ENST00000479751.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR09014225.hg.1 |
ENST00000484764.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR09014225.hg.1 |
NM_001039592.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR09014225.hg.1 |
NM_172312.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900168612.hg.1 |
ENST00000340291.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900168612.hg.1 |
ENST00000396638.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900168612.hg.1 |
ENST00000460836.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900168612.hg.1 |
ENST00000463889.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900168612.hg.1 |
ENST00000471631.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900168612.hg.1 |
ENST00000472605.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900168612.hg.1 |
ENST00000475644.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900168612.hg.1 |
ENST00000479751.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900168612.hg.1 |
ENST00000484764.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900168612.hg.1 |
ENST00000495667.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900168612.hg.1 |
ENST00000497810.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900168612.hg.1 |
NM_001039592.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900168612.hg.1 |
NM_172312.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900168612.hg.1 |
ENST00000340291.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900168612.hg.1 |
ENST00000396638.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900168612.hg.1 |
ENST00000460836.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900168612.hg.1 |
ENST00000463889.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900168612.hg.1 |
ENST00000471631.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900168612.hg.1 |
ENST00000472605.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900168612.hg.1 |
ENST00000475644.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900168612.hg.1 |
ENST00000479751.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900168612.hg.1 |
ENST00000484764.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900168612.hg.1 |
ENST00000495667.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900168612.hg.1 |
ENST00000497810.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900168612.hg.1 |
NM_001039592.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900168612.hg.1 |
NM_172312.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900168612.hg.1 |
TC0900009956.hg.1 |
Clariom D TC |