Assay Details
| Gene Symbol: VAMP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_014232.2 | NP_055047.2 | 3 - 4 | 373 | 67 | |
| XM_005256775.4 | XP_005256832.1 | 3 - 4 | 703 | 67 | ||
| GenBank mRNA | AJ225044.1 | - | 2 - 3 | 277 | 67 | |
| AK021522.1 | - | 3 - 4 | 355 | 67 | ||
| AK225777.1 | - | 3 - 4 | 373 | 67 | ||
| AK289555.1 | - | 3 - 4 | 346 | 67 | ||
| BC002737.2 | - | 3 - 4 | 342 | 3628842 | 67 | |
| BC019608.1 | - | 3 - 4 | 355 | 4508990 | 67 | |
| BC033870.1 | - | 3 - 4 | 361 | 5203115 | 67 | |
More Information
Set Membership: |
|

VAMP2
Genomic Map