Assay Details
| Gene Symbol: FBXW11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_012300.2 | NP_036432.2 | 12 - 13 | 2027 | 92 | |
| NM_033644.2 | NP_387448.2 | 12 - 13 | 1988 | 92 | ||
| NM_033645.2 | NP_387449.2 | 11 - 12 | 1925 | 92 | ||
| XM_005265855.4 | XP_005265912.1 | 13 - 14 | 2077 | 92 | ||
| XM_005265856.2 | XP_005265913.1 | 14 - 15 | 2047 | 92 | ||
| XM_005265857.2 | XP_005265914.1 | 11 - 12 | 1585 | 92 | ||
| XM_005265858.2 | XP_005265915.1 | 13 - 14 | 2089 | 92 | ||
| XM_017009279.1 | XP_016864768.1 | 12 - 13 | 1887 | 92 | ||
| XM_017009280.1 | XP_016864769.1 | 13 - 14 | 1903 | 92 | ||
| XM_017009281.1 | XP_016864770.1 | 13 - 14 | 1971 | 92 | ||
| GenBank mRNA | AB014596.1 | - | 12 - 13 | 1681 | 92 | |
| AB033279.1 | - | 11 - 12 | 1655 | 92 | ||
| AB033280.1 | - | 12 - 13 | 1773 | 92 | ||
| AB033281.1 | - | 12 - 13 | 1795 | 92 | ||
| AK294956.1 | - | 12 - 13 | 1670 | 92 | ||
| BC026213.1 | - | 12 - 13 | 1988 | 4419029 | 92 | |
More Information
Set Membership: |
|

FBXW11
Genomic Map