| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500008883.hg.1 |
ENST00000239450.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500008883.hg.1 |
ENST00000239450.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500008883.hg.1 |
ENST00000239450.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500008883.hg.1 |
ENST00000239450.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500008883.hg.1 |
ENST00000239450.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500008883.hg.1 |
ENST00000239450.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500008883.hg.1 |
ENST00000239450.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500008883.hg.1 |
ENST00000239450.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500008883.hg.1 |
ENST00000622978.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500008883.hg.1 |
ENST00000622978.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500008883.hg.1 |
ENST00000622978.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500008883.hg.1 |
ENST00000622978.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500008883.hg.1 |
ENST00000622978.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500008883.hg.1 |
ENST00000624949.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500008883.hg.1 |
ENST00000624949.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500008883.hg.1 |
ENST00000624949.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500008883.hg.1 |
ENST00000624949.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500008883.hg.1 |
NM_018932.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500008883.hg.1 |
NM_018932.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500008883.hg.1 |
NM_018932.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500008883.hg.1 |
NM_018932.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500008883.hg.1 |
NM_018932.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500008883.hg.1 |
NM_018932.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500008883.hg.1 |
NM_018932.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500008880.hg.1 |
ENST00000231137.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500008880.hg.1 |
ENST00000231137.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500008880.hg.1 |
ENST00000231137.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500008880.hg.1 |
ENST00000231137.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500008880.hg.1 |
ENST00000231137.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500008880.hg.1 |
ENST00000231137.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500008880.hg.1 |
ENST00000231137.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500008880.hg.1 |
NM_018940.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500008880.hg.1 |
NM_018940.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500008880.hg.1 |
NM_018940.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500008880.hg.1 |
NM_018940.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500008880.hg.1 |
NM_018940.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500008880.hg.1 |
NM_018940.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500008880.hg.1 |
NM_018940.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0500008883.hg.1 |
ENST00000239450.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500008883.hg.1 |
ENST00000239450.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500008883.hg.1 |
ENST00000239450.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500008883.hg.1 |
ENST00000239450.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500008883.hg.1 |
ENST00000239450.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500008883.hg.1 |
ENST00000239450.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500008883.hg.1 |
ENST00000239450.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500008883.hg.1 |
ENST00000239450.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500008883.hg.1 |
ENST00000622978.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500008883.hg.1 |
ENST00000622978.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500008883.hg.1 |
ENST00000622978.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500008883.hg.1 |
ENST00000622978.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500008883.hg.1 |
ENST00000622978.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500008883.hg.1 |
ENST00000624949.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500008883.hg.1 |
ENST00000624949.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500008883.hg.1 |
ENST00000624949.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500008883.hg.1 |
ENST00000624949.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500008883.hg.1 |
NM_018932.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0500008883.hg.1 |
NM_018932.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0500008883.hg.1 |
NM_018932.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0500008883.hg.1 |
NM_018932.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0500008883.hg.1 |
NM_018932.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0500008883.hg.1 |
NM_018932.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0500008883.hg.1 |
NM_018932.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0500008880.hg.1 |
ENST00000231137.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500008880.hg.1 |
ENST00000231137.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500008880.hg.1 |
ENST00000231137.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500008880.hg.1 |
ENST00000231137.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500008880.hg.1 |
ENST00000231137.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500008880.hg.1 |
ENST00000231137.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500008880.hg.1 |
ENST00000231137.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500008880.hg.1 |
NM_018940.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0500008880.hg.1 |
NM_018940.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0500008880.hg.1 |
NM_018940.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0500008880.hg.1 |
NM_018940.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0500008880.hg.1 |
NM_018940.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0500008880.hg.1 |
NM_018940.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0500008880.hg.1 |
NM_018940.3 |
Refseq |