Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR05010795.hg.1 |
TC05000752.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR05010795.hg.1 |
NM_019119.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR05010798.hg.1 |
TC05000752.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR05010798.hg.1 |
NM_019119.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013245.hg.1 |
ENST00000316105.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013245.hg.1 |
ENST00000316105.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013245.hg.1 |
ENST00000316105.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013245.hg.1 |
ENST00000316105.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013245.hg.1 |
ENST00000316105.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013245.hg.1 |
ENST00000316105.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013245.hg.1 |
ENST00000316105.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013245.hg.1 |
ENST00000316105.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013245.hg.1 |
ENST00000316105.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013245.hg.1 |
ENST00000316105.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013245.hg.1 |
ENST00000623266.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013245.hg.1 |
ENST00000623266.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013245.hg.1 |
ENST00000623266.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013245.hg.1 |
ENST00000623266.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013245.hg.1 |
ENST00000623266.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013245.hg.1 |
ENST00000624909.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013245.hg.1 |
ENST00000624909.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013245.hg.1 |
ENST00000624909.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013245.hg.1 |
ENST00000624909.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013245.hg.1 |
NM_019119.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013245.hg.1 |
NM_019119.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013245.hg.1 |
NM_019119.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013245.hg.1 |
NM_019119.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013245.hg.1 |
NM_019119.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013245.hg.1 |
NM_019119.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013245.hg.1 |
NM_019119.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013245.hg.1 |
NM_019119.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013245.hg.1 |
NM_019119.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013245.hg.1 |
NM_019119.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500194380.hg.1 |
ENST00000316105.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500194380.hg.1 |
ENST00000623266.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500194380.hg.1 |
ENST00000624909.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500194380.hg.1 |
NM_019119.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500194380.hg.1 |
ENST00000316105.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500194380.hg.1 |
ENST00000623266.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500194380.hg.1 |
ENST00000624909.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500194380.hg.1 |
NM_019119.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500194380.hg.1 |
TC0500013245.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0500194383.hg.1 |
ENST00000316105.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500194383.hg.1 |
ENST00000624909.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500194383.hg.1 |
NM_019119.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500194383.hg.1 |
ENST00000316105.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500194383.hg.1 |
ENST00000624909.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500194383.hg.1 |
NM_019119.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500194383.hg.1 |
TC0500013245.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0500194386.hg.1 |
ENST00000316105.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500194386.hg.1 |
NM_019119.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500194386.hg.1 |
ENST00000316105.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500194386.hg.1 |
NM_019119.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500194386.hg.1 |
TC0500013245.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0500194379.hg.1 |
ENST00000316105.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500194379.hg.1 |
ENST00000623266.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500194379.hg.1 |
NM_019119.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500194379.hg.1 |
ENST00000316105.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500194379.hg.1 |
ENST00000623266.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500194379.hg.1 |
NM_019119.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500194379.hg.1 |
TC0500013245.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0500194381.hg.1 |
ENST00000316105.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500194381.hg.1 |
ENST00000623266.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500194381.hg.1 |
ENST00000624909.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500194381.hg.1 |
NM_019119.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500194381.hg.1 |
ENST00000316105.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500194381.hg.1 |
ENST00000623266.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500194381.hg.1 |
ENST00000624909.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500194381.hg.1 |
NM_019119.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500194381.hg.1 |
TC0500013245.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0500194382.hg.1 |
ENST00000316105.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500194382.hg.1 |
NM_019119.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500194382.hg.1 |
ENST00000316105.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500194382.hg.1 |
NM_019119.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500194382.hg.1 |
TC0500013245.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0500194384.hg.1 |
ENST00000316105.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500194384.hg.1 |
ENST00000623266.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500194384.hg.1 |
ENST00000624909.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500194384.hg.1 |
NM_019119.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500194384.hg.1 |
ENST00000316105.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500194384.hg.1 |
ENST00000623266.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500194384.hg.1 |
ENST00000624909.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500194384.hg.1 |
NM_019119.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500194384.hg.1 |
TC0500013245.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0500194385.hg.1 |
ENST00000316105.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500194385.hg.1 |
ENST00000623266.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500194385.hg.1 |
NM_019119.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500194385.hg.1 |
ENST00000316105.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500194385.hg.1 |
ENST00000623266.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500194385.hg.1 |
NM_019119.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500194385.hg.1 |
TC0500013245.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0500013245.hg.1 |
ENST00000316105.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500013245.hg.1 |
ENST00000316105.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500013245.hg.1 |
ENST00000316105.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500013245.hg.1 |
ENST00000316105.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500013245.hg.1 |
ENST00000316105.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500013245.hg.1 |
ENST00000316105.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500013245.hg.1 |
ENST00000316105.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500013245.hg.1 |
ENST00000316105.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500013245.hg.1 |
ENST00000316105.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500013245.hg.1 |
ENST00000316105.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500013245.hg.1 |
ENST00000623266.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500013245.hg.1 |
ENST00000623266.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500013245.hg.1 |
ENST00000623266.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500013245.hg.1 |
ENST00000623266.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500013245.hg.1 |
ENST00000623266.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500013245.hg.1 |
ENST00000624909.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500013245.hg.1 |
ENST00000624909.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500013245.hg.1 |
ENST00000624909.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500013245.hg.1 |
ENST00000624909.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500013245.hg.1 |
NM_019119.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500013245.hg.1 |
NM_019119.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500013245.hg.1 |
NM_019119.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500013245.hg.1 |
NM_019119.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500013245.hg.1 |
NM_019119.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500013245.hg.1 |
NM_019119.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500013245.hg.1 |
NM_019119.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500013245.hg.1 |
NM_019119.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500013245.hg.1 |
NM_019119.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500013245.hg.1 |
NM_019119.4 |
Refseq |