Assay Details
| Gene Symbol: ERCC8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK022657.1 | - | 1 | 849 | 82 | |
| AK226129.1 | - | 7 | 1622 | 82 | ||
| BC093802.1 | - | 1 | 446 | 82 | ||
| BC112227.1 | - | 1 | 446 | 82 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

ERCC8
Genomic Map