Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR17011810.hg.1 |
ENST00000269399.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR17011810.hg.1 |
ENST00000310942.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR17011810.hg.1 |
NM_005189.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR17011810.hg.1 |
NM_032647.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR17011810.hg.1 |
TC17000911.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR17011811.hg.1 |
ENST00000269399.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR17011811.hg.1 |
NM_032647.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR17011811.hg.1 |
TC17000911.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009093.hg.1 |
ENST00000269399.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009093.hg.1 |
ENST00000269399.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009093.hg.1 |
ENST00000269399.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009093.hg.1 |
ENST00000269399.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009093.hg.1 |
ENST00000269399.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009093.hg.1 |
ENST00000269399.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009093.hg.1 |
ENST00000269399.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009093.hg.1 |
ENST00000310942.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009093.hg.1 |
ENST00000310942.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009093.hg.1 |
ENST00000310942.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009093.hg.1 |
ENST00000310942.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009093.hg.1 |
ENST00000310942.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009093.hg.1 |
ENST00000310942.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009093.hg.1 |
ENST00000310942.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009093.hg.1 |
ENST00000310942.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009093.hg.1 |
ENST00000310942.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009093.hg.1 |
ENST00000310942.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009093.hg.1 |
ENST00000571484.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009093.hg.1 |
ENST00000571484.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009093.hg.1 |
ENST00000571484.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009093.hg.1 |
ENST00000571484.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009093.hg.1 |
ENST00000571484.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009093.hg.1 |
NM_005189.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009093.hg.1 |
NM_005189.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009093.hg.1 |
NM_005189.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009093.hg.1 |
NM_005189.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009093.hg.1 |
NM_005189.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009093.hg.1 |
NM_005189.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009093.hg.1 |
NM_005189.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009093.hg.1 |
NM_005189.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009093.hg.1 |
NM_032647.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009093.hg.1 |
NM_032647.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009093.hg.1 |
NM_032647.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009093.hg.1 |
NM_032647.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009093.hg.1 |
NM_032647.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009093.hg.1 |
NM_032647.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009093.hg.1 |
NM_032647.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700009093.hg.1 |
ENST00000269399.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700009093.hg.1 |
ENST00000269399.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700009093.hg.1 |
ENST00000269399.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700009093.hg.1 |
ENST00000269399.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700009093.hg.1 |
ENST00000269399.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700009093.hg.1 |
ENST00000269399.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700009093.hg.1 |
ENST00000269399.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700009093.hg.1 |
ENST00000310942.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700009093.hg.1 |
ENST00000310942.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700009093.hg.1 |
ENST00000310942.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700009093.hg.1 |
ENST00000310942.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700009093.hg.1 |
ENST00000310942.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700009093.hg.1 |
ENST00000310942.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700009093.hg.1 |
ENST00000310942.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700009093.hg.1 |
ENST00000310942.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700009093.hg.1 |
ENST00000310942.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700009093.hg.1 |
ENST00000310942.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700009093.hg.1 |
ENST00000571484.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700009093.hg.1 |
ENST00000571484.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700009093.hg.1 |
ENST00000571484.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700009093.hg.1 |
ENST00000571484.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700009093.hg.1 |
ENST00000571484.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700009093.hg.1 |
NM_005189.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700009093.hg.1 |
NM_005189.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700009093.hg.1 |
NM_005189.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700009093.hg.1 |
NM_005189.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700009093.hg.1 |
NM_005189.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700009093.hg.1 |
NM_005189.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700009093.hg.1 |
NM_005189.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700009093.hg.1 |
NM_005189.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700009093.hg.1 |
NM_032647.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700009093.hg.1 |
NM_032647.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700009093.hg.1 |
NM_032647.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700009093.hg.1 |
NM_032647.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700009093.hg.1 |
NM_032647.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700009093.hg.1 |
NM_032647.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700009093.hg.1 |
NM_032647.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700168675.hg.1 |
ENST00000269399.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700168675.hg.1 |
NM_032647.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700168675.hg.1 |
ENST00000269399.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700168675.hg.1 |
NM_032647.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700168675.hg.1 |
TC1700009093.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1700168673.hg.1 |
ENST00000269399.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700168673.hg.1 |
ENST00000310942.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700168673.hg.1 |
NM_005189.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700168673.hg.1 |
NM_032647.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700168673.hg.1 |
ENST00000269399.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700168673.hg.1 |
ENST00000310942.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700168673.hg.1 |
NM_005189.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700168673.hg.1 |
NM_032647.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700168673.hg.1 |
TC1700009093.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1700168674.hg.1 |
ENST00000269399.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700168674.hg.1 |
NM_032647.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700168674.hg.1 |
ENST00000269399.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700168674.hg.1 |
NM_032647.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700168674.hg.1 |
TC1700009093.hg.1 |
Clariom D TC |