Assay Details
| Gene Symbol: SIGLEC12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_053003.3 | NP_443729.1 | 1 - 2 | 481 | 118 | |
| XM_011527442.2 | XP_011525744.1 | 1 - 2 | 481 | 118 | ||
| GenBank mRNA | AF282256.1 | - | 1 - 2 | 461 | 118 | |
| AK095919.1 | - | 1 - 2 | 481 | 118 | ||
| AY358140.1 | - | 1 - 2 | 439 | 118 | ||
| BC035809.1 | - | 1 - 2 | 478 | 5755526 | 118 | |
| DA562082.1 | - | 1 - 2 | 481 | 118 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC19001781.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR19024202.hg.1 | ENST00000291707.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR19024202.hg.1 | NM_033329.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR19024202.hg.1 | NM_053003.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR19024202.hg.1 | TC19001781.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008626.hg.1 | ENST00000600623.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR19024204.hg.1 | ENST00000291707.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR19024204.hg.1 | NM_053003.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR19024204.hg.1 | TC19001781.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011259.hg.1 | ENST00000291707.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011259.hg.1 | ENST00000291707.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011259.hg.1 | ENST00000291707.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011259.hg.1 | ENST00000291707.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011259.hg.1 | ENST00000291707.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011259.hg.1 | ENST00000291707.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011259.hg.1 | ENST00000291707.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011259.hg.1 | ENST00000291707.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011259.hg.1 | ENST00000291707.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011259.hg.1 | ENST00000291707.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011259.hg.1 | ENST00000291707.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011259.hg.1 | ENST00000596742.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011259.hg.1 | ENST00000596742.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011259.hg.1 | ENST00000596742.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011259.hg.1 | ENST00000596742.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011259.hg.1 | ENST00000596742.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011259.hg.1 | ENST00000596742.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011259.hg.1 | ENST00000596742.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011259.hg.1 | ENST00000596742.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011259.hg.1 | ENST00000596742.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011259.hg.1 | ENST00000596742.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011259.hg.1 | ENST00000598614.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011259.hg.1 | ENST00000598614.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011259.hg.1 | ENST00000598614.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011259.hg.1 | ENST00000598614.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011259.hg.1 | ENST00000598614.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011259.hg.1 | ENST00000598614.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011259.hg.1 | ENST00000598614.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011259.hg.1 | ENST00000598614.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011259.hg.1 | ENST00000598614.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011259.hg.1 | NM_033329.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011259.hg.1 | NM_033329.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011259.hg.1 | NM_033329.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011259.hg.1 | NM_033329.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011259.hg.1 | NM_033329.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011259.hg.1 | NM_033329.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011259.hg.1 | NM_033329.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011259.hg.1 | NM_033329.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011259.hg.1 | NM_033329.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011259.hg.1 | NM_033329.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011259.hg.1 | NM_033329.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011259.hg.1 | NM_053003.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011259.hg.1 | NM_053003.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011259.hg.1 | NM_053003.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011259.hg.1 | NM_053003.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011259.hg.1 | NM_053003.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011259.hg.1 | NM_053003.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011259.hg.1 | NM_053003.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011259.hg.1 | NM_053003.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011259.hg.1 | NM_053003.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011259.hg.1 | NM_053003.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011259.hg.1 | NM_053003.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011259.hg.1 | NM_053003.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900008626.hg.1 | ENST00000600623.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011259.hg.1 | ENST00000291707.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011259.hg.1 | ENST00000291707.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011259.hg.1 | ENST00000291707.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011259.hg.1 | ENST00000291707.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011259.hg.1 | ENST00000291707.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011259.hg.1 | ENST00000291707.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011259.hg.1 | ENST00000291707.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011259.hg.1 | ENST00000291707.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011259.hg.1 | ENST00000291707.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011259.hg.1 | ENST00000291707.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011259.hg.1 | ENST00000291707.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011259.hg.1 | ENST00000596742.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011259.hg.1 | ENST00000596742.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011259.hg.1 | ENST00000596742.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011259.hg.1 | ENST00000596742.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011259.hg.1 | ENST00000596742.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011259.hg.1 | ENST00000596742.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011259.hg.1 | ENST00000596742.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011259.hg.1 | ENST00000596742.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011259.hg.1 | ENST00000596742.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011259.hg.1 | ENST00000596742.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011259.hg.1 | ENST00000598614.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011259.hg.1 | ENST00000598614.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011259.hg.1 | ENST00000598614.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011259.hg.1 | ENST00000598614.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011259.hg.1 | ENST00000598614.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011259.hg.1 | ENST00000598614.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011259.hg.1 | ENST00000598614.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011259.hg.1 | ENST00000598614.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011259.hg.1 | ENST00000598614.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011259.hg.1 | NM_033329.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011259.hg.1 | NM_033329.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011259.hg.1 | NM_033329.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011259.hg.1 | NM_033329.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011259.hg.1 | NM_033329.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011259.hg.1 | NM_033329.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011259.hg.1 | NM_033329.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011259.hg.1 | NM_033329.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011259.hg.1 | NM_033329.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011259.hg.1 | NM_033329.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011259.hg.1 | NM_033329.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011259.hg.1 | NM_053003.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011259.hg.1 | NM_053003.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011259.hg.1 | NM_053003.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011259.hg.1 | NM_053003.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011259.hg.1 | NM_053003.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011259.hg.1 | NM_053003.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011259.hg.1 | NM_053003.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011259.hg.1 | NM_053003.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011259.hg.1 | NM_053003.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011259.hg.1 | NM_053003.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011259.hg.1 | NM_053003.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011259.hg.1 | NM_053003.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900194798.hg.1 | ENST00000291707.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900194798.hg.1 | ENST00000598614.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900194798.hg.1 | NM_033329.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900194798.hg.1 | NM_053003.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900194798.hg.1 | TC1900011259.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1900194798.hg.1 | ENST00000291707.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900194798.hg.1 | ENST00000598614.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900194798.hg.1 | NM_033329.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900194798.hg.1 | NM_053003.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900194801.hg.1 | ENST00000291707.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900194801.hg.1 | ENST00000596742.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900194801.hg.1 | NM_053003.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900194801.hg.1 | TC1900011259.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1900194801.hg.1 | ENST00000291707.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900194801.hg.1 | ENST00000596742.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900194801.hg.1 | NM_053003.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900194802.hg.1 | ENST00000291707.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900194802.hg.1 | ENST00000596742.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900194802.hg.1 | NM_053003.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900194802.hg.1 | TC1900011259.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1900194802.hg.1 | ENST00000291707.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900194802.hg.1 | ENST00000596742.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900194802.hg.1 | NM_053003.3 | Refseq |

SIGLEC12
Genomic Map