Assay Details
| Gene Symbol: TSSK2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_053006.4 | NP_443732.3 | 1 | 1470 | 130 | |
| GenBank mRNA | AF362953.1 | - | 1 | 950 | 130 | |
| BC037781.2 | - | 1 | 1001 | 5273397 | 130 | |
| CR456587.1 | - | 1 | 892 | 130 | ||
| L77564.1 | - | 1 | 1470 | 130 | ||
| Gene Symbol: DGCR14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_022719.2 | NP_073210.1 | 10 | 130 | ||
| NR_134304.1 | 10 | 130 | ||||
| GenBank mRNA | AK095932.1 | - | 1 | 130 | ||
| BC037829.1 | - | 1 | 130 | |||
| BC069275.1 | - | 1 | 130 | |||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC22000041.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
protein phosphorylation
multicellular organism development
spermatid development
intracellular signal transduction
protein autophosphorylation
mRNA splicing, via spliceosome
nervous system development
magnesium ion binding
protein serine/threonine kinase activity
protein binding
ATP binding
molecular_function
protein phosphorylation
multicellular organism development
spermatid development
intracellular signal transduction
protein autophosphorylation
mRNA splicing, via spliceosome
nervous system development
magnesium ion binding
protein serine/threonine kinase activity
protein binding
ATP binding
molecular_function
MicroArray Probes:
Agilent
Affymetrix
ArrayStar
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P32098297 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P11428113 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P11428112 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P11428114 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P32098285 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P11428116 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P32098287 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P11428118 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P32098296 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P11428110 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P11428117 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P11428119 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P11428111 | - | - |
| Agilent Whole Human Genome Microarray 4x44K v2 | A_23_P40515 | - | - |
| Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k | GT_44k_23_P40515 | - | - |
| Agilent Whole Human Genome Oligo Microarray 4x44K | 36948 | - | - |
| Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray | 4049 | - | - |
| Agilent Whole Human Genome Microarray 4x44K v2 | A_23_P40515 | - | - |
| SurePrint G3 Human GE Microarray G4851A | A_23_P40515 | - | - |
| SurePrint G3 Human GE Microarray G4851A | A_33_P3225011 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11428110 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11428113 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11428118 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11428119 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11428117 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32098297 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32098296 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11428116 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11428114 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11428111 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32098285 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32098285 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32098296 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11428116 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11428113 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11428117 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11428118 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11428111 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32098297 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11428119 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11428114 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11428110 | - | - |
| Whole Human Genome Oligo Microarray G4112A | A_23_P40515 | - | - |
| Agilent Human Promoter ChIP-on-Chip A) | A_17_P11428110 | - | - |
| Agilent Human Promoter ChIP-on-Chip A) | A_17_P11428120 | - | - |
| Agilent Human Promoter ChIP-on-Chip A) | A_17_P11428116 | - | - |
| Agilent Human Promoter ChIP-on-Chip A) | A_17_P11428118 | - | - |
| Agilent Human Promoter ChIP-on-Chip A) | A_17_P11428111 | - | - |
| Agilent Human Promoter ChIP-on-Chip A) | A_17_P11428117 | - | - |
| Agilent Human Promoter ChIP-on-Chip A) | A_17_P11428113 | - | - |
| Agilent Human Promoter ChIP-on-Chip A) | A_17_P11428123 | - | - |
| Agilent Human Promoter ChIP-on-Chip A) | A_17_P11428114 | - | - |
| Agilent Human Promoter ChIP-on-Chip A) | A_17_P11428121 | - | - |
| Agilent Human CpG Island ChIP-on-Chip Microarray 244K (G4492A) | A_17_P11428116 | - | - |
| Agilent Human CpG Island ChIP-on-Chip Microarray 244K (G4492A) | A_17_P11428118 | - | - |
| Agilent Human CpG Island ChIP-on-Chip Microarray 244K (G4492A) | A_17_P11428119 | - | - |
| Agilent Human CpG Island ChIP-on-Chip Microarray 244K (G4492A) | A_17_P11428117 | - | - |
| Agilent Whole Human Genome Microarray 4x44K G4112F | A_23_P40515 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P11428061 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P11428071 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P11428190 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P11428184 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P11428113 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P11428180 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P17254641 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P11428174 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P11428062 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P11428187 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P11428072 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P11428067 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P11428200 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P11428123 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P11428179 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P11428120 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P11428057 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P11428059 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P17254640 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P17254638 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P11428114 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P11428110 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P11428192 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P11428118 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P11428202 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P11428075 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P11428197 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P17254654 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P11428177 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P11428070 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P11428121 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P11428063 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P17254650 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P11428068 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P11428181 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P11428116 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Gene 1.0 ST Array | NM_053006_at | - | - |
| Affymetrix Human Gene 1.0 ST Array | 8071197 | - | - |
| Affymetrix Human Gene 1.0 ST Array | 8074368 | - | - |
| Affymetrix Human Gene 1.0 ST Array | 8074367 | - | - |
| Affymetrix Human Gene 1.0 ST Array | 8071198 | - | - |
| Affymetrix Human Gene 1.0 ST Array | 8071199 | - | - |
| Affymetrix Human Gene 1.0 ST Array | 8074366 | - | - |
| Affymetrix Human Gene 1.0 ST Array | 8074365 | - | - |
| Affymetrix Human Gene 1.0 ST Array | 8071200 | - | - |
| Affymetrix Human Gene 1.1 ST Array | 8074364 | - | - |
| Affymetrix Human Gene 1.1 ST Array | 8071196 | - | - |
| Affymetrix HT HG-U133+ PM Array Plate | 214371_PM_at | - | - |
| Affymetrix HT HG-U133+ PM Array Plate | 217275_PM_at | - | - |
| Affymetrix Human Genome U219 Array | 11727218_PM_at | - | - |
| Affymetrix Human Genome U219 Array | 11727218_at | - | - |
| Affymetrix Human Transcriptome Array 2.0 | PSR22000349.hg.1 | ENST00000399635.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR22000349.hg.1 | NM_053006.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR22000349.hg.1 | TC22000041.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR22000348.hg.1 | ENST00000399635.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR22000348.hg.1 | NM_053006.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR22000348.hg.1 | TC22000041.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006587.hg.1 | ENST00000399635.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006587.hg.1 | ENST00000399635.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006587.hg.1 | ENST00000399635.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006587.hg.1 | ENST00000399635.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006587.hg.1 | NM_053006.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006587.hg.1 | NM_053006.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006587.hg.1 | NM_053006.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006587.hg.1 | NM_053006.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200006587.hg.1 | ENST00000399635.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200006587.hg.1 | ENST00000399635.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200006587.hg.1 | ENST00000399635.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200006587.hg.1 | ENST00000399635.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200006587.hg.1 | NM_053006.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200006587.hg.1 | NM_053006.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200006587.hg.1 | NM_053006.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200006587.hg.1 | NM_053006.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2200145111.hg.1 | TC2200006587.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR2200145111.hg.1 | ENST00000399635.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200145111.hg.1 | NM_053006.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2200145111.hg.1 | ENST00000399635.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200145111.hg.1 | NM_053006.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2200145109.hg.1 | TC2200006587.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR2200145109.hg.1 | ENST00000399635.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200145109.hg.1 | NM_053006.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2200145109.hg.1 | ENST00000399635.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200145109.hg.1 | NM_053006.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2200145110.hg.1 | TC2200006587.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR2200145110.hg.1 | ENST00000399635.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200145110.hg.1 | NM_053006.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2200145110.hg.1 | ENST00000399635.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200145110.hg.1 | NM_053006.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2200145112.hg.1 | TC2200006587.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR2200145112.hg.1 | ENST00000399635.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200145112.hg.1 | NM_053006.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2200145112.hg.1 | ENST00000399635.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200145112.hg.1 | NM_053006.4 | Refseq |
| Human Genome U133 Plus 2.0 Array | 214371_PM_at | - | - |
| Human Genome U133 Plus 2.0 Array | 214371_at | - | - |
| Affymetrix GeneChip Human Genome U133 Plus 2.0 Array based on a custom CDF (REFSEQ_7) | NM_053006_at | - | - |
| AffymetrixPathwork Diagnostics Pathchip | pw524055 | - | - |
| Human Genome U95 Set | 39534_PM_at | - | - |
| Human Genome U95 Set | 39534_at | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 214371_at2 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 214371_at1 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 214371_at7 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 214371_at9 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 214371_at3 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 214371_at10 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 214371_at8 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 214371_at5 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 214371_at4 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 214371_at11 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 214371_at6 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Arraystar Human LncRNA microarray V2.0 | ASHG19A3A020078 | - | - |
| Arraystar Human LncRNA microarray V2.0 | ASHG19A3A020078 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Human Genome Survey Microarray Version 2.0 | 175663 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| HumanHT-12 v4 Expression BeadChip | ILMN_1738843 | BC037781.2 | human_transcriptome |
| HumanHT-12 v4 Expression BeadChip | ILMN_1738843 | NM_053006.4 | human_transcriptome |
| Illumina HumanWG-6 v2.0 expression beadchip | ILMN_1738843 | BC037781.2 | human_transcriptome |
| Illumina HumanWG-6 v2.0 expression beadchip | ILMN_1738843 | NM_053006.4 | human_transcriptome |
| HumanHT-12 V4.0 expression beadchip | ILMN_1738843 | BC037781.2 | human_transcriptome |
| HumanHT-12 V4.0 expression beadchip | ILMN_1738843 | NM_053006.4 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_1738843 | BC037781.2 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_1738843 | NM_053006.4 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip | ILMN_1738843 | BC037781.2 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip | ILMN_1738843 | NM_053006.4 | human_transcriptome |
| Sentrix Human-6 Expression BeadChip | GI_34222349-S | L77564.1 | human_transcriptome |
| Sentrix Human-6 Expression BeadChip | GI_34222349-S | BC037781.2 | human_transcriptome |
| Sentrix Human-6 Expression BeadChip | GI_34222349-S | AF362953.1 | human_transcriptome |
| Sentrix Human-6 Expression BeadChip | GI_34222349-S | NM_053006.4 | human_transcriptome |
| Sentrix Human-6 Expression BeadChip | GI_34222349-S | BC069275.1 | human_transcriptome |
| Illumina HumanRef-8 v2 Expression BeadChip | GI_11196-S | - | - |
| Illumina human-6 v1.0 expression beadchip | 0001340377 | - | - |
| Illumina humanRef-8 v1.0 expression beadchip | 0001340377 | - | - |
| Illumina human-6 v2.0 expression beadchip | ILMN_1738843 | BC037781.2 | human_transcriptome |
| Illumina human-6 v2.0 expression beadchip | ILMN_1738843 | NM_053006.4 | human_transcriptome |
| Illumina humanRef-8 v2.0 expression beadchip | ILMN_1738843 | BC037781.2 | human_transcriptome |
| Illumina humanRef-8 v2.0 expression beadchip | ILMN_1738843 | NM_053006.4 | human_transcriptome |
| Illumina Sentrix Human-6 v2 Expression BeadChip | 580382 | - | - |
| Illumina HumanRef-6 v2.0 expression beadchip | ILMN_11196 | - | - |
| Illumina HumanRef-8 v3.0 expression beadchip | ILMN_1738843 | BC037781.2 | human_transcriptome |
| Illumina HumanRef-8 v3.0 expression beadchip | ILMN_1738843 | NM_053006.4 | human_transcriptome |
| Illumina HumanWG-6 v3.0 expression beadchip | ILMN_1738843 | BC037781.2 | human_transcriptome |
| Illumina HumanWG-6 v3.0 expression beadchip | ILMN_1738843 | NM_053006.4 | human_transcriptome |
| Illumina HumanHT-12 V3.0 expression beadchip | ILMN_1738843 | BC037781.2 | human_transcriptome |
| Illumina HumanHT-12 V3.0 expression beadchip | ILMN_1738843 | NM_053006.4 | human_transcriptome |
| Illumina HumanWG-6_V2_0_R2 | ILMN_1738843 | BC037781.2 | human_transcriptome |
| Illumina HumanWG-6_V2_0_R2 | ILMN_1738843 | NM_053006.4 | human_transcriptome |
| Illumina HumanRef-8 WG-DASL v3.0 | ILMN_1738843 | BC037781.2 | human_transcriptome |
| Illumina HumanRef-8 WG-DASL v3.0 | ILMN_1738843 | NM_053006.4 | human_transcriptome |

TSSK2
Genomic Map